Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 77624949-77625494 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLNS1A
Entrez Gene ID
1207
Gene Name
chloride nucleotide-sensitive channel 1A
Gene Aliases
CLCI, CLNS1B, ICln
UniGene
Hs.430733
Gene Chromosome Location
Chr.11: 77616151-77637806 on Build GRCh38
Transcripts
NM_001293.2 NM_001311199.1 NM_001311200.1 NM_001311201.1 NM_001311202.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7139697 c.424G>A p.E142K Chr.11: 77625011-77625011 on Build GRCh38
1357116 c.451G>T p.E151* Chr.11: 77624984-77624984 on Build GRCh38
6630586 c.390C>T p.C130C Chr.11: 77625045-77625045 on Build GRCh38
6484699 c.418G>A p.E140K Chr.11: 77625017-77625017 on Build GRCh38
932189 c.391G>A p.E131K Chr.11: 77625044-77625044 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs144382400 Chr.11: 77624993-77624993 on Build GRCh38
rs751869860 Chr.11: 77625103-77625103 on Build GRCh38
rs748760788 Chr.11: 77625045-77625045 on Build GRCh38
rs946876584 Chr.11: 77625256-77625256 on Build GRCh38
rs147214856 Chr.11: 77624972-77624972 on Build GRCh38
rs760007966 Chr.11: 77624984-77624984 on Build GRCh38
rs995089220 Chr.11: 77625147-77625147 on Build GRCh38
rs1042239523 Chr.11: 77625228-77625228 on Build GRCh38
rs772979604 Chr.11: 77625081-77625082 on Build GRCh38
rs769617390 Chr.11: 77625316-77625316 on Build GRCh38
rs572041764 Chr.11: 77625279-77625279 on Build GRCh38
rs762618847 Chr.11: 77625374-77625374 on Build GRCh38
rs747798115 Chr.11: 77625054-77625054 on Build GRCh38
rs778688458 Chr.11: 77625051-77625051 on Build GRCh38
rs750434003 Chr.11: 77625030-77625030 on Build GRCh38
rs569426404 Chr.11: 77625249-77625249 on Build GRCh38
rs779459403 Chr.11: 77625044-77625044 on Build GRCh38
rs1015701276 Chr.11: 77625451-77625451 on Build GRCh38
rs763033179 Chr.11: 77625077-77625077 on Build GRCh38
rs771328257 Chr.11: 77625062-77625062 on Build GRCh38
rs940268610 Chr.11: 77624997-77624997 on Build GRCh38
rs768203603 Chr.11: 77625047-77625047 on Build GRCh38
rs868770474 Chr.11: 77625060-77625060 on Build GRCh38
rs554921700 Chr.11: 77625278-77625278 on Build GRCh38
rs1055546897 Chr.11: 77625186-77625186 on Build GRCh38
rs535651129 Chr.11: 77625207-77625207 on Build GRCh38
rs139711418 Chr.11: 77625121-77625121 on Build GRCh38
rs141952158 Chr.11: 77625018-77625018 on Build GRCh38
rs868318879 Chr.11: 77625295-77625295 on Build GRCh38
rs140625962 Chr.11: 77624973-77624973 on Build GRCh38
rs764076653 Chr.11: 77625086-77625086 on Build GRCh38
rs540072253 Chr.11: 77625033-77625033 on Build GRCh38
rs758580440 Chr.11: 77625001-77625001 on Build GRCh38
rs368773419 Chr.11: 77625002-77625002 on Build GRCh38
rs1051036502 Chr.11: 77625293-77625293 on Build GRCh38
rs990624319 Chr.11: 77625214-77625214 on Build GRCh38
rs746506869 Chr.11: 77625056-77625056 on Build GRCh38
rs751817097 Chr.11: 77625006-77625006 on Build GRCh38
rs898961686 Chr.11: 77625144-77625144 on Build GRCh38
rs994650890 Chr.11: 77625457-77625457 on Build GRCh38
rs914982301 Chr.11: 77625215-77625215 on Build GRCh38
rs760059138 Chr.11: 77625066-77625066 on Build GRCh38
rs773449647 Chr.11: 77624971-77624971 on Build GRCh38
rs951825455 Chr.11: 77625384-77625384 on Build GRCh38
rs182584204 Chr.11: 77625331-77625331 on Build GRCh38
rs781101003 Chr.11: 77625029-77625029 on Build GRCh38
rs371312037 Chr.11: 77625071-77625071 on Build GRCh38
rs577715904 Chr.11: 77625453-77625453 on Build GRCh38
rs781581866 Chr.11: 77625089-77625089 on Build GRCh38
rs889830707 Chr.11: 77625300-77625300 on Build GRCh38
rs978248188 Chr.11: 77625259-77625259 on Build GRCh38
rs983197726 Chr.11: 77625432-77625432 on Build GRCh38
rs1007010953 Chr.11: 77625109-77625109 on Build GRCh38
rs747493269 Chr.11: 77625014-77625014 on Build GRCh38
rs374532472 Chr.11: 77625000-77625000 on Build GRCh38
rs534601941 Chr.11: 77625326-77625326 on Build GRCh38
rs772966252 Chr.11: 77625360-77625360 on Build GRCh38
rs995824139 Chr.11: 77625355-77625355 on Build GRCh38
rs777192891 Chr.11: 77624980-77624980 on Build GRCh38
rs779464286 Chr.11: 77624982-77624984 on Build GRCh38
rs535053321 Chr.11: 77625260-77625260 on Build GRCh38
rs770551188 Chr.11: 77625435-77625435 on Build GRCh38
rs747779974 Chr.11: 77625157-77625157 on Build GRCh38
rs549420048 Chr.11: 77625206-77625206 on Build GRCh38
rs1008801077 Chr.11: 77625291-77625291 on Build GRCh38
rs775437809 Chr.11: 77625076-77625076 on Build GRCh38
rs942696292 Chr.11: 77625329-77625329 on Build GRCh38
rs762102529 Chr.11: 77625122-77625122 on Build GRCh38
rs768165632 Chr.11: 77625070-77625071 on Build GRCh38
rs201097455 Chr.11: 77625021-77625021 on Build GRCh38

View Full Product Details