Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 78120940-78121449 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
UniGene
Hs.503368
Gene Chromosome Location
Chr.11: 78100942-78139660 on Build GRCh38
Transcripts
NM_001007027.2 NM_024079.4 XM_005274247.3 XM_011545252.2 XM_017018274.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4444373 c.460G>A p.G154R Chr.11: 78121083-78121083 on Build GRCh38
6070289 c.444A>G p.L148L Chr.11: 78121099-78121099 on Build GRCh38
5970393 c.382A>G p.I128V Chr.11: 78121161-78121161 on Build GRCh38
5967864 c.420G>C p.K140N Chr.11: 78121123-78121123 on Build GRCh38
6070288 c.444A>G p.L148L Chr.11: 78121099-78121099 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774531870 Chr.11: 78121232-78121232 on Build GRCh38
rs139832787 Chr.11: 78121064-78121064 on Build GRCh38
rs781047936 Chr.11: 78121227-78121227 on Build GRCh38
rs370944777 Chr.11: 78121088-78121088 on Build GRCh38
rs534739528 Chr.11: 78121105-78121105 on Build GRCh38
rs773750460 Chr.11: 78121244-78121244 on Build GRCh38
rs940120502 Chr.11: 78121287-78121288 on Build GRCh38
rs762415620 Chr.11: 78121233-78121233 on Build GRCh38
rs944305211 Chr.11: 78120981-78120981 on Build GRCh38
rs991765683 Chr.11: 78121043-78121043 on Build GRCh38
rs518624 Chr.11: 78121030-78121030 on Build GRCh38
rs1013259366 Chr.11: 78121271-78121271 on Build GRCh38
rs201359142 Chr.11: 78121083-78121083 on Build GRCh38
rs113030322 Chr.11: 78121108-78121108 on Build GRCh38
rs767659108 Chr.11: 78121174-78121174 on Build GRCh38
rs151266773 Chr.11: 78121117-78121117 on Build GRCh38
rs521298 Chr.11: 78121307-78121307 on Build GRCh38
rs760174988 Chr.11: 78121056-78121056 on Build GRCh38
rs754838558 Chr.11: 78121194-78121194 on Build GRCh38
rs957666759 Chr.11: 78121424-78121424 on Build GRCh38
rs905153886 Chr.11: 78121368-78121368 on Build GRCh38
rs777371572 Chr.11: 78121216-78121216 on Build GRCh38
rs199913102 Chr.11: 78121308-78121308 on Build GRCh38
rs141978141 Chr.11: 78121078-78121078 on Build GRCh38
rs606459 Chr.11: 78121326-78121326 on Build GRCh38
rs111831728 Chr.11: 78121161-78121161 on Build GRCh38
rs182551019 Chr.11: 78121365-78121365 on Build GRCh38
rs387906277 Chr.11: 78121130-78121130 on Build GRCh38
rs368103442 Chr.11: 78120990-78120990 on Build GRCh38
rs778870497 Chr.11: 78121240-78121250 on Build GRCh38
rs772535935 Chr.11: 78121106-78121106 on Build GRCh38
rs577713411 Chr.11: 78121231-78121231 on Build GRCh38
rs771172816 Chr.11: 78121021-78121021 on Build GRCh38
rs774926172 Chr.11: 78121164-78121164 on Build GRCh38
rs748126939 Chr.11: 78121203-78121203 on Build GRCh38
rs908672162 Chr.11: 78121281-78121281 on Build GRCh38
rs753631154 Chr.11: 78121146-78121147 on Build GRCh38
rs772979769 Chr.11: 78121048-78121048 on Build GRCh38
rs753578554 Chr.11: 78121353-78121353 on Build GRCh38
rs377603174 Chr.11: 78121062-78121062 on Build GRCh38
rs373231474 Chr.11: 78121041-78121041 on Build GRCh38
rs749124205 Chr.11: 78121100-78121100 on Build GRCh38
rs746539782 Chr.11: 78121222-78121222 on Build GRCh38
rs772497365 Chr.11: 78121235-78121235 on Build GRCh38
rs760798823 Chr.11: 78121250-78121250 on Build GRCh38
rs374645837 Chr.11: 78121189-78121189 on Build GRCh38
rs1019711324 Chr.11: 78121162-78121162 on Build GRCh38
rs768543831 Chr.11: 78121028-78121028 on Build GRCh38
rs776390397 Chr.11: 78121160-78121160 on Build GRCh38
rs143349528 Chr.11: 78121113-78121113 on Build GRCh38
rs377707742 Chr.11: 78121093-78121093 on Build GRCh38
rs1000704604 Chr.11: 78121382-78121382 on Build GRCh38
rs772100224 Chr.11: 78121259-78121262 on Build GRCh38
rs756243372 Chr.11: 78121097-78121097 on Build GRCh38
rs769636151 Chr.11: 78121018-78121018 on Build GRCh38
rs1057191137 Chr.11: 78121288-78121291 on Build GRCh38
rs375077103 Chr.11: 78120968-78120968 on Build GRCh38
rs606461 Chr.11: 78121328-78121328 on Build GRCh38
rs749447530 Chr.11: 78121010-78121010 on Build GRCh38
rs764314364 Chr.11: 78121230-78121230 on Build GRCh38
rs745511204 Chr.11: 78121228-78121228 on Build GRCh38
rs1038257482 Chr.11: 78121211-78121211 on Build GRCh38
rs200626083 Chr.11: 78121304-78121304 on Build GRCh38
rs201483866 Chr.11: 78121180-78121180 on Build GRCh38
rs769196188 Chr.11: 78121344-78121344 on Build GRCh38
rs764540263 Chr.11: 78121163-78121163 on Build GRCh38
rs979106666 Chr.11: 78121405-78121405 on Build GRCh38
rs758216857 Chr.11: 78121206-78121206 on Build GRCh38
rs766439033 Chr.11: 78121255-78121255 on Build GRCh38
rs562139 Chr.11: 78121306-78121306 on Build GRCh38
rs757122764 Chr.11: 78121204-78121206 on Build GRCh38
rs606425 Chr.11: 78121303-78121303 on Build GRCh38
rs1006790690 Chr.11: 78120987-78120987 on Build GRCh38
rs542904458 Chr.11: 78121293-78121293 on Build GRCh38
rs144501219 Chr.11: 78121084-78121084 on Build GRCh38
rs745818926 Chr.11: 78121244-78121244 on Build GRCh38
rs376124604 Chr.11: 78121288-78121288 on Build GRCh38
rs376537848 Chr.11: 78121022-78121022 on Build GRCh38
rs763583384 Chr.11: 78121065-78121065 on Build GRCh38
rs762169500 Chr.11: 78121167-78121167 on Build GRCh38
rs754049229 Chr.11: 78121192-78121192 on Build GRCh38
rs370958120 Chr.11: 78121046-78121046 on Build GRCh38
rs573862897 Chr.11: 78121404-78121404 on Build GRCh38
rs969038892 Chr.11: 78121081-78121081 on Build GRCh38
rs778607862 Chr.11: 78121201-78121201 on Build GRCh38
rs770665646 Chr.11: 78121146-78121146 on Build GRCh38
rs112109685 Chr.11: 78121102-78121102 on Build GRCh38
rs989262748 Chr.11: 78121287-78121287 on Build GRCh38
rs770541267 Chr.11: 78121223-78121223 on Build GRCh38
rs748027702 Chr.11: 78121104-78121104 on Build GRCh38
rs575902068 Chr.11: 78121257-78121257 on Build GRCh38
rs966510518 Chr.11: 78121319-78121319 on Build GRCh38
rs1020718978 Chr.11: 78121313-78121313 on Build GRCh38
rs7105633 Chr.11: 78121423-78121423 on Build GRCh38
rs368832117 Chr.11: 78121011-78121011 on Build GRCh38

View Full Product Details