Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 82901611-82902112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDIAS
Entrez Gene ID
220042
Gene Name
DNA damage induced apoptosis suppressor
Gene Aliases
C11orf82, noxin
UniGene
Hs.165607
Gene Chromosome Location
Chr.11: 82901695-82934659 on Build GRCh38
Transcripts
NM_145018.3 XM_011544835.1 XM_011544838.2 XM_017017352.1 XM_017017353.1 NM_001319214.1 XM_005274093.1

Gene Symbol
PRCP
Entrez Gene ID
5547
Gene Name
prolylcarboxypeptidase
Gene Aliases
HUMPCP, PCP
UniGene
Hs.523936
Gene Chromosome Location
Chr.11: 82822940-82901740 on Build GRCh38
Transcripts
NM_145018.3 XM_011544835.1 XM_011544838.2 XM_017017352.1 XM_017017353.1 NM_001319214.1 XM_005274093.1

SNP Detail

SNP ID Chromosome Location
rs1015903968 Chr.11: 82901724-82901724 on Build GRCh38
rs1026773126 Chr.11: 82901750-82901750 on Build GRCh38
rs944290296 Chr.11: 82901668-82901668 on Build GRCh38
rs574782651 Chr.11: 82901772-82901772 on Build GRCh38
rs569604690 Chr.11: 82901914-82901914 on Build GRCh38
rs929048139 Chr.11: 82901692-82901692 on Build GRCh38
rs918872310 Chr.11: 82902014-82902014 on Build GRCh38
rs148532882 Chr.11: 82901858-82901858 on Build GRCh38
rs114550967 Chr.11: 82901798-82901798 on Build GRCh38
rs932115192 Chr.11: 82902047-82902047 on Build GRCh38
rs181756671 Chr.11: 82901777-82901777 on Build GRCh38
rs890481365 Chr.11: 82901807-82901807 on Build GRCh38
rs890419657 Chr.11: 82901681-82901681 on Build GRCh38
rs1054974840 Chr.11: 82901843-82901843 on Build GRCh38
rs1051821329 Chr.11: 82901801-82901801 on Build GRCh38
rs990494881 Chr.11: 82901762-82901762 on Build GRCh38
rs3763813 Chr.11: 82901979-82901979 on Build GRCh38
rs773047262 Chr.11: 82901878-82901878 on Build GRCh38
rs949047422 Chr.11: 82901771-82901771 on Build GRCh38
rs973406210 Chr.11: 82901998-82901998 on Build GRCh38
rs61902275 Chr.11: 82901845-82901845 on Build GRCh38
rs78295364 Chr.11: 82901846-82901846 on Build GRCh38
rs996262285 Chr.11: 82901660-82901660 on Build GRCh38
rs959117615 Chr.11: 82901758-82901758 on Build GRCh38
rs923631108 Chr.11: 82902027-82902027 on Build GRCh38
rs534755281 Chr.11: 82902024-82902024 on Build GRCh38
rs766623930 Chr.11: 82901934-82901934 on Build GRCh38
rs759490019 Chr.11: 82902075-82902075 on Build GRCh38
rs887621923 Chr.11: 82901729-82901729 on Build GRCh38
rs1049190855 Chr.11: 82901813-82901813 on Build GRCh38
rs954109144 Chr.11: 82901778-82901778 on Build GRCh38
rs1002065486 Chr.11: 82901737-82901737 on Build GRCh38
rs563684831 Chr.11: 82901811-82901811 on Build GRCh38
rs922110828 Chr.11: 82902040-82902040 on Build GRCh38
rs7943593 Chr.11: 82901653-82901653 on Build GRCh38
rs776652880 Chr.11: 82901799-82901799 on Build GRCh38
rs142821764 Chr.11: 82901982-82901982 on Build GRCh38
rs954161162 Chr.11: 82901885-82901885 on Build GRCh38
rs532529335 Chr.11: 82901820-82901820 on Build GRCh38
rs978193517 Chr.11: 82902006-82902006 on Build GRCh38
rs974341990 Chr.11: 82901730-82901730 on Build GRCh38
rs779690095 Chr.11: 82901663-82901663 on Build GRCh38
rs760862002 Chr.11: 82901915-82901915 on Build GRCh38
rs984491953 Chr.11: 82902055-82902055 on Build GRCh38
rs966712412 Chr.11: 82901775-82901775 on Build GRCh38
rs543870992 Chr.11: 82901809-82901809 on Build GRCh38
rs918934908 Chr.11: 82901802-82901802 on Build GRCh38
rs1013472407 Chr.11: 82901763-82901763 on Build GRCh38
rs547590016 Chr.11: 82901827-82901827 on Build GRCh38
rs3763814 Chr.11: 82901800-82901800 on Build GRCh38
rs1032301637 Chr.11: 82901881-82901885 on Build GRCh38
rs1008160128 Chr.11: 82901702-82901702 on Build GRCh38
rs961300819 Chr.11: 82901726-82901726 on Build GRCh38
rs977765901 Chr.11: 82901774-82901774 on Build GRCh38
rs917595446 Chr.11: 82901764-82901764 on Build GRCh38
rs897455599 Chr.11: 82901686-82901687 on Build GRCh38
rs559638028 Chr.11: 82901704-82901704 on Build GRCh38
rs1053943027 Chr.11: 82901635-82901635 on Build GRCh38
rs902417056 Chr.11: 82901853-82901853 on Build GRCh38
rs536660324 Chr.11: 82901977-82901977 on Build GRCh38
rs958537313 Chr.11: 82902028-82902028 on Build GRCh38
rs773560929 Chr.11: 82901854-82901854 on Build GRCh38
rs1033511548 Chr.11: 82901748-82901748 on Build GRCh38
rs937738028 Chr.11: 82901839-82901839 on Build GRCh38
rs909002329 Chr.11: 82901818-82901818 on Build GRCh38
rs924214911 Chr.11: 82901776-82901776 on Build GRCh38
rs989699311 Chr.11: 82902031-82902031 on Build GRCh38
rs959169560 Chr.11: 82901867-82901867 on Build GRCh38

View Full Product Details