Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83211301-83211849 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD42
Entrez Gene ID
338699
Gene Name
ankyrin repeat domain 42
Gene Aliases
PPP1R79, SARP
UniGene
Hs.503438
Gene Chromosome Location
Chr.11: 83193739-83256099 on Build GRCh38
Transcripts
NM_001300972.1 NM_001300973.1 NM_001300975.1 NM_001300976.1 NM_001300977.1 NM_182603.3 NR_125355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5373720 c.575G>A p.G192E Chr.11: 83211419-83211419 on Build GRCh38
7412226 c.500C>T p.P167L Chr.11: 83211428-83211428 on Build GRCh38
6324776 c.502G>T p.V168F Chr.11: 83211430-83211430 on Build GRCh38
5980175 c.554T>C p.I185T Chr.11: 83211398-83211398 on Build GRCh38
6324777 c.586G>T p.E196* Chr.11: 83211430-83211430 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768602964 Chr.11: 83211378-83211378 on Build GRCh38
rs751907301 Chr.11: 83211479-83211479 on Build GRCh38
rs774258551 Chr.11: 83211398-83211398 on Build GRCh38
rs938942062 Chr.11: 83211743-83211743 on Build GRCh38
rs796895171 Chr.11: 83211502-83211502 on Build GRCh38
rs976465609 Chr.11: 83211577-83211577 on Build GRCh38
rs371567372 Chr.11: 83211427-83211427 on Build GRCh38
rs781389549 Chr.11: 83211350-83211350 on Build GRCh38
rs765094239 Chr.11: 83211440-83211440 on Build GRCh38
rs929424767 Chr.11: 83211356-83211356 on Build GRCh38
rs568363326 Chr.11: 83211626-83211626 on Build GRCh38
rs919305719 Chr.11: 83211331-83211331 on Build GRCh38
rs1045058251 Chr.11: 83211566-83211566 on Build GRCh38
rs909300914 Chr.11: 83211655-83211655 on Build GRCh38
rs1044598563 Chr.11: 83211695-83211695 on Build GRCh38
rs765861847 Chr.11: 83211437-83211437 on Build GRCh38
rs7106979 Chr.11: 83211801-83211801 on Build GRCh38
rs200518936 Chr.11: 83211401-83211401 on Build GRCh38
rs761893987 Chr.11: 83211411-83211411 on Build GRCh38
rs1019454510 Chr.11: 83211542-83211542 on Build GRCh38
rs56933662 Chr.11: 83211528-83211528 on Build GRCh38
rs112861384 Chr.11: 83211659-83211659 on Build GRCh38
rs754591308 Chr.11: 83211446-83211446 on Build GRCh38
rs1031357174 Chr.11: 83211438-83211438 on Build GRCh38
rs534968134 Chr.11: 83211800-83211800 on Build GRCh38
rs149846444 Chr.11: 83211594-83211594 on Build GRCh38
rs372502718 Chr.11: 83211465-83211465 on Build GRCh38
rs775587249 Chr.11: 83211365-83211365 on Build GRCh38
rs1011721136 Chr.11: 83211649-83211649 on Build GRCh38
rs757735593 Chr.11: 83211347-83211347 on Build GRCh38
rs894615393 Chr.11: 83211633-83211633 on Build GRCh38
rs1010155889 Chr.11: 83211704-83211704 on Build GRCh38
rs984934055 Chr.11: 83211654-83211654 on Build GRCh38
rs1049676081 Chr.11: 83211367-83211367 on Build GRCh38
rs866828553 Chr.11: 83211512-83211512 on Build GRCh38
rs749472747 Chr.11: 83211366-83211366 on Build GRCh38
rs74982233 Chr.11: 83211635-83211635 on Build GRCh38
rs1053186632 Chr.11: 83211744-83211744 on Build GRCh38
rs147507852 Chr.11: 83211338-83211338 on Build GRCh38
rs965276353 Chr.11: 83211573-83211573 on Build GRCh38
rs978412950 Chr.11: 83211797-83211797 on Build GRCh38
rs988615599 Chr.11: 83211392-83211392 on Build GRCh38
rs887980199 Chr.11: 83211372-83211372 on Build GRCh38
rs752589049 Chr.11: 83211669-83211669 on Build GRCh38
rs1036415077 Chr.11: 83211469-83211469 on Build GRCh38
rs377748743 Chr.11: 83211416-83211416 on Build GRCh38
rs953918245 Chr.11: 83211523-83211523 on Build GRCh38
rs1013173533 Chr.11: 83211387-83211387 on Build GRCh38
rs191959822 Chr.11: 83211756-83211756 on Build GRCh38
rs912880379 Chr.11: 83211394-83211394 on Build GRCh38
rs745929198 Chr.11: 83211355-83211355 on Build GRCh38
rs1051579969 Chr.11: 83211417-83211419 on Build GRCh38
rs929367948 Chr.11: 83211650-83211650 on Build GRCh38
rs34954592 Chr.11: 83211441-83211442 on Build GRCh38
rs764886996 Chr.11: 83211606-83211606 on Build GRCh38
rs145426555 Chr.11: 83211415-83211415 on Build GRCh38
rs796348054 Chr.11: 83211633-83211634 on Build GRCh38
rs889538183 Chr.11: 83211822-83211822 on Build GRCh38
rs530084973 Chr.11: 83211632-83211632 on Build GRCh38
rs921820096 Chr.11: 83211414-83211415 on Build GRCh38
rs769845467 Chr.11: 83211364-83211364 on Build GRCh38
rs985262033 Chr.11: 83211539-83211539 on Build GRCh38
rs760111567 Chr.11: 83211431-83211431 on Build GRCh38
rs7106850 Chr.11: 83211682-83211682 on Build GRCh38
rs907433057 Chr.11: 83211706-83211706 on Build GRCh38
rs372106815 Chr.11: 83211664-83211664 on Build GRCh38
rs140132570 Chr.11: 83211385-83211385 on Build GRCh38
rs1021870735 Chr.11: 83211522-83211522 on Build GRCh38
rs537590915 Chr.11: 83211758-83211758 on Build GRCh38

View Full Product Details