Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83227623-83228171 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD42
Entrez Gene ID
338699
Gene Name
ankyrin repeat domain 42
Gene Aliases
PPP1R79, SARP
UniGene
Hs.503438
Gene Chromosome Location
Chr.11: 83193739-83256099 on Build GRCh38
Transcripts
NM_001300972.1 NM_001300973.1 NM_001300975.1 NM_182603.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
932557 c.761A>C p.K254T Chr.11: 83227804-83227804 on Build GRCh38
278852 c.788T>G p.I263S Chr.11: 83227831-83227831 on Build GRCh38
127317 c.813A>C p.S271S Chr.11: 83227856-83227856 on Build GRCh38
4037109 c.816T>A p.T272T Chr.11: 83227859-83227859 on Build GRCh38
932556 c.759G>T p.K253N Chr.11: 83227802-83227802 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761199145 Chr.11: 83228007-83228007 on Build GRCh38
rs773448447 Chr.11: 83227820-83227820 on Build GRCh38
rs374797808 Chr.11: 83227816-83227816 on Build GRCh38
rs77990513 Chr.11: 83227743-83227743 on Build GRCh38
rs531387185 Chr.11: 83228073-83228073 on Build GRCh38
rs781513569 Chr.11: 83227908-83227908 on Build GRCh38
rs536189501 Chr.11: 83227840-83227840 on Build GRCh38
rs749850438 Chr.11: 83227783-83227783 on Build GRCh38
rs753036830 Chr.11: 83227815-83227815 on Build GRCh38
rs988418290 Chr.11: 83227931-83227931 on Build GRCh38
rs756837444 Chr.11: 83227899-83227899 on Build GRCh38
rs757011073 Chr.11: 83227839-83227839 on Build GRCh38
rs548364350 Chr.11: 83228127-83228127 on Build GRCh38
rs552503571 Chr.11: 83227928-83227928 on Build GRCh38
rs756956049 Chr.11: 83228057-83228057 on Build GRCh38
rs564892668 Chr.11: 83228075-83228075 on Build GRCh38
rs887665761 Chr.11: 83228088-83228088 on Build GRCh38
rs200403348 Chr.11: 83227713-83227713 on Build GRCh38
rs752600872 Chr.11: 83227661-83227661 on Build GRCh38
rs919119732 Chr.11: 83228054-83228054 on Build GRCh38
rs750703814 Chr.11: 83228064-83228064 on Build GRCh38
rs765157526 Chr.11: 83228068-83228071 on Build GRCh38
rs1038468282 Chr.11: 83227914-83227914 on Build GRCh38
rs528251148 Chr.11: 83227868-83227868 on Build GRCh38
rs752621704 Chr.11: 83227848-83227848 on Build GRCh38
rs755857564 Chr.11: 83227910-83227910 on Build GRCh38
rs11820761 Chr.11: 83228081-83228081 on Build GRCh38
rs372844297 Chr.11: 83227760-83227760 on Build GRCh38
rs777376318 Chr.11: 83227889-83227889 on Build GRCh38
rs766240576 Chr.11: 83227836-83227836 on Build GRCh38
rs772172870 Chr.11: 83227923-83227923 on Build GRCh38
rs967757315 Chr.11: 83227811-83227811 on Build GRCh38
rs1045031999 Chr.11: 83228103-83228103 on Build GRCh38
rs1005972993 Chr.11: 83228101-83228101 on Build GRCh38
rs186174858 Chr.11: 83227907-83227907 on Build GRCh38
rs769031334 Chr.11: 83227786-83227786 on Build GRCh38
rs148077511 Chr.11: 83227825-83227825 on Build GRCh38
rs1053181698 Chr.11: 83228006-83228006 on Build GRCh38
rs955366671 Chr.11: 83227851-83227851 on Build GRCh38
rs986725013 Chr.11: 83228115-83228115 on Build GRCh38
rs111986663 Chr.11: 83227773-83227773 on Build GRCh38
rs765484686 Chr.11: 83227817-83227818 on Build GRCh38
rs376922138 Chr.11: 83227912-83227912 on Build GRCh38
rs1040519676 Chr.11: 83228042-83228042 on Build GRCh38
rs758413904 Chr.11: 83227677-83227677 on Build GRCh38
rs1034729529 Chr.11: 83227746-83227746 on Build GRCh38
rs763982681 Chr.11: 83227970-83227970 on Build GRCh38
rs780249161 Chr.11: 83227781-83227781 on Build GRCh38
rs919899148 Chr.11: 83227918-83227918 on Build GRCh38
rs1022059512 Chr.11: 83228029-83228029 on Build GRCh38
rs944512565 Chr.11: 83228027-83228027 on Build GRCh38
rs754843366 Chr.11: 83227710-83227710 on Build GRCh38
rs985781661 Chr.11: 83227998-83227998 on Build GRCh38
rs974140028 Chr.11: 83228138-83228138 on Build GRCh38
rs751297081 Chr.11: 83227890-83227890 on Build GRCh38
rs138224683 Chr.11: 83227987-83227987 on Build GRCh38
rs948776535 Chr.11: 83227925-83227925 on Build GRCh38
rs758003398 Chr.11: 83227857-83227857 on Build GRCh38
rs1050620664 Chr.11: 83227708-83227708 on Build GRCh38
rs757911708 Chr.11: 83227731-83227731 on Build GRCh38
rs1009246511 Chr.11: 83228015-83228015 on Build GRCh38
rs202238819 Chr.11: 83227828-83227828 on Build GRCh38
rs544836083 Chr.11: 83227694-83227694 on Build GRCh38
rs79522535 Chr.11: 83227748-83227748 on Build GRCh38
rs903504461 Chr.11: 83228047-83228047 on Build GRCh38
rs956847717 Chr.11: 83227927-83227927 on Build GRCh38
rs542327405 Chr.11: 83227952-83227952 on Build GRCh38
rs144403536 Chr.11: 83228124-83228124 on Build GRCh38
rs938842686 Chr.11: 83227994-83227994 on Build GRCh38
rs748607846 Chr.11: 83227919-83227919 on Build GRCh38
rs367715172 Chr.11: 83227900-83227900 on Build GRCh38

View Full Product Details