Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83261069-83261587 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
UniGene
Hs.368866
Gene Chromosome Location
Chr.11: 83259093-83286407 on Build GRCh38
Transcripts
NM_001286116.1 NM_001286117.1 NM_001286118.1 NM_001286119.1 NM_001286120.1 NM_021825.4 XM_005274154.2 XM_005274156.3 XM_005274159.3 XM_017018103.1 XM_017018104.1

SNP Detail

SNP ID Chromosome Location
rs960293793 Chr.11: 83261157-83261157 on Build GRCh38
rs912861429 Chr.11: 83261178-83261178 on Build GRCh38
rs1001201333 Chr.11: 83261130-83261130 on Build GRCh38
rs201544618 Chr.11: 83261313-83261313 on Build GRCh38
rs539527164 Chr.11: 83261493-83261493 on Build GRCh38
rs779499990 Chr.11: 83261409-83261409 on Build GRCh38
rs140839516 Chr.11: 83261359-83261359 on Build GRCh38
rs1035959469 Chr.11: 83261114-83261114 on Build GRCh38
rs768099538 Chr.11: 83261304-83261304 on Build GRCh38
rs988884590 Chr.11: 83261174-83261174 on Build GRCh38
rs978638942 Chr.11: 83261535-83261535 on Build GRCh38
rs1001723574 Chr.11: 83261440-83261440 on Build GRCh38
rs780835347 Chr.11: 83261490-83261490 on Build GRCh38
rs865899704 Chr.11: 83261327-83261327 on Build GRCh38
rs987930225 Chr.11: 83261305-83261305 on Build GRCh38
rs563441807 Chr.11: 83261339-83261339 on Build GRCh38
rs1003681788 Chr.11: 83261427-83261427 on Build GRCh38
rs886481701 Chr.11: 83261385-83261385 on Build GRCh38
rs968654353 Chr.11: 83261251-83261251 on Build GRCh38
rs988515714 Chr.11: 83261179-83261179 on Build GRCh38
rs978384048 Chr.11: 83261508-83261508 on Build GRCh38
rs1045230683 Chr.11: 83261310-83261310 on Build GRCh38
rs571008021 Chr.11: 83261301-83261301 on Build GRCh38
rs929876777 Chr.11: 83261357-83261357 on Build GRCh38
rs905336915 Chr.11: 83261414-83261414 on Build GRCh38
rs547267517 Chr.11: 83261314-83261314 on Build GRCh38
rs959947089 Chr.11: 83261162-83261162 on Build GRCh38
rs978586611 Chr.11: 83261255-83261255 on Build GRCh38
rs189840861 Chr.11: 83261439-83261439 on Build GRCh38
rs1006209719 Chr.11: 83261407-83261407 on Build GRCh38
rs112822194 Chr.11: 83261211-83261211 on Build GRCh38
rs530885375 Chr.11: 83261099-83261099 on Build GRCh38
rs910983604 Chr.11: 83261558-83261558 on Build GRCh38
rs960575286 Chr.11: 83261464-83261464 on Build GRCh38
rs1020604899 Chr.11: 83261223-83261223 on Build GRCh38
rs536174261 Chr.11: 83261421-83261421 on Build GRCh38
rs556969769 Chr.11: 83261429-83261429 on Build GRCh38
rs1010306633 Chr.11: 83261467-83261467 on Build GRCh38
rs530699060 Chr.11: 83261501-83261501 on Build GRCh38
rs911950589 Chr.11: 83261562-83261562 on Build GRCh38
rs989108596 Chr.11: 83261482-83261482 on Build GRCh38
rs1036393696 Chr.11: 83261332-83261332 on Build GRCh38
rs747520175 Chr.11: 83261158-83261158 on Build GRCh38
rs940667883 Chr.11: 83261314-83261319 on Build GRCh38
rs987343826 Chr.11: 83261561-83261561 on Build GRCh38
rs765813845 Chr.11: 83261379-83261382 on Build GRCh38
rs527576 Chr.11: 83261256-83261256 on Build GRCh38
rs1035453935 Chr.11: 83261139-83261139 on Build GRCh38
rs1028926243 Chr.11: 83261539-83261539 on Build GRCh38
rs1035822633 Chr.11: 83261455-83261455 on Build GRCh38
rs901088929 Chr.11: 83261347-83261347 on Build GRCh38
rs879504534 Chr.11: 83261190-83261190 on Build GRCh38
rs748608866 Chr.11: 83261422-83261422 on Build GRCh38
rs1040826550 Chr.11: 83261319-83261319 on Build GRCh38
rs773782061 Chr.11: 83261529-83261529 on Build GRCh38

View Full Product Details