Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83261434-83261961 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
UniGene
Hs.368866
Gene Chromosome Location
Chr.11: 83259093-83286407 on Build GRCh38
Transcripts
NM_001286116.1 NM_001286117.1 NM_001286118.1 NM_001286119.1 NM_001286120.1 NM_021825.4 XM_005274154.2 XM_005274156.3 XM_005274159.3 XM_017018103.1 XM_017018104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322125 c.741G>C p.L247F Chr.11: 83261935-83261935 on Build GRCh38
467492 c.729G>A p.L243L Chr.11: 83261947-83261947 on Build GRCh38
6134863 c.725G>C p.R242T Chr.11: 83261924-83261924 on Build GRCh38
6134862 c.752G>C p.R251T Chr.11: 83261924-83261924 on Build GRCh38
4574840 c.732A>G p.A244A Chr.11: 83261944-83261944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs761912727 Chr.11: 83261884-83261884 on Build GRCh38
rs1001583166 Chr.11: 83261781-83261781 on Build GRCh38
rs769533899 Chr.11: 83261717-83261717 on Build GRCh38
rs563319955 Chr.11: 83261808-83261808 on Build GRCh38
rs1057460257 Chr.11: 83261790-83261790 on Build GRCh38
rs761109071 Chr.11: 83261575-83261575 on Build GRCh38
rs539527164 Chr.11: 83261493-83261493 on Build GRCh38
rs879727969 Chr.11: 83261939-83261939 on Build GRCh38
rs564467675 Chr.11: 83261846-83261846 on Build GRCh38
rs765016739 Chr.11: 83261927-83261927 on Build GRCh38
rs926889198 Chr.11: 83261915-83261915 on Build GRCh38
rs762784761 Chr.11: 83261916-83261916 on Build GRCh38
rs540008405 Chr.11: 83261767-83261767 on Build GRCh38
rs978638942 Chr.11: 83261535-83261535 on Build GRCh38
rs922495211 Chr.11: 83261652-83261652 on Build GRCh38
rs754651505 Chr.11: 83261921-83261921 on Build GRCh38
rs780835347 Chr.11: 83261490-83261490 on Build GRCh38
rs182121966 Chr.11: 83261791-83261791 on Build GRCh38
rs929796217 Chr.11: 83261679-83261679 on Build GRCh38
rs552628241 Chr.11: 83261820-83261820 on Build GRCh38
rs942428533 Chr.11: 83261570-83261570 on Build GRCh38
rs767392092 Chr.11: 83261888-83261888 on Build GRCh38
rs762392819 Chr.11: 83261730-83261731 on Build GRCh38
rs926897511 Chr.11: 83261707-83261707 on Build GRCh38
rs572991452 Chr.11: 83261647-83261647 on Build GRCh38
rs978384048 Chr.11: 83261508-83261508 on Build GRCh38
rs552418186 Chr.11: 83261728-83261729 on Build GRCh38
rs1009946301 Chr.11: 83261750-83261750 on Build GRCh38
rs1037683487 Chr.11: 83261732-83261732 on Build GRCh38
rs28930683 Chr.11: 83261813-83261813 on Build GRCh38
rs187174441 Chr.11: 83261873-83261873 on Build GRCh38
rs1018741719 Chr.11: 83261806-83261806 on Build GRCh38
rs150150415 Chr.11: 83261573-83261573 on Build GRCh38
rs370521192 Chr.11: 83261896-83261896 on Build GRCh38
rs999879277 Chr.11: 83261786-83261786 on Build GRCh38
rs942025238 Chr.11: 83261719-83261719 on Build GRCh38
rs144006437 Chr.11: 83261918-83261918 on Build GRCh38
rs1017862461 Chr.11: 83261814-83261814 on Build GRCh38
rs909820909 Chr.11: 83261919-83261919 on Build GRCh38
rs207472085 Chr.11: 83261802-83261802 on Build GRCh38
rs910983604 Chr.11: 83261558-83261558 on Build GRCh38
rs960575286 Chr.11: 83261464-83261464 on Build GRCh38
rs373174461 Chr.11: 83261851-83261851 on Build GRCh38
rs1057118120 Chr.11: 83261937-83261937 on Build GRCh38
rs963675092 Chr.11: 83261819-83261819 on Build GRCh38
rs920557053 Chr.11: 83261576-83261576 on Build GRCh38
rs747874610 Chr.11: 83261720-83261720 on Build GRCh38
rs1010306633 Chr.11: 83261467-83261467 on Build GRCh38
rs374190990 Chr.11: 83261882-83261882 on Build GRCh38
rs530699060 Chr.11: 83261501-83261501 on Build GRCh38
rs911950589 Chr.11: 83261562-83261562 on Build GRCh38
rs370520085 Chr.11: 83261861-83261861 on Build GRCh38
rs768916127 Chr.11: 83261881-83261881 on Build GRCh38
rs1010802341 Chr.11: 83261801-83261801 on Build GRCh38
rs766080249 Chr.11: 83261904-83261904 on Build GRCh38
rs989108596 Chr.11: 83261482-83261482 on Build GRCh38
rs758749604 Chr.11: 83261729-83261730 on Build GRCh38
rs930654501 Chr.11: 83261671-83261671 on Build GRCh38
rs775991525 Chr.11: 83261862-83261862 on Build GRCh38
rs747032998 Chr.11: 83261908-83261909 on Build GRCh38
rs987343826 Chr.11: 83261561-83261561 on Build GRCh38
rs374958155 Chr.11: 83261584-83261584 on Build GRCh38
rs561931080 Chr.11: 83261783-83261783 on Build GRCh38
rs1028926243 Chr.11: 83261539-83261539 on Build GRCh38
rs760449609 Chr.11: 83261900-83261900 on Build GRCh38
rs568695215 Chr.11: 83261938-83261938 on Build GRCh38
rs951276934 Chr.11: 83261910-83261910 on Build GRCh38
rs1046998635 Chr.11: 83261704-83261704 on Build GRCh38
rs773782061 Chr.11: 83261529-83261529 on Build GRCh38

View Full Product Details