Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 83278405-83278908 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC90B
Entrez Gene ID
60492
Gene Name
coiled-coil domain containing 90B
Gene Aliases
MDS011, MDS025
UniGene
Hs.368866
Gene Chromosome Location
Chr.11: 83259093-83286407 on Build GRCh38
Transcripts
NM_001286116.1 NM_001286117.1 NM_001286118.1 NM_001286119.1 NM_001286120.1 NM_021825.4 XM_005274154.2 XM_005274156.3 XM_005274159.3 XM_017018103.1 XM_017018104.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7145769 c.272C>T p.S91L Chr.11: 83278778-83278778 on Build GRCh38
6622665 c.234A>G p.L78L Chr.11: 83278789-83278789 on Build GRCh38
6622664 c.261A>G p.L87L Chr.11: 83278789-83278789 on Build GRCh38
6375030 c.280G>T p.V94F Chr.11: 83278743-83278743 on Build GRCh38
3453949 c.286G>C p.D96H Chr.11: 83278764-83278764 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373524286 Chr.11: 83278800-83278800 on Build GRCh38
rs908563529 Chr.11: 83278438-83278438 on Build GRCh38
rs781382555 Chr.11: 83278782-83278782 on Build GRCh38
rs757668854 Chr.11: 83278844-83278844 on Build GRCh38
rs117072241 Chr.11: 83278729-83278729 on Build GRCh38
rs750064661 Chr.11: 83278740-83278741 on Build GRCh38
rs919089934 Chr.11: 83278775-83278775 on Build GRCh38
rs779703567 Chr.11: 83278479-83278479 on Build GRCh38
rs1003678259 Chr.11: 83278520-83278520 on Build GRCh38
rs200299445 Chr.11: 83278701-83278701 on Build GRCh38
rs974331237 Chr.11: 83278503-83278503 on Build GRCh38
rs561360815 Chr.11: 83278631-83278631 on Build GRCh38
rs746162020 Chr.11: 83278853-83278853 on Build GRCh38
rs761416733 Chr.11: 83278736-83278737 on Build GRCh38
rs751454918 Chr.11: 83278789-83278790 on Build GRCh38
rs1009146733 Chr.11: 83278717-83278717 on Build GRCh38
rs941472382 Chr.11: 83278464-83278464 on Build GRCh38
rs764820458 Chr.11: 83278838-83278838 on Build GRCh38
rs554253117 Chr.11: 83278884-83278884 on Build GRCh38
rs905379920 Chr.11: 83278510-83278510 on Build GRCh38
rs554455408 Chr.11: 83278854-83278855 on Build GRCh38
rs193040119 Chr.11: 83278803-83278803 on Build GRCh38
rs948990385 Chr.11: 83278574-83278574 on Build GRCh38
rs754545270 Chr.11: 83278749-83278749 on Build GRCh38
rs756476568 Chr.11: 83278857-83278857 on Build GRCh38
rs781735874 Chr.11: 83278850-83278850 on Build GRCh38
rs372037684 Chr.11: 83278833-83278833 on Build GRCh38
rs77724203 Chr.11: 83278589-83278589 on Build GRCh38
rs757834789 Chr.11: 83278776-83278776 on Build GRCh38
rs752293988 Chr.11: 83278839-83278839 on Build GRCh38
rs191647222 Chr.11: 83278708-83278708 on Build GRCh38
rs907350807 Chr.11: 83278592-83278592 on Build GRCh38
rs149196251 Chr.11: 83278692-83278692 on Build GRCh38
rs939122439 Chr.11: 83278615-83278615 on Build GRCh38
rs753345047 Chr.11: 83278743-83278743 on Build GRCh38
rs780464727 Chr.11: 83278863-83278863 on Build GRCh38
rs200672198 Chr.11: 83278793-83278793 on Build GRCh38
rs1009802913 Chr.11: 83278624-83278624 on Build GRCh38
rs967709948 Chr.11: 83278655-83278655 on Build GRCh38
rs538799582 Chr.11: 83278731-83278731 on Build GRCh38
rs767201605 Chr.11: 83278721-83278721 on Build GRCh38
rs1035176928 Chr.11: 83278549-83278549 on Build GRCh38
rs374510472 Chr.11: 83278784-83278784 on Build GRCh38
rs768913821 Chr.11: 83278869-83278869 on Build GRCh38
rs775512473 Chr.11: 83278686-83278686 on Build GRCh38
rs143522058 Chr.11: 83278820-83278820 on Build GRCh38
rs766067420 Chr.11: 83278822-83278822 on Build GRCh38
rs754741615 Chr.11: 83278835-83278835 on Build GRCh38
rs373329593 Chr.11: 83278879-83278879 on Build GRCh38
rs766270509 Chr.11: 83278746-83278747 on Build GRCh38
rs754907184 Chr.11: 83278822-83278822 on Build GRCh38
rs778355526 Chr.11: 83278755-83278755 on Build GRCh38
rs747463863 Chr.11: 83278769-83278769 on Build GRCh38
rs939975293 Chr.11: 83278471-83278471 on Build GRCh38
rs536980942 Chr.11: 83278814-83278814 on Build GRCh38
rs188393866 Chr.11: 83278732-83278732 on Build GRCh38
rs148847806 Chr.11: 83278792-83278792 on Build GRCh38
rs764747212 Chr.11: 83278737-83278738 on Build GRCh38
rs920133268 Chr.11: 83278534-83278534 on Build GRCh38
rs761878692 Chr.11: 83278710-83278710 on Build GRCh38
rs758160898 Chr.11: 83278742-83278743 on Build GRCh38

View Full Product Details