Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85657720-85658267 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREBZF
Entrez Gene ID
58487
Gene Name
CREB/ATF bZIP transcription factor
Gene Aliases
SMILE, ZF
UniGene
Hs.535319
Gene Chromosome Location
Chr.11: 85657564-85665148 on Build GRCh38
Transcripts
NM_001039618.2 NR_028024.1 NR_028025.1 NR_028026.1 NR_028027.1 XM_006718642.3 XM_011545195.2 XM_017018086.1 XM_017018087.1 XM_017018088.1 XM_017018089.1 XM_017018090.1 XM_017018091.1 XM_017018092.1

SNP Detail

SNP ID Chromosome Location
rs192846370 Chr.11: 85657889-85657889 on Build GRCh38
rs979687496 Chr.11: 85658165-85658165 on Build GRCh38
rs1050157328 Chr.11: 85658220-85658220 on Build GRCh38
rs138720736 Chr.11: 85657968-85657968 on Build GRCh38
rs145809435 Chr.11: 85657868-85657868 on Build GRCh38
rs34417921 Chr.11: 85658213-85658214 on Build GRCh38
rs530711652 Chr.11: 85657750-85657750 on Build GRCh38
rs994802172 Chr.11: 85658090-85658090 on Build GRCh38
rs528111379 Chr.11: 85657984-85657984 on Build GRCh38
rs977441982 Chr.11: 85658180-85658180 on Build GRCh38
rs923881852 Chr.11: 85657876-85657876 on Build GRCh38
rs868065831 Chr.11: 85658170-85658170 on Build GRCh38
rs560020557 Chr.11: 85658052-85658052 on Build GRCh38
rs761001727 Chr.11: 85657981-85657981 on Build GRCh38
rs533380475 Chr.11: 85658200-85658200 on Build GRCh38
rs923828072 Chr.11: 85658205-85658205 on Build GRCh38
rs969504305 Chr.11: 85657766-85657766 on Build GRCh38
rs571693932 Chr.11: 85658111-85658111 on Build GRCh38
rs1991923 Chr.11: 85658154-85658154 on Build GRCh38
rs958831472 Chr.11: 85657771-85657771 on Build GRCh38
rs775598305 Chr.11: 85658131-85658131 on Build GRCh38
rs1028131175 Chr.11: 85657760-85657760 on Build GRCh38
rs756367575 Chr.11: 85658169-85658169 on Build GRCh38
rs149035927 Chr.11: 85657864-85657864 on Build GRCh38
rs928147174 Chr.11: 85657817-85657817 on Build GRCh38
rs1050627337 Chr.11: 85657979-85657979 on Build GRCh38
rs1035539258 Chr.11: 85658140-85658140 on Build GRCh38
rs890122910 Chr.11: 85657947-85657947 on Build GRCh38
rs551189762 Chr.11: 85657861-85657861 on Build GRCh38
rs898101835 Chr.11: 85657985-85657985 on Build GRCh38
rs989682148 Chr.11: 85658235-85658235 on Build GRCh38
rs550812290 Chr.11: 85657767-85657767 on Build GRCh38
rs574024467 Chr.11: 85657903-85657903 on Build GRCh38
rs991864393 Chr.11: 85657783-85657783 on Build GRCh38
rs542258778 Chr.11: 85658125-85658125 on Build GRCh38
rs1027830311 Chr.11: 85658059-85658059 on Build GRCh38
rs914174240 Chr.11: 85657885-85657885 on Build GRCh38
rs187785918 Chr.11: 85658192-85658192 on Build GRCh38
rs1053634427 Chr.11: 85657895-85657895 on Build GRCh38
rs568147610 Chr.11: 85658139-85658139 on Build GRCh38
rs577778865 Chr.11: 85657995-85657995 on Build GRCh38
rs574638027 Chr.11: 85658053-85658053 on Build GRCh38
rs74733979 Chr.11: 85658011-85658011 on Build GRCh38
rs1050642342 Chr.11: 85657930-85657930 on Build GRCh38
rs1036249089 Chr.11: 85658009-85658009 on Build GRCh38
rs187543864 Chr.11: 85657820-85657820 on Build GRCh38
rs960882808 Chr.11: 85658153-85658153 on Build GRCh38
rs569352051 Chr.11: 85657774-85657774 on Build GRCh38
rs570306625 Chr.11: 85657906-85657906 on Build GRCh38
rs914115984 Chr.11: 85658155-85658155 on Build GRCh38
rs971425838 Chr.11: 85657842-85657842 on Build GRCh38
rs192464519 Chr.11: 85658229-85658229 on Build GRCh38
rs539878262 Chr.11: 85657815-85657815 on Build GRCh38
rs769786838 Chr.11: 85658102-85658102 on Build GRCh38
rs1009163608 Chr.11: 85657988-85657988 on Build GRCh38
rs747389934 Chr.11: 85657790-85657790 on Build GRCh38
rs183097653 Chr.11: 85658160-85658160 on Build GRCh38
rs969829208 Chr.11: 85658128-85658128 on Build GRCh38
rs536726741 Chr.11: 85658149-85658149 on Build GRCh38

View Full Product Details