Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 85911247-85911773 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC83
Entrez Gene ID
220047
Gene Name
coiled-coil domain containing 83
Gene Aliases
CT148, HSD9
UniGene
Hs.567774
Gene Chromosome Location
Chr.11: 85855066-85920021 on Build GRCh38
Transcripts
NM_001286159.1 NM_173556.4 XM_011544839.2 XM_011544840.2 XM_011544841.1 XM_011544842.1 XM_011544843.1 XM_011544844.2 XM_017017354.1 XM_017017355.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4556438 c.691G>A p.E231K Chr.11: 85911299-85911299 on Build GRCh38
6324778 c.762C>T p.S254S Chr.11: 85911370-85911370 on Build GRCh38
6622505 c.733G>A p.E245K Chr.11: 85911341-85911341 on Build GRCh38
5528286 c.718C>T p.H240Y Chr.11: 85911326-85911326 on Build GRCh38
4037209 c.752A>T p.D251V Chr.11: 85911360-85911360 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs929889194 Chr.11: 85911687-85911687 on Build GRCh38
rs968125282 Chr.11: 85911614-85911614 on Build GRCh38
rs747360405 Chr.11: 85911399-85911399 on Build GRCh38
rs1005305119 Chr.11: 85911468-85911468 on Build GRCh38
rs141952527 Chr.11: 85911396-85911396 on Build GRCh38
rs1042311703 Chr.11: 85911744-85911744 on Build GRCh38
rs199964272 Chr.11: 85911317-85911317 on Build GRCh38
rs943359892 Chr.11: 85911667-85911667 on Build GRCh38
rs141851299 Chr.11: 85911324-85911324 on Build GRCh38
rs895997077 Chr.11: 85911483-85911483 on Build GRCh38
rs773306677 Chr.11: 85911445-85911445 on Build GRCh38
rs901125869 Chr.11: 85911435-85911445 on Build GRCh38
rs760747316 Chr.11: 85911579-85911579 on Build GRCh38
rs913952219 Chr.11: 85911364-85911364 on Build GRCh38
rs746134258 Chr.11: 85911410-85911410 on Build GRCh38
rs748549556 Chr.11: 85911382-85911382 on Build GRCh38
rs1032015299 Chr.11: 85911645-85911645 on Build GRCh38
rs761630181 Chr.11: 85911436-85911436 on Build GRCh38
rs368591219 Chr.11: 85911475-85911475 on Build GRCh38
rs137945995 Chr.11: 85911315-85911315 on Build GRCh38
rs764178619 Chr.11: 85911353-85911353 on Build GRCh38
rs1002885140 Chr.11: 85911488-85911488 on Build GRCh38
rs563282501 Chr.11: 85911470-85911470 on Build GRCh38
rs767217143 Chr.11: 85911380-85911380 on Build GRCh38
rs1014865862 Chr.11: 85911563-85911563 on Build GRCh38
rs924937940 Chr.11: 85911367-85911367 on Build GRCh38
rs144109835 Chr.11: 85911725-85911725 on Build GRCh38
rs1001840307 Chr.11: 85911750-85911750 on Build GRCh38
rs114865543 Chr.11: 85911690-85911690 on Build GRCh38
rs148135893 Chr.11: 85911387-85911387 on Build GRCh38
rs762973972 Chr.11: 85911347-85911347 on Build GRCh38
rs753335846 Chr.11: 85911388-85911388 on Build GRCh38
rs146914384 Chr.11: 85911737-85911737 on Build GRCh38
rs556278072 Chr.11: 85911577-85911577 on Build GRCh38
rs571898796 Chr.11: 85911523-85911523 on Build GRCh38
rs755537419 Chr.11: 85911383-85911383 on Build GRCh38
rs970083973 Chr.11: 85911280-85911280 on Build GRCh38
rs943840339 Chr.11: 85911407-85911407 on Build GRCh38
rs965140700 Chr.11: 85911671-85911671 on Build GRCh38
rs374592744 Chr.11: 85911474-85911474 on Build GRCh38
rs781324817 Chr.11: 85911409-85911409 on Build GRCh38
rs140532988 Chr.11: 85911482-85911483 on Build GRCh38
rs532629255 Chr.11: 85911531-85911531 on Build GRCh38
rs961166633 Chr.11: 85911545-85911545 on Build GRCh38
rs550253573 Chr.11: 85911497-85911497 on Build GRCh38
rs901019989 Chr.11: 85911419-85911419 on Build GRCh38
rs977030892 Chr.11: 85911673-85911673 on Build GRCh38
rs143833799 Chr.11: 85911359-85911359 on Build GRCh38
rs1046971401 Chr.11: 85911698-85911698 on Build GRCh38
rs771136585 Chr.11: 85911313-85911313 on Build GRCh38
rs922511339 Chr.11: 85911676-85911676 on Build GRCh38
rs969070269 Chr.11: 85911472-85911472 on Build GRCh38
rs530470461 Chr.11: 85911471-85911471 on Build GRCh38
rs866797711 Chr.11: 85911349-85911349 on Build GRCh38
rs146932939 Chr.11: 85911303-85911303 on Build GRCh38
rs537726488 Chr.11: 85911334-85911334 on Build GRCh38
rs199965022 Chr.11: 85911420-85911420 on Build GRCh38
rs761771100 Chr.11: 85911363-85911363 on Build GRCh38
rs1039593490 Chr.11: 85911403-85911403 on Build GRCh38
rs752913568 Chr.11: 85911374-85911374 on Build GRCh38
rs937312205 Chr.11: 85911718-85911718 on Build GRCh38
rs988300941 Chr.11: 85911561-85911561 on Build GRCh38
rs148480771 Chr.11: 85911475-85911476 on Build GRCh38
rs781191130 Chr.11: 85911550-85911550 on Build GRCh38
rs554013433 Chr.11: 85911293-85911293 on Build GRCh38
rs968072157 Chr.11: 85911586-85911586 on Build GRCh38
rs758905499 Chr.11: 85911393-85911393 on Build GRCh38
rs772689151 Chr.11: 85911451-85911451 on Build GRCh38
rs17853979 Chr.11: 85911288-85911288 on Build GRCh38
rs186677307 Chr.11: 85911627-85911627 on Build GRCh38
rs768715037 Chr.11: 85911425-85911425 on Build GRCh38
rs775400239 Chr.11: 85911668-85911668 on Build GRCh38
rs369317422 Chr.11: 85911435-85911435 on Build GRCh38
rs1005336369 Chr.11: 85911717-85911717 on Build GRCh38
rs541207034 Chr.11: 85911401-85911401 on Build GRCh38
rs769714410 Chr.11: 85911418-85911418 on Build GRCh38
rs758639912 Chr.11: 85911282-85911282 on Build GRCh38
rs756388710 Chr.11: 85911407-85911408 on Build GRCh38
rs957669224 Chr.11: 85911652-85911652 on Build GRCh38
rs749621813 Chr.11: 85911422-85911423 on Build GRCh38
rs543317861 Chr.11: 85911381-85911381 on Build GRCh38
rs566112077 Chr.11: 85911621-85911621 on Build GRCh38
rs778056020 Chr.11: 85911415-85911416 on Build GRCh38
rs372435399 Chr.11: 85911423-85911423 on Build GRCh38
rs762916378 Chr.11: 85911708-85911708 on Build GRCh38
rs575991895 Chr.11: 85911375-85911375 on Build GRCh38
rs767998604 Chr.11: 85911364-85911365 on Build GRCh38

View Full Product Details