Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 90200914-90201421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHORDC1
Entrez Gene ID
26973
Gene Name
cysteine and histidine rich domain containing 1
Gene Aliases
CHP1
UniGene
Hs.112482
Gene Chromosome Location
Chr.11: 90200429-90223364 on Build GRCh38
Transcripts
NM_001144073.1 NM_012124.2 XM_017017540.1 XM_017017541.1

SNP Detail

SNP ID Chromosome Location
rs562269092 Chr.11: 90201060-90201060 on Build GRCh38
rs1037588288 Chr.11: 90201117-90201117 on Build GRCh38
rs762220011 Chr.11: 90201246-90201246 on Build GRCh38
rs1055153656 Chr.11: 90201009-90201009 on Build GRCh38
rs371199562 Chr.11: 90201344-90201344 on Build GRCh38
rs879390858 Chr.11: 90200954-90200954 on Build GRCh38
rs397840939 Chr.11: 90201357-90201358 on Build GRCh38
rs551242768 Chr.11: 90201228-90201228 on Build GRCh38
rs540049400 Chr.11: 90201126-90201126 on Build GRCh38
rs1007242789 Chr.11: 90201247-90201247 on Build GRCh38
rs781526191 Chr.11: 90200941-90200941 on Build GRCh38
rs967355494 Chr.11: 90201389-90201389 on Build GRCh38
rs1021247913 Chr.11: 90201384-90201387 on Build GRCh38
rs993380758 Chr.11: 90201151-90201151 on Build GRCh38
rs982686104 Chr.11: 90200978-90200980 on Build GRCh38
rs752101425 Chr.11: 90201229-90201229 on Build GRCh38
rs3834437 Chr.11: 90201356-90201357 on Build GRCh38
rs561512283 Chr.11: 90201207-90201207 on Build GRCh38
rs773261026 Chr.11: 90201366-90201366 on Build GRCh38
rs767879241 Chr.11: 90201248-90201248 on Build GRCh38
rs1030568612 Chr.11: 90201235-90201235 on Build GRCh38
rs192320274 Chr.11: 90201314-90201314 on Build GRCh38
rs570552796 Chr.11: 90201383-90201383 on Build GRCh38
rs559517464 Chr.11: 90200975-90200975 on Build GRCh38
rs770101539 Chr.11: 90201008-90201008 on Build GRCh38
rs528756528 Chr.11: 90200993-90200993 on Build GRCh38
rs188042057 Chr.11: 90201132-90201132 on Build GRCh38
rs10741326 Chr.11: 90201014-90201014 on Build GRCh38

View Full Product Details