Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 95771168-95771664 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM76B
Entrez Gene ID
143684
Gene Name
family with sequence similarity 76 member B
Gene Aliases
-
UniGene
Hs.288304
Gene Chromosome Location
Chr.11: 95768942-95790396 on Build GRCh38
Transcripts
NM_144664.4 XM_005273775.4 XM_005273776.4 XM_011542610.2 XM_011542611.2 XM_011542612.1 XM_011542613.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6251816 c.1010C>A p.T337K Chr.11: 95771571-95771571 on Build GRCh38
6588135 c.941G>C p.R314T Chr.11: 95771640-95771640 on Build GRCh38
7159709 c.951C>G p.L317L Chr.11: 95771630-95771630 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs903282426 Chr.11: 95771327-95771327 on Build GRCh38
rs982534774 Chr.11: 95771508-95771508 on Build GRCh38
rs760436895 Chr.11: 95771324-95771324 on Build GRCh38
rs754187254 Chr.11: 95771514-95771514 on Build GRCh38
rs757486610 Chr.11: 95771621-95771621 on Build GRCh38
rs373243898 Chr.11: 95771573-95771573 on Build GRCh38
rs760838810 Chr.11: 95771602-95771602 on Build GRCh38
rs769398471 Chr.11: 95771542-95771542 on Build GRCh38
rs776217401 Chr.11: 95771261-95771261 on Build GRCh38
rs549895793 Chr.11: 95771378-95771378 on Build GRCh38
rs921187052 Chr.11: 95771605-95771605 on Build GRCh38
rs998919136 Chr.11: 95771363-95771363 on Build GRCh38
rs757476270 Chr.11: 95771493-95771493 on Build GRCh38
rs771349510 Chr.11: 95771564-95771566 on Build GRCh38
rs752781261 Chr.11: 95771597-95771597 on Build GRCh38
rs771498007 Chr.11: 95771427-95771427 on Build GRCh38
rs1013624007 Chr.11: 95771419-95771419 on Build GRCh38
rs1050878199 Chr.11: 95771249-95771250 on Build GRCh38
rs757652925 Chr.11: 95771515-95771515 on Build GRCh38
rs375781739 Chr.11: 95771566-95771566 on Build GRCh38
rs763831667 Chr.11: 95771362-95771362 on Build GRCh38
rs963354133 Chr.11: 95771289-95771289 on Build GRCh38
rs570661453 Chr.11: 95771270-95771270 on Build GRCh38
rs779212467 Chr.11: 95771521-95771521 on Build GRCh38
rs1026411909 Chr.11: 95771487-95771487 on Build GRCh38
rs200981048 Chr.11: 95771523-95771523 on Build GRCh38
rs894549193 Chr.11: 95771198-95771198 on Build GRCh38
rs764352249 Chr.11: 95771613-95771613 on Build GRCh38
rs567819637 Chr.11: 95771414-95771414 on Build GRCh38
rs899074102 Chr.11: 95771529-95771529 on Build GRCh38
rs946225294 Chr.11: 95771392-95771392 on Build GRCh38
rs775064699 Chr.11: 95771637-95771638 on Build GRCh38
rs184419001 Chr.11: 95771260-95771260 on Build GRCh38
rs752653812 Chr.11: 95771268-95771268 on Build GRCh38
rs987227897 Chr.11: 95771374-95771374 on Build GRCh38
rs906335975 Chr.11: 95771503-95771503 on Build GRCh38
rs1033455013 Chr.11: 95771366-95771366 on Build GRCh38
rs906360641 Chr.11: 95771269-95771269 on Build GRCh38
rs747681751 Chr.11: 95771538-95771538 on Build GRCh38
rs558150856 Chr.11: 95771632-95771632 on Build GRCh38
rs376848080 Chr.11: 95771643-95771643 on Build GRCh38
rs759491549 Chr.11: 95771575-95771575 on Build GRCh38
rs912026253 Chr.11: 95771379-95771379 on Build GRCh38
rs772858364 Chr.11: 95771557-95771557 on Build GRCh38
rs933823012 Chr.11: 95771211-95771211 on Build GRCh38
rs754097720 Chr.11: 95771617-95771617 on Build GRCh38
rs893116550 Chr.11: 95771373-95771375 on Build GRCh38
rs754084051 Chr.11: 95771461-95771461 on Build GRCh38
rs139116319 Chr.11: 95771436-95771436 on Build GRCh38
rs767580908 Chr.11: 95771595-95771595 on Build GRCh38
rs936292992 Chr.11: 95771428-95771428 on Build GRCh38
rs746381454 Chr.11: 95771518-95771518 on Build GRCh38
rs189354757 Chr.11: 95771315-95771315 on Build GRCh38
rs749750689 Chr.11: 95771515-95771516 on Build GRCh38
rs1015266923 Chr.11: 95771287-95771287 on Build GRCh38
rs779304798 Chr.11: 95771623-95771623 on Build GRCh38
rs535138380 Chr.11: 95771433-95771433 on Build GRCh38
rs1004777464 Chr.11: 95771279-95771279 on Build GRCh38
rs568874939 Chr.11: 95771507-95771508 on Build GRCh38
rs369049259 Chr.11: 95771568-95771568 on Build GRCh38
rs973046599 Chr.11: 95771318-95771318 on Build GRCh38
rs779201665 Chr.11: 95771516-95771516 on Build GRCh38
rs565308842 Chr.11: 95771483-95771483 on Build GRCh38

View Full Product Details