Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 99956720-99957226 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNTN5
Entrez Gene ID
53942
Gene Name
contactin 5
Gene Aliases
HNB-2s, NB-2
UniGene
Hs.656783
Gene Chromosome Location
Chr.11: 99020904-100358885 on Build GRCh38
Transcripts
NM_001243270.1 NM_001243271.1 NM_014361.3 NM_175566.2 XM_011542871.1 XM_011542873.1 XM_017017926.1 XM_017017927.1 XM_017017928.1 XM_017017929.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3870501 c.787G>A p.D263N Chr.11: 99956919-99956919 on Build GRCh38
2062250 c.728G>T p.S243I Chr.11: 99956860-99956860 on Build GRCh38
933722 c.784T>G p.S262A Chr.11: 99956916-99956916 on Build GRCh38
5603239 c.678C>T p.I226I Chr.11: 99956810-99956810 on Build GRCh38
6631808 c.869G>A p.R290H Chr.11: 99957001-99957001 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200945111 Chr.11: 99957026-99957026 on Build GRCh38
rs749442209 Chr.11: 99956852-99956852 on Build GRCh38
rs752003777 Chr.11: 99957041-99957041 on Build GRCh38
rs1048462384 Chr.11: 99957082-99957082 on Build GRCh38
rs549484645 Chr.11: 99957032-99957032 on Build GRCh38
rs774698713 Chr.11: 99956813-99956813 on Build GRCh38
rs371755605 Chr.11: 99956856-99956856 on Build GRCh38
rs140553641 Chr.11: 99957079-99957080 on Build GRCh38
rs1010788117 Chr.11: 99957140-99957140 on Build GRCh38
rs376992677 Chr.11: 99956989-99956989 on Build GRCh38
rs779073851 Chr.11: 99956944-99956944 on Build GRCh38
rs749565155 Chr.11: 99956792-99956792 on Build GRCh38
rs998379407 Chr.11: 99957131-99957131 on Build GRCh38
rs190220140 Chr.11: 99956862-99956862 on Build GRCh38
rs897258814 Chr.11: 99957079-99957079 on Build GRCh38
rs749373857 Chr.11: 99957105-99957109 on Build GRCh38
rs997873541 Chr.11: 99957081-99957081 on Build GRCh38
rs1030669886 Chr.11: 99957088-99957088 on Build GRCh38
rs764099716 Chr.11: 99956921-99956921 on Build GRCh38
rs750705149 Chr.11: 99956956-99956956 on Build GRCh38
rs574144977 Chr.11: 99956761-99956761 on Build GRCh38
rs748684510 Chr.11: 99957059-99957059 on Build GRCh38
rs1022549168 Chr.11: 99957152-99957152 on Build GRCh38
rs918880916 Chr.11: 99956914-99956914 on Build GRCh38
rs182352064 Chr.11: 99957197-99957197 on Build GRCh38
rs748840124 Chr.11: 99956995-99956995 on Build GRCh38
rs758676568 Chr.11: 99956960-99956960 on Build GRCh38
rs577962852 Chr.11: 99956851-99956851 on Build GRCh38
rs202063081 Chr.11: 99956806-99956806 on Build GRCh38
rs200102673 Chr.11: 99957018-99957018 on Build GRCh38
rs978434230 Chr.11: 99956990-99956990 on Build GRCh38
rs766211231 Chr.11: 99956844-99956844 on Build GRCh38
rs1034423867 Chr.11: 99957184-99957184 on Build GRCh38
rs760662211 Chr.11: 99956908-99956908 on Build GRCh38
rs751415649 Chr.11: 99956846-99956846 on Build GRCh38
rs764046886 Chr.11: 99957025-99957025 on Build GRCh38
rs557580919 Chr.11: 99956755-99956755 on Build GRCh38
rs199802165 Chr.11: 99956888-99956888 on Build GRCh38
rs752723139 Chr.11: 99956848-99956848 on Build GRCh38
rs572094429 Chr.11: 99956818-99956818 on Build GRCh38
rs369374416 Chr.11: 99956892-99956892 on Build GRCh38
rs540994363 Chr.11: 99956823-99956823 on Build GRCh38
rs1020840120 Chr.11: 99957143-99957143 on Build GRCh38
rs965992934 Chr.11: 99957126-99957126 on Build GRCh38
rs781749561 Chr.11: 99957052-99957052 on Build GRCh38
rs746068381 Chr.11: 99957173-99957173 on Build GRCh38
rs375312209 Chr.11: 99957078-99957079 on Build GRCh38
rs932360963 Chr.11: 99957132-99957132 on Build GRCh38
rs960113690 Chr.11: 99957189-99957189 on Build GRCh38
rs199557862 Chr.11: 99956858-99956858 on Build GRCh38
rs200560123 Chr.11: 99956842-99956842 on Build GRCh38
rs771277271 Chr.11: 99956811-99956811 on Build GRCh38
rs1007289509 Chr.11: 99957102-99957107 on Build GRCh38
rs780533843 Chr.11: 99956961-99956961 on Build GRCh38
rs199898141 Chr.11: 99956991-99956991 on Build GRCh38
rs766153259 Chr.11: 99956891-99956891 on Build GRCh38
rs942463692 Chr.11: 99957065-99957065 on Build GRCh38
rs1039443653 Chr.11: 99957078-99957078 on Build GRCh38
rs773493367 Chr.11: 99956899-99956899 on Build GRCh38
rs773856381 Chr.11: 99957001-99957001 on Build GRCh38
rs888584011 Chr.11: 99957051-99957051 on Build GRCh38
rs563536789 Chr.11: 99956866-99956866 on Build GRCh38
rs917139202 Chr.11: 99957068-99957068 on Build GRCh38
rs969224120 Chr.11: 99957177-99957177 on Build GRCh38
rs755508153 Chr.11: 99957048-99957048 on Build GRCh38
rs775365705 Chr.11: 99957019-99957019 on Build GRCh38
rs201219815 Chr.11: 99956863-99956863 on Build GRCh38
rs949935458 Chr.11: 99957072-99957072 on Build GRCh38
rs145075779 Chr.11: 99957182-99957182 on Build GRCh38
rs757312285 Chr.11: 99956985-99956985 on Build GRCh38
rs200899470 Chr.11: 99956977-99956977 on Build GRCh38
rs751262157 Chr.11: 99956778-99956778 on Build GRCh38
rs777370237 Chr.11: 99956986-99956986 on Build GRCh38
rs370878735 Chr.11: 99956934-99956934 on Build GRCh38
rs767375808 Chr.11: 99956883-99956883 on Build GRCh38
rs370012927 Chr.11: 99957080-99957080 on Build GRCh38
rs761196266 Chr.11: 99956833-99956833 on Build GRCh38
rs766335817 Chr.11: 99956768-99956768 on Build GRCh38
rs371756154 Chr.11: 99956904-99956904 on Build GRCh38
rs772853483 Chr.11: 99956878-99956878 on Build GRCh38
rs764850156 Chr.11: 99956837-99956837 on Build GRCh38
rs199670730 Chr.11: 99956938-99956938 on Build GRCh38
rs756128950 Chr.11: 99956849-99956849 on Build GRCh38
rs765358335 Chr.11: 99957027-99957027 on Build GRCh38
rs759302485 Chr.11: 99957014-99957014 on Build GRCh38
rs772984856 Chr.11: 99956840-99956840 on Build GRCh38
rs529063149 Chr.11: 99956980-99956980 on Build GRCh38
rs202177289 Chr.11: 99956756-99956756 on Build GRCh38
rs780226403 Chr.11: 99957034-99957034 on Build GRCh38
rs200237479 Chr.11: 99956771-99956771 on Build GRCh38
rs200451910 Chr.11: 99956898-99956898 on Build GRCh38
rs201937561 Chr.11: 99956933-99956933 on Build GRCh38
rs112148988 Chr.11: 99956779-99956779 on Build GRCh38
rs537252996 Chr.11: 99956751-99956751 on Build GRCh38
rs35581997 Chr.11: 99956939-99956940 on Build GRCh38
rs892410607 Chr.11: 99957115-99957115 on Build GRCh38
rs746247673 Chr.11: 99956861-99956861 on Build GRCh38
rs754877840 Chr.11: 99956775-99956775 on Build GRCh38
rs201012221 Chr.11: 99956997-99956997 on Build GRCh38
rs201760493 Chr.11: 99957010-99957010 on Build GRCh38
rs771764286 Chr.11: 99957013-99957013 on Build GRCh38
rs752639831 Chr.11: 99956922-99956922 on Build GRCh38
rs779028608 Chr.11: 99957000-99957000 on Build GRCh38
rs758651484 Chr.11: 99957033-99957033 on Build GRCh38
rs201895126 Chr.11: 99956982-99956982 on Build GRCh38
rs967093176 Chr.11: 99956963-99956963 on Build GRCh38
rs201326352 Chr.11: 99956847-99956847 on Build GRCh38

View Full Product Details