Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 102321779-102322304 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIRC3
Entrez Gene ID
330
Gene Name
baculoviral IAP repeat containing 3
Gene Aliases
AIP1, API2, CIAP2, HAIP1, HIAP1, MALT2, MIHC, RNF49, c-IAP2
UniGene
Hs.127799
Gene Chromosome Location
Chr.11: 102317450-102339403 on Build GRCh38
Transcripts
NM_001165.4 NM_182962.2 XM_017017643.1

SNP Detail

SNP ID Chromosome Location
rs17885474 Chr.11: 102322034-102322034 on Build GRCh38
rs970544182 Chr.11: 102322232-102322232 on Build GRCh38
rs964906122 Chr.11: 102321959-102321959 on Build GRCh38
rs961853957 Chr.11: 102321844-102321844 on Build GRCh38
rs774268137 Chr.11: 102321937-102321937 on Build GRCh38
rs17878442 Chr.11: 102321843-102321843 on Build GRCh38
rs144117251 Chr.11: 102322099-102322099 on Build GRCh38
rs907899073 Chr.11: 102322033-102322033 on Build GRCh38
rs952675705 Chr.11: 102322055-102322055 on Build GRCh38
rs926322020 Chr.11: 102322244-102322244 on Build GRCh38
rs1042136702 Chr.11: 102322144-102322144 on Build GRCh38
rs1021042993 Chr.11: 102321900-102321900 on Build GRCh38
rs1019190191 Chr.11: 102321948-102321948 on Build GRCh38
rs1053952660 Chr.11: 102321995-102321995 on Build GRCh38
rs562056022 Chr.11: 102321856-102321856 on Build GRCh38
rs952236005 Chr.11: 102322265-102322265 on Build GRCh38
rs936959090 Chr.11: 102321989-102321989 on Build GRCh38
rs980923260 Chr.11: 102322240-102322240 on Build GRCh38
rs759236630 Chr.11: 102321986-102321986 on Build GRCh38
rs563952067 Chr.11: 102322220-102322220 on Build GRCh38
rs377068531 Chr.11: 102322010-102322010 on Build GRCh38
rs971867497 Chr.11: 102321854-102321854 on Build GRCh38
rs946492771 Chr.11: 102322131-102322131 on Build GRCh38
rs747748073 Chr.11: 102321839-102321839 on Build GRCh38
rs375905102 Chr.11: 102322198-102322198 on Build GRCh38
rs961926736 Chr.11: 102322176-102322176 on Build GRCh38
rs1028356626 Chr.11: 102322043-102322043 on Build GRCh38
rs11558755 Chr.11: 102321915-102321915 on Build GRCh38
rs763241396 Chr.11: 102322207-102322207 on Build GRCh38
rs572461468 Chr.11: 102321971-102321971 on Build GRCh38
rs878944899 Chr.11: 102322192-102322192 on Build GRCh38
rs926280355 Chr.11: 102321987-102321987 on Build GRCh38
rs1009193566 Chr.11: 102321928-102321928 on Build GRCh38
rs770752739 Chr.11: 102321932-102321932 on Build GRCh38
rs971930394 Chr.11: 102322181-102322181 on Build GRCh38
rs937899802 Chr.11: 102322084-102322084 on Build GRCh38
rs1016206818 Chr.11: 102322157-102322157 on Build GRCh38
rs997335392 Chr.11: 102322042-102322042 on Build GRCh38
rs900775847 Chr.11: 102322200-102322200 on Build GRCh38
rs1006165318 Chr.11: 102322089-102322089 on Build GRCh38
rs893600561 Chr.11: 102322121-102322121 on Build GRCh38
rs776841389 Chr.11: 102322247-102322247 on Build GRCh38
rs917687602 Chr.11: 102321903-102321903 on Build GRCh38
rs1018595185 Chr.11: 102322193-102322193 on Build GRCh38
rs1025190085 Chr.11: 102322216-102322216 on Build GRCh38
rs749261597 Chr.11: 102321901-102321901 on Build GRCh38
rs530207374 Chr.11: 102321855-102321855 on Build GRCh38
rs1041583894 Chr.11: 102322023-102322023 on Build GRCh38
rs17880156 Chr.11: 102322013-102322013 on Build GRCh38
rs1055386513 Chr.11: 102322108-102322108 on Build GRCh38
rs528920822 Chr.11: 102322246-102322246 on Build GRCh38

View Full Product Details