Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 104887075-104887574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASP12
Entrez Gene ID
100506742
Gene Name
caspase 12 (gene/pseudogene)
Gene Aliases
CASP-12, CASP12P1
UniGene
Hs.476989
Gene Chromosome Location
Chr.11: 104885718-104898670 on Build GRCh38
Transcripts
NM_001191016.2 NR_034061.2 NR_034063.2 NR_034064.2 NR_034065.2 NR_034066.2 NR_034067.2 NR_034068.2 NR_034070.2 NR_034071.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5920074 c.913G>A p.E305K Chr.11: 104887210-104887210 on Build GRCh38
2109029 c.827C>A p.S276Y Chr.11: 104887296-104887296 on Build GRCh38
3738831 c.823G>A p.V275I Chr.11: 104887300-104887300 on Build GRCh38
2109028 c.833G>T p.R278I Chr.11: 104887290-104887290 on Build GRCh38
2109025 c.910G>A p.E304K Chr.11: 104887213-104887213 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs955581237 Chr.11: 104887416-104887416 on Build GRCh38
rs966479682 Chr.11: 104887354-104887354 on Build GRCh38
rs1003926944 Chr.11: 104887463-104887463 on Build GRCh38
rs186169944 Chr.11: 104887274-104887274 on Build GRCh38
rs113334746 Chr.11: 104887537-104887537 on Build GRCh38
rs1003845910 Chr.11: 104887280-104887280 on Build GRCh38
rs190492754 Chr.11: 104887467-104887467 on Build GRCh38
rs952502775 Chr.11: 104887193-104887193 on Build GRCh38
rs5794352 Chr.11: 104887109-104887110 on Build GRCh38
rs869077153 Chr.11: 104887110-104887111 on Build GRCh38
rs484438 Chr.11: 104887308-104887308 on Build GRCh38
rs747097131 Chr.11: 104887282-104887282 on Build GRCh38
rs974076194 Chr.11: 104887483-104887483 on Build GRCh38
rs897281997 Chr.11: 104887207-104887207 on Build GRCh38
rs778426059 Chr.11: 104887328-104887328 on Build GRCh38
rs971304221 Chr.11: 104887223-104887223 on Build GRCh38
rs570925421 Chr.11: 104887536-104887536 on Build GRCh38
rs918656139 Chr.11: 104887490-104887490 on Build GRCh38
rs148869736 Chr.11: 104887540-104887540 on Build GRCh38
rs952491352 Chr.11: 104887435-104887435 on Build GRCh38
rs1042875734 Chr.11: 104887362-104887362 on Build GRCh38
rs200757065 Chr.11: 104887370-104887370 on Build GRCh38
rs563371877 Chr.11: 104887322-104887322 on Build GRCh38
rs899791012 Chr.11: 104887422-104887422 on Build GRCh38
rs1010616639 Chr.11: 104887327-104887327 on Build GRCh38
rs1033912231 Chr.11: 104887527-104887527 on Build GRCh38
rs908675376 Chr.11: 104887217-104887217 on Build GRCh38
rs911186819 Chr.11: 104887180-104887180 on Build GRCh38
rs181763844 Chr.11: 104887202-104887202 on Build GRCh38
rs189514213 Chr.11: 104887184-104887184 on Build GRCh38
rs142269030 Chr.11: 104887271-104887271 on Build GRCh38
rs963658881 Chr.11: 104887153-104887153 on Build GRCh38
rs199619566 Chr.11: 104887107-104887108 on Build GRCh38
rs147819564 Chr.11: 104887351-104887351 on Build GRCh38
rs56670368 Chr.11: 104887391-104887391 on Build GRCh38
rs548034856 Chr.11: 104887533-104887533 on Build GRCh38
rs941380897 Chr.11: 104887187-104887187 on Build GRCh38
rs146027803 Chr.11: 104887276-104887276 on Build GRCh38
rs748340970 Chr.11: 104887335-104887335 on Build GRCh38
rs772888343 Chr.11: 104887433-104887433 on Build GRCh38
rs996482425 Chr.11: 104887167-104887167 on Build GRCh38
rs978239663 Chr.11: 104887385-104887385 on Build GRCh38
rs1019493808 Chr.11: 104887151-104887151 on Build GRCh38
rs930085204 Chr.11: 104887532-104887532 on Build GRCh38
rs780191160 Chr.11: 104887173-104887173 on Build GRCh38
rs555290718 Chr.11: 104887179-104887179 on Build GRCh38
rs1015775708 Chr.11: 104887464-104887464 on Build GRCh38
rs147443677 Chr.11: 104887384-104887385 on Build GRCh38
rs12276274 Chr.11: 104887434-104887434 on Build GRCh38
rs778626129 Chr.11: 104887248-104887248 on Build GRCh38

View Full Product Details