Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107522180-107522717 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALKBH8
Entrez Gene ID
91801
Gene Name
alkB homolog 8, tRNA methyltransferase
Gene Aliases
ABH8, TRM9, TRMT9
UniGene
Hs.503763
Gene Chromosome Location
Chr.11: 107502726-107565740 on Build GRCh38
Transcripts
NM_001301010.1 NM_138775.2 XM_005271728.2 XM_005271729.3 XM_017018553.1 XM_017018554.1 XM_017018555.1 XM_017018556.1 XM_017018557.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6338886 c.1185G>A p.P395P Chr.11: 107522401-107522401 on Build GRCh38
922601 c.1146T>A p.I382I Chr.11: 107522440-107522440 on Build GRCh38
4845477 c.1253G>A p.G418E Chr.11: 107522333-107522333 on Build GRCh38
2109933 c.1043T>A p.V348D Chr.11: 107522543-107522543 on Build GRCh38
6338887 c.1185G>A p.P395P Chr.11: 107522401-107522401 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373004685 Chr.11: 107522304-107522304 on Build GRCh38
rs371431984 Chr.11: 107522367-107522367 on Build GRCh38
rs557574244 Chr.11: 107522357-107522357 on Build GRCh38
rs956000961 Chr.11: 107522345-107522345 on Build GRCh38
rs145855865 Chr.11: 107522637-107522637 on Build GRCh38
rs557502763 Chr.11: 107522678-107522678 on Build GRCh38
rs556827511 Chr.11: 107522215-107522215 on Build GRCh38
rs971095710 Chr.11: 107522311-107522311 on Build GRCh38
rs770811496 Chr.11: 107522456-107522456 on Build GRCh38
rs138767142 Chr.11: 107522679-107522679 on Build GRCh38
rs905850520 Chr.11: 107522247-107522247 on Build GRCh38
rs1038387744 Chr.11: 107522480-107522480 on Build GRCh38
rs556026632 Chr.11: 107522465-107522465 on Build GRCh38
rs775774880 Chr.11: 107522487-107522487 on Build GRCh38
rs1006857399 Chr.11: 107522231-107522231 on Build GRCh38
rs1002600420 Chr.11: 107522542-107522542 on Build GRCh38
rs907914367 Chr.11: 107522230-107522230 on Build GRCh38
rs183757750 Chr.11: 107522428-107522428 on Build GRCh38
rs764532634 Chr.11: 107522512-107522512 on Build GRCh38
rs777087425 Chr.11: 107522530-107522530 on Build GRCh38
rs757179773 Chr.11: 107522559-107522560 on Build GRCh38
rs906963620 Chr.11: 107522513-107522513 on Build GRCh38
rs747954438 Chr.11: 107522402-107522402 on Build GRCh38
rs923362235 Chr.11: 107522435-107522435 on Build GRCh38
rs1057132578 Chr.11: 107522602-107522602 on Build GRCh38
rs1037824508 Chr.11: 107522244-107522244 on Build GRCh38
rs768861805 Chr.11: 107522603-107522604 on Build GRCh38
rs778987140 Chr.11: 107522585-107522585 on Build GRCh38
rs762740092 Chr.11: 107522625-107522625 on Build GRCh38
rs914480436 Chr.11: 107522364-107522364 on Build GRCh38
rs537937283 Chr.11: 107522275-107522275 on Build GRCh38
rs568806408 Chr.11: 107522248-107522248 on Build GRCh38
rs907695606 Chr.11: 107522515-107522515 on Build GRCh38
rs188523089 Chr.11: 107522548-107522548 on Build GRCh38
rs775064272 Chr.11: 107522287-107522287 on Build GRCh38
rs753631187 Chr.11: 107522269-107522269 on Build GRCh38
rs749242418 Chr.11: 107522455-107522455 on Build GRCh38
rs964324969 Chr.11: 107522464-107522464 on Build GRCh38
rs760741899 Chr.11: 107522508-107522508 on Build GRCh38
rs745922825 Chr.11: 107522481-107522481 on Build GRCh38
rs948408312 Chr.11: 107522496-107522496 on Build GRCh38
rs895393777 Chr.11: 107522493-107522493 on Build GRCh38
rs373418821 Chr.11: 107522332-107522332 on Build GRCh38
rs904720798 Chr.11: 107522411-107522411 on Build GRCh38
rs1010781069 Chr.11: 107522395-107522395 on Build GRCh38
rs933376320 Chr.11: 107522450-107522450 on Build GRCh38
rs751166313 Chr.11: 107522596-107522596 on Build GRCh38
rs908014696 Chr.11: 107522463-107522463 on Build GRCh38
rs765666925 Chr.11: 107522331-107522331 on Build GRCh38
rs574270690 Chr.11: 107522482-107522482 on Build GRCh38
rs1031399735 Chr.11: 107522588-107522588 on Build GRCh38
rs568927883 Chr.11: 107522270-107522270 on Build GRCh38
rs754720775 Chr.11: 107522373-107522373 on Build GRCh38
rs777368983 Chr.11: 107522446-107522446 on Build GRCh38
rs891649222 Chr.11: 107522601-107522601 on Build GRCh38
rs768513222 Chr.11: 107522324-107522324 on Build GRCh38
rs377341673 Chr.11: 107522358-107522358 on Build GRCh38
rs192250667 Chr.11: 107522348-107522348 on Build GRCh38
rs371011851 Chr.11: 107522379-107522379 on Build GRCh38

View Full Product Details