Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107635549-107636052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD1
Entrez Gene ID
55531
Gene Name
ELMO domain containing 1
Gene Aliases
-
UniGene
Hs.495779
Gene Chromosome Location
Chr.11: 107591091-107666779 on Build GRCh38
Transcripts
NM_001130037.1 NM_001308018.1 NM_018712.3 XM_017017994.1 XM_017017995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
218293 c.328G>A p.V110I Chr.11: 107635673-107635673 on Build GRCh38
3443089 c.382G>A p.D128N Chr.11: 107635727-107635727 on Build GRCh38
6305400 c.405A>T p.E135D Chr.11: 107635750-107635750 on Build GRCh38
3443090 c.395C>T p.P132L Chr.11: 107635740-107635740 on Build GRCh38
4680802 c.367C>T p.R123C Chr.11: 107635712-107635712 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778757578 Chr.11: 107635720-107635720 on Build GRCh38
rs755713175 Chr.11: 107635605-107635605 on Build GRCh38
rs749621912 Chr.11: 107635764-107635765 on Build GRCh38
rs772308524 Chr.11: 107635632-107635632 on Build GRCh38
rs760759593 Chr.11: 107635639-107635639 on Build GRCh38
rs971381218 Chr.11: 107635915-107635915 on Build GRCh38
rs144076001 Chr.11: 107635832-107635832 on Build GRCh38
rs79251702 Chr.11: 107635782-107635782 on Build GRCh38
rs572733694 Chr.11: 107635846-107635846 on Build GRCh38
rs763263117 Chr.11: 107635684-107635684 on Build GRCh38
rs538131426 Chr.11: 107635712-107635712 on Build GRCh38
rs959370321 Chr.11: 107635866-107635866 on Build GRCh38
rs147335987 Chr.11: 107636007-107636007 on Build GRCh38
rs745637426 Chr.11: 107635737-107635737 on Build GRCh38
rs548183462 Chr.11: 107635667-107635667 on Build GRCh38
rs979580122 Chr.11: 107635988-107635988 on Build GRCh38
rs752311387 Chr.11: 107635698-107635698 on Build GRCh38
rs764176228 Chr.11: 107635655-107635655 on Build GRCh38
rs775607521 Chr.11: 107635638-107635638 on Build GRCh38
rs762305899 Chr.11: 107635674-107635674 on Build GRCh38
rs776681372 Chr.11: 107635765-107635765 on Build GRCh38
rs1032542635 Chr.11: 107635998-107635998 on Build GRCh38
rs1014005090 Chr.11: 107635900-107635900 on Build GRCh38
rs533751129 Chr.11: 107635869-107635869 on Build GRCh38
rs755627533 Chr.11: 107635700-107635700 on Build GRCh38
rs748589998 Chr.11: 107635605-107635606 on Build GRCh38
rs750774743 Chr.11: 107635683-107635683 on Build GRCh38
rs759848005 Chr.11: 107635606-107635606 on Build GRCh38
rs774281259 Chr.11: 107635628-107635628 on Build GRCh38
rs1033736950 Chr.11: 107635867-107635867 on Build GRCh38
rs377547455 Chr.11: 107635678-107635678 on Build GRCh38
rs866163360 Chr.11: 107635820-107635820 on Build GRCh38
rs766666964 Chr.11: 107635797-107635797 on Build GRCh38
rs767154171 Chr.11: 107635696-107635696 on Build GRCh38
rs921017276 Chr.11: 107636016-107636016 on Build GRCh38
rs752326535 Chr.11: 107635809-107635809 on Build GRCh38
rs967396267 Chr.11: 107635959-107635959 on Build GRCh38
rs773619382 Chr.11: 107635786-107635786 on Build GRCh38
rs747240188 Chr.11: 107635750-107635750 on Build GRCh38
rs768936625 Chr.11: 107635759-107635759 on Build GRCh38
rs918174602 Chr.11: 107635592-107635592 on Build GRCh38
rs753463613 Chr.11: 107635713-107635713 on Build GRCh38
rs935762912 Chr.11: 107635693-107635693 on Build GRCh38
rs966862481 Chr.11: 107635636-107635636 on Build GRCh38
rs757996724 Chr.11: 107635660-107635660 on Build GRCh38
rs770782129 Chr.11: 107635623-107635623 on Build GRCh38
rs991233540 Chr.11: 107635880-107635880 on Build GRCh38
rs771786761 Chr.11: 107635739-107635739 on Build GRCh38
rs7101759 Chr.11: 107635823-107635823 on Build GRCh38
rs760398972 Chr.11: 107635810-107635810 on Build GRCh38
rs751184468 Chr.11: 107635839-107635839 on Build GRCh38
rs555242017 Chr.11: 107635906-107635906 on Build GRCh38
rs761927187 Chr.11: 107635781-107635781 on Build GRCh38
rs934385606 Chr.11: 107635672-107635672 on Build GRCh38
rs749251046 Chr.11: 107635619-107635619 on Build GRCh38
rs11826462 Chr.11: 107635635-107635635 on Build GRCh38
rs202156688 Chr.11: 107635673-107635673 on Build GRCh38
rs780265801 Chr.11: 107635745-107635745 on Build GRCh38
rs978397428 Chr.11: 107635979-107635979 on Build GRCh38
rs943042896 Chr.11: 107635717-107635717 on Build GRCh38
rs978664312 Chr.11: 107635643-107635643 on Build GRCh38
rs571269110 Chr.11: 107635787-107635787 on Build GRCh38
rs1026096264 Chr.11: 107635796-107635796 on Build GRCh38
rs1025015825 Chr.11: 107635914-107635914 on Build GRCh38
rs200189303 Chr.11: 107635631-107635631 on Build GRCh38
rs924142080 Chr.11: 107635669-107635669 on Build GRCh38
rs756777381 Chr.11: 107635879-107635879 on Build GRCh38
rs949543706 Chr.11: 107635637-107635637 on Build GRCh38

View Full Product Details