Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 107650243-107650740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMOD1
Entrez Gene ID
55531
Gene Name
ELMO domain containing 1
Gene Aliases
-
UniGene
Hs.495779
Gene Chromosome Location
Chr.11: 107591091-107666779 on Build GRCh38
Transcripts
NM_001130037.1 NM_001308018.1 NM_018712.3 XM_017017994.1 XM_017017995.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594534 c.576C>T p.A192A Chr.11: 107650356-107650356 on Build GRCh38
6067471 c.555-2A>G p.? Chr.11: 107650333-107650333 on Build GRCh38
1166127 c.562G>A p.A188T Chr.11: 107650342-107650342 on Build GRCh38
6565278 c.555-7G>T p.? Chr.11: 107650328-107650328 on Build GRCh38
5453812 c.564G>A p.A188A Chr.11: 107650344-107650344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs986894606 Chr.11: 107650375-107650375 on Build GRCh38
rs74760388 Chr.11: 107650287-107650287 on Build GRCh38
rs35794825 Chr.11: 107650692-107650692 on Build GRCh38
rs938221882 Chr.11: 107650421-107650421 on Build GRCh38
rs533354427 Chr.11: 107650695-107650695 on Build GRCh38
rs1051689695 Chr.11: 107650468-107650468 on Build GRCh38
rs746382771 Chr.11: 107650372-107650372 on Build GRCh38
rs563123810 Chr.11: 107650602-107650602 on Build GRCh38
rs199532976 Chr.11: 107650394-107650394 on Build GRCh38
rs4753809 Chr.11: 107650554-107650554 on Build GRCh38
rs1028690830 Chr.11: 107650618-107650618 on Build GRCh38
rs867787369 Chr.11: 107650366-107650366 on Build GRCh38
rs568688091 Chr.11: 107650328-107650328 on Build GRCh38
rs759464252 Chr.11: 107650432-107650432 on Build GRCh38
rs200602594 Chr.11: 107650355-107650355 on Build GRCh38
rs12362443 Chr.11: 107650364-107650364 on Build GRCh38
rs574941339 Chr.11: 107650310-107650310 on Build GRCh38
rs902965135 Chr.11: 107650469-107650469 on Build GRCh38
rs1014788865 Chr.11: 107650543-107650543 on Build GRCh38
rs1015007353 Chr.11: 107650299-107650299 on Build GRCh38
rs570892039 Chr.11: 107650476-107650476 on Build GRCh38
rs368957163 Chr.11: 107650344-107650344 on Build GRCh38
rs562549325 Chr.11: 107650504-107650504 on Build GRCh38
rs1004932244 Chr.11: 107650288-107650288 on Build GRCh38
rs750164953 Chr.11: 107650383-107650384 on Build GRCh38
rs567441629 Chr.11: 107650275-107650275 on Build GRCh38
rs757380889 Chr.11: 107650350-107650350 on Build GRCh38
rs764825044 Chr.11: 107650306-107650311 on Build GRCh38
rs755112050 Chr.11: 107650601-107650601 on Build GRCh38
rs188228118 Chr.11: 107650463-107650463 on Build GRCh38
rs747460460 Chr.11: 107650391-107650391 on Build GRCh38
rs994943117 Chr.11: 107650589-107650589 on Build GRCh38
rs1045483384 Chr.11: 107650461-107650461 on Build GRCh38
rs1024020703 Chr.11: 107650593-107650593 on Build GRCh38
rs766724122 Chr.11: 107650276-107650276 on Build GRCh38
rs372554853 Chr.11: 107650327-107650327 on Build GRCh38
rs573619987 Chr.11: 107650387-107650387 on Build GRCh38
rs928213646 Chr.11: 107650606-107650606 on Build GRCh38
rs539871678 Chr.11: 107650424-107650424 on Build GRCh38
rs762567061 Chr.11: 107650317-107650317 on Build GRCh38
rs544855323 Chr.11: 107650683-107650683 on Build GRCh38
rs956982096 Chr.11: 107650617-107650617 on Build GRCh38
rs779196499 Chr.11: 107650610-107650610 on Build GRCh38
rs201176533 Chr.11: 107650341-107650341 on Build GRCh38
rs752553992 Chr.11: 107650342-107650342 on Build GRCh38
rs775179576 Chr.11: 107650289-107650289 on Build GRCh38
rs765001932 Chr.11: 107650309-107650309 on Build GRCh38
rs754705721 Chr.11: 107650337-107650337 on Build GRCh38
rs947164286 Chr.11: 107650422-107650422 on Build GRCh38
rs915431474 Chr.11: 107650696-107650696 on Build GRCh38
rs767500622 Chr.11: 107650453-107650453 on Build GRCh38
rs868773951 Chr.11: 107650385-107650385 on Build GRCh38
rs772952763 Chr.11: 107650395-107650395 on Build GRCh38
rs767548312 Chr.11: 107650400-107650400 on Build GRCh38
rs374759236 Chr.11: 107650425-107650425 on Build GRCh38
rs751074571 Chr.11: 107650351-107650351 on Build GRCh38
rs763943002 Chr.11: 107650343-107650343 on Build GRCh38
rs772773616 Chr.11: 107650379-107650379 on Build GRCh38
rs762563995 Chr.11: 107650294-107650294 on Build GRCh38
rs988252127 Chr.11: 107650666-107650666 on Build GRCh38
rs749295217 Chr.11: 107650347-107650347 on Build GRCh38

View Full Product Details