Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108052654-108053163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL5
Entrez Gene ID
8065
Gene Name
cullin 5
Gene Aliases
VACM-1, VACM1
UniGene
Hs.440320
Gene Chromosome Location
Chr.11: 108008682-108107766 on Build GRCh38
Transcripts
NM_003478.3 XM_005271682.2 XM_011543013.1 XM_017018363.1 XM_017018364.1 XM_017018365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1604028 c.421G>A p.D141N Chr.11: 108052669-108052669 on Build GRCh38
2110132 c.473G>T p.S158I Chr.11: 108052721-108052721 on Build GRCh38
3375591 c.500G>C p.R167T Chr.11: 108052748-108052748 on Build GRCh38
5688398 c.500G>T p.R167I Chr.11: 108052748-108052748 on Build GRCh38
4891523 c.458A>G p.N153S Chr.11: 108052706-108052706 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771559839 Chr.11: 108052760-108052760 on Build GRCh38
rs912441151 Chr.11: 108053054-108053054 on Build GRCh38
rs770316737 Chr.11: 108052721-108052721 on Build GRCh38
rs773514285 Chr.11: 108052726-108052726 on Build GRCh38
rs544845863 Chr.11: 108053074-108053074 on Build GRCh38
rs557429040 Chr.11: 108052910-108052910 on Build GRCh38
rs149575444 Chr.11: 108052776-108052776 on Build GRCh38
rs4453201 Chr.11: 108053031-108053031 on Build GRCh38
rs539289117 Chr.11: 108052909-108052909 on Build GRCh38
rs547685418 Chr.11: 108053140-108053140 on Build GRCh38
rs988781972 Chr.11: 108052968-108052968 on Build GRCh38
rs144188707 Chr.11: 108052904-108052904 on Build GRCh38
rs947277375 Chr.11: 108053129-108053129 on Build GRCh38
rs571288383 Chr.11: 108053025-108053025 on Build GRCh38
rs755193857 Chr.11: 108052830-108052830 on Build GRCh38
rs761804083 Chr.11: 108052817-108052817 on Build GRCh38
rs1050098256 Chr.11: 108052687-108052687 on Build GRCh38
rs1027076818 Chr.11: 108052860-108052860 on Build GRCh38
rs779768714 Chr.11: 108052824-108052824 on Build GRCh38
rs866216242 Chr.11: 108052981-108052981 on Build GRCh38
rs1023615802 Chr.11: 108053027-108053027 on Build GRCh38
rs994386446 Chr.11: 108052978-108052978 on Build GRCh38
rs182241012 Chr.11: 108053021-108053021 on Build GRCh38
rs943978034 Chr.11: 108053055-108053055 on Build GRCh38
rs985333243 Chr.11: 108053046-108053046 on Build GRCh38
rs938534709 Chr.11: 108052804-108052804 on Build GRCh38
rs765542200 Chr.11: 108052699-108052700 on Build GRCh38
rs1036670726 Chr.11: 108052751-108052751 on Build GRCh38
rs1036619762 Chr.11: 108053067-108053067 on Build GRCh38
rs555311702 Chr.11: 108053023-108053023 on Build GRCh38
rs931015185 Chr.11: 108053082-108053082 on Build GRCh38
rs768493085 Chr.11: 108052822-108052822 on Build GRCh38
rs531628007 Chr.11: 108052965-108052965 on Build GRCh38
rs758204829 Chr.11: 108052823-108052823 on Build GRCh38
rs781175345 Chr.11: 108052706-108052706 on Build GRCh38
rs186349446 Chr.11: 108052767-108052767 on Build GRCh38
rs943690818 Chr.11: 108052718-108052718 on Build GRCh38
rs113729240 Chr.11: 108053102-108053102 on Build GRCh38
rs1036335960 Chr.11: 108052934-108052934 on Build GRCh38
rs751781080 Chr.11: 108052825-108052825 on Build GRCh38
rs749228387 Chr.11: 108052734-108052734 on Build GRCh38
rs528015818 Chr.11: 108053091-108053091 on Build GRCh38
rs1048615988 Chr.11: 108053115-108053115 on Build GRCh38
rs765292447 Chr.11: 108052822-108052822 on Build GRCh38
rs866833026 Chr.11: 108053008-108053008 on Build GRCh38
rs567566270 Chr.11: 108052710-108052710 on Build GRCh38
rs868850662 Chr.11: 108053020-108053020 on Build GRCh38
rs988630802 Chr.11: 108053076-108053076 on Build GRCh38
rs901304627 Chr.11: 108052966-108052966 on Build GRCh38
rs891756165 Chr.11: 108052889-108052889 on Build GRCh38
rs867861963 Chr.11: 108052685-108052685 on Build GRCh38
rs189587327 Chr.11: 108052945-108052945 on Build GRCh38
rs1007029312 Chr.11: 108052925-108052925 on Build GRCh38

View Full Product Details