Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108054527-108055029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUL5
Entrez Gene ID
8065
Gene Name
cullin 5
Gene Aliases
VACM-1, VACM1
UniGene
Hs.440320
Gene Chromosome Location
Chr.11: 108008682-108107766 on Build GRCh38
Transcripts
NM_003478.3 XM_005271682.2 XM_011543013.1 XM_017018363.1 XM_017018364.1 XM_017018365.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1585375 c.758G>A p.R253Q Chr.11: 108054933-108054933 on Build GRCh38
6189599 c.633G>C p.E211D Chr.11: 108054726-108054726 on Build GRCh38
6531297 c.731G>C p.R244P Chr.11: 108054906-108054906 on Build GRCh38
4017578 c.729A>T p.K243N Chr.11: 108054904-108054904 on Build GRCh38
5999753 c.746T>C p.L249S Chr.11: 108054921-108054921 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs567468310 Chr.11: 108054705-108054705 on Build GRCh38
rs755176411 Chr.11: 108054708-108054708 on Build GRCh38
rs771594093 Chr.11: 108054855-108054855 on Build GRCh38
rs777288052 Chr.11: 108054953-108054953 on Build GRCh38
rs751341507 Chr.11: 108054707-108054707 on Build GRCh38
rs749782953 Chr.11: 108054761-108054761 on Build GRCh38
rs758804397 Chr.11: 108054801-108054802 on Build GRCh38
rs772886015 Chr.11: 108054816-108054816 on Build GRCh38
rs756378846 Chr.11: 108054861-108054861 on Build GRCh38
rs774137907 Chr.11: 108054833-108054833 on Build GRCh38
rs762844081 Chr.11: 108054693-108054693 on Build GRCh38
rs769598770 Chr.11: 108054947-108054947 on Build GRCh38
rs2271852 Chr.11: 108054576-108054576 on Build GRCh38
rs768301255 Chr.11: 108054772-108054772 on Build GRCh38
rs748759725 Chr.11: 108054975-108054975 on Build GRCh38
rs759577436 Chr.11: 108054992-108054992 on Build GRCh38
rs77669929 Chr.11: 108054814-108054814 on Build GRCh38
rs200802906 Chr.11: 108054623-108054623 on Build GRCh38
rs566273598 Chr.11: 108054837-108054837 on Build GRCh38
rs142787597 Chr.11: 108054919-108054919 on Build GRCh38
rs147410964 Chr.11: 108054946-108054946 on Build GRCh38
rs770834911 Chr.11: 108054982-108054982 on Build GRCh38
rs11212499 Chr.11: 108054566-108054566 on Build GRCh38
rs372484238 Chr.11: 108054621-108054621 on Build GRCh38
rs769402039 Chr.11: 108054815-108054815 on Build GRCh38
rs774150726 Chr.11: 108054983-108054983 on Build GRCh38
rs536837281 Chr.11: 108054841-108054841 on Build GRCh38
rs767739701 Chr.11: 108054712-108054712 on Build GRCh38
rs7125285 Chr.11: 108055002-108055002 on Build GRCh38
rs991939318 Chr.11: 108054634-108054634 on Build GRCh38
rs773265157 Chr.11: 108054675-108054675 on Build GRCh38
rs768352677 Chr.11: 108054820-108054820 on Build GRCh38
rs770226989 Chr.11: 108054601-108054601 on Build GRCh38
rs751985714 Chr.11: 108054954-108054955 on Build GRCh38
rs938110436 Chr.11: 108054857-108054857 on Build GRCh38
rs549904533 Chr.11: 108054768-108054768 on Build GRCh38
rs371804055 Chr.11: 108054618-108054618 on Build GRCh38
rs199562808 Chr.11: 108054858-108054858 on Build GRCh38
rs746180978 Chr.11: 108054927-108054927 on Build GRCh38
rs757736723 Chr.11: 108054914-108054914 on Build GRCh38
rs753897966 Chr.11: 108054889-108054889 on Build GRCh38
rs1032749848 Chr.11: 108054616-108054616 on Build GRCh38
rs3847562 Chr.11: 108054600-108054600 on Build GRCh38
rs183961591 Chr.11: 108054933-108054933 on Build GRCh38
rs2271853 Chr.11: 108054830-108054830 on Build GRCh38
rs768226352 Chr.11: 108054630-108054630 on Build GRCh38
rs776431315 Chr.11: 108054796-108054796 on Build GRCh38
rs531278262 Chr.11: 108054750-108054750 on Build GRCh38
rs939870045 Chr.11: 108054963-108054963 on Build GRCh38
rs1004096560 Chr.11: 108054588-108054588 on Build GRCh38
rs750738840 Chr.11: 108054625-108054626 on Build GRCh38
rs138939451 Chr.11: 108054753-108054753 on Build GRCh38
rs879170357 Chr.11: 108054632-108054632 on Build GRCh38
rs1050889767 Chr.11: 108054563-108054565 on Build GRCh38
rs780610258 Chr.11: 108054937-108054937 on Build GRCh38
rs752706904 Chr.11: 108054748-108054748 on Build GRCh38
rs966857348 Chr.11: 108054663-108054663 on Build GRCh38
rs778258178 Chr.11: 108054605-108054605 on Build GRCh38
rs771833226 Chr.11: 108055001-108055001 on Build GRCh38
rs771315306 Chr.11: 108054619-108054619 on Build GRCh38
rs146877430 Chr.11: 108054611-108054611 on Build GRCh38
rs776236248 Chr.11: 108054656-108054656 on Build GRCh38
rs903130235 Chr.11: 108054977-108054977 on Build GRCh38
rs764129188 Chr.11: 108054880-108054880 on Build GRCh38
rs369821434 Chr.11: 108054854-108054854 on Build GRCh38
rs538677426 Chr.11: 108054829-108054829 on Build GRCh38
rs914032445 Chr.11: 108054640-108054640 on Build GRCh38
rs200991955 Chr.11: 108054600-108054605 on Build GRCh38
rs372301056 Chr.11: 108054762-108054762 on Build GRCh38
rs764904082 Chr.11: 108054664-108054664 on Build GRCh38
rs766898170 Chr.11: 108054859-108054859 on Build GRCh38
rs1049076804 Chr.11: 108054849-108054849 on Build GRCh38

View Full Product Details