Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108227353-108227895 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATM
Entrez Gene ID
472
Gene Name
ATM serine/threonine kinase
Gene Aliases
AT1, ATA, ATC, ATD, ATDC, ATE, TEL1, TELO1
UniGene
Hs.367437
Gene Chromosome Location
Chr.11: 108222484-108369102 on Build GRCh38
Transcripts
NM_000051.3 XM_005271561.4 XM_005271562.4 XM_006718843.3 XM_011542840.2 XM_011542842.2 XM_011542843.2 XM_011542844.2 XM_017017789.1 XM_017017790.1 XM_017017791.1 XM_017017792.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
370728 c.170G>T p.W57L Chr.11: 108227873-108227873 on Build GRCh38
21316 c.68G>A p.R23Q Chr.11: 108227692-108227692 on Build GRCh38
5574415 c.1_19del19 p.? Chr.11: 108227625-108227643 on Build GRCh38
7325226 c.125A>G p.H42R Chr.11: 108227828-108227828 on Build GRCh38
6972760 c.121A>T p.K41* Chr.11: 108227824-108227824 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776457787 Chr.11: 108227715-108227715 on Build GRCh38
rs148061139 Chr.11: 108227797-108227797 on Build GRCh38
rs770834907 Chr.11: 108227841-108227841 on Build GRCh38
rs786204088 Chr.11: 108227697-108227697 on Build GRCh38
rs772591447 Chr.11: 108227830-108227830 on Build GRCh38
rs876658520 Chr.11: 108227792-108227792 on Build GRCh38
rs876659078 Chr.11: 108227860-108227860 on Build GRCh38
rs182382127 Chr.11: 108227542-108227542 on Build GRCh38
rs141376974 Chr.11: 108227540-108227540 on Build GRCh38
rs970407823 Chr.11: 108227838-108227838 on Build GRCh38
rs4987897 Chr.11: 108227415-108227415 on Build GRCh38
rs771887195 Chr.11: 108227667-108227667 on Build GRCh38
rs869312754 Chr.11: 108227594-108227594 on Build GRCh38
rs786203888 Chr.11: 108227854-108227854 on Build GRCh38
rs199853729 Chr.11: 108227687-108227687 on Build GRCh38
rs730881361 Chr.11: 108227779-108227779 on Build GRCh38
rs199854180 Chr.11: 108227734-108227734 on Build GRCh38
rs1047093670 Chr.11: 108227498-108227498 on Build GRCh38
rs3218684 Chr.11: 108227836-108227836 on Build GRCh38
rs879254052 Chr.11: 108227632-108227632 on Build GRCh38
rs965882083 Chr.11: 108227472-108227472 on Build GRCh38
rs890413530 Chr.11: 108227601-108227601 on Build GRCh38
rs201773026 Chr.11: 108227828-108227828 on Build GRCh38
rs775561876 Chr.11: 108227674-108227675 on Build GRCh38
rs751378494 Chr.11: 108227617-108227617 on Build GRCh38
rs746235533 Chr.11: 108227691-108227691 on Build GRCh38
rs766166610 Chr.11: 108227611-108227611 on Build GRCh38
rs7112053 Chr.11: 108227631-108227631 on Build GRCh38
rs778611534 Chr.11: 108227579-108227580 on Build GRCh38
rs547110435 Chr.11: 108227737-108227737 on Build GRCh38
rs863224583 Chr.11: 108227796-108227796 on Build GRCh38
rs774768437 Chr.11: 108227672-108227672 on Build GRCh38
rs2066734 Chr.11: 108227733-108227734 on Build GRCh38
rs876659116 Chr.11: 108227822-108227825 on Build GRCh38
rs745673370 Chr.11: 108227582-108227583 on Build GRCh38
rs768359847 Chr.11: 108227701-108227701 on Build GRCh38
rs1025659147 Chr.11: 108227525-108227526 on Build GRCh38
rs786203606 Chr.11: 108227626-108227626 on Build GRCh38
rs780616292 Chr.11: 108227832-108227832 on Build GRCh38
rs587779858 Chr.11: 108227692-108227692 on Build GRCh38
rs876660725 Chr.11: 108227859-108227859 on Build GRCh38
rs907531304 Chr.11: 108227386-108227386 on Build GRCh38
rs786201365 Chr.11: 108227636-108227636 on Build GRCh38
rs1039736256 Chr.11: 108227544-108227544 on Build GRCh38
rs1040197895 Chr.11: 108227870-108227870 on Build GRCh38
rs200744434 Chr.11: 108227591-108227591 on Build GRCh38
rs995984668 Chr.11: 108227573-108227573 on Build GRCh38
rs876660842 Chr.11: 108227639-108227640 on Build GRCh38
rs876659088 Chr.11: 108227642-108227642 on Build GRCh38
rs730881359 Chr.11: 108227625-108227625 on Build GRCh38
rs3218693 Chr.11: 108227516-108227516 on Build GRCh38
rs150143957 Chr.11: 108227834-108227834 on Build GRCh38
rs755213267 Chr.11: 108227623-108227623 on Build GRCh38
rs55861249 Chr.11: 108227806-108227806 on Build GRCh38
rs778201041 Chr.11: 108227662-108227662 on Build GRCh38
rs549786728 Chr.11: 108227578-108227578 on Build GRCh38
rs1052417994 Chr.11: 108227532-108227532 on Build GRCh38
rs754770960 Chr.11: 108227782-108227782 on Build GRCh38
rs147009251 Chr.11: 108227789-108227789 on Build GRCh38
rs768832884 Chr.11: 108227751-108227753 on Build GRCh38
rs755116080 Chr.11: 108227491-108227491 on Build GRCh38
rs876660671 Chr.11: 108227652-108227652 on Build GRCh38
rs779297339 Chr.11: 108227818-108227818 on Build GRCh38
rs587781661 Chr.11: 108227829-108227829 on Build GRCh38
rs890075956 Chr.11: 108227606-108227606 on Build GRCh38
rs56326800 Chr.11: 108227539-108227539 on Build GRCh38
rs754136203 Chr.11: 108227430-108227430 on Build GRCh38
rs1052875923 Chr.11: 108227552-108227557 on Build GRCh38
rs766284348 Chr.11: 108227762-108227762 on Build GRCh38
rs3218703 Chr.11: 108227546-108227546 on Build GRCh38
rs766396464 Chr.11: 108227732-108227733 on Build GRCh38
rs928307261 Chr.11: 108227550-108227550 on Build GRCh38
rs751310537 Chr.11: 108227777-108227777 on Build GRCh38
rs876659324 Chr.11: 108227807-108227807 on Build GRCh38
rs781404312 Chr.11: 108227627-108227627 on Build GRCh38
rs776227873 Chr.11: 108227580-108227580 on Build GRCh38
rs948216400 Chr.11: 108227438-108227438 on Build GRCh38
rs1800054 Chr.11: 108227849-108227849 on Build GRCh38
rs786203926 Chr.11: 108227678-108227678 on Build GRCh38
rs786203370 Chr.11: 108227841-108227844 on Build GRCh38
rs730881362 Chr.11: 108227857-108227857 on Build GRCh38
rs748158168 Chr.11: 108227630-108227630 on Build GRCh38
rs761358619 Chr.11: 108227718-108227718 on Build GRCh38
rs876658161 Chr.11: 108227673-108227673 on Build GRCh38
rs973130115 Chr.11: 108227416-108227416 on Build GRCh38
rs1025690128 Chr.11: 108227751-108227751 on Build GRCh38
rs771378101 Chr.11: 108227666-108227666 on Build GRCh38
rs890010215 Chr.11: 108227402-108227402 on Build GRCh38
rs786202953 Chr.11: 108227781-108227781 on Build GRCh38
rs762382111 Chr.11: 108227837-108227837 on Build GRCh38
rs762943692 Chr.11: 108227744-108227744 on Build GRCh38
rs765229257 Chr.11: 108227603-108227603 on Build GRCh38
rs763900742 Chr.11: 108227576-108227576 on Build GRCh38
rs876659304 Chr.11: 108227693-108227693 on Build GRCh38
rs374303671 Chr.11: 108227607-108227607 on Build GRCh38
rs749776879 Chr.11: 108227665-108227665 on Build GRCh38
rs935580590 Chr.11: 108227530-108227530 on Build GRCh38
rs761415893 Chr.11: 108227596-108227596 on Build GRCh38
rs876660033 Chr.11: 108227850-108227850 on Build GRCh38
rs3218690 Chr.11: 108227865-108227865 on Build GRCh38
rs56096834 Chr.11: 108227551-108227551 on Build GRCh38
rs730881360 Chr.11: 108227629-108227629 on Build GRCh38
rs774185390 Chr.11: 108227843-108227843 on Build GRCh38
rs368161489 Chr.11: 108227798-108227798 on Build GRCh38
rs141586345 Chr.11: 108227661-108227661 on Build GRCh38
rs893791140 Chr.11: 108227537-108227537 on Build GRCh38

View Full Product Details