Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108511026-108511529 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
UniGene
Hs.28540
Gene Chromosome Location
Chr.11: 108505431-108607514 on Build GRCh38
Transcripts
NM_001144763.1 NM_001144764.1 NM_001144765.1 NM_001308019.1 NM_015065.2 XM_011542696.2 XM_011542698.2 XM_011542700.2 XM_017017397.1 XM_017017398.1 XM_017017399.1 XM_017017400.1 XM_017017401.1 XM_017017402.1 XM_017017403.1 XM_017017404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2110898 c.4046C>A p.S1349Y Chr.11: 108511461-108511461 on Build GRCh38
5371610 c.4388C>T p.P1463L Chr.11: 108511119-108511119 on Build GRCh38
4017678 c.4166T>C p.L1389S Chr.11: 108511341-108511341 on Build GRCh38
1251602 c.4232A>G p.E1411G Chr.11: 108511275-108511275 on Build GRCh38
7280224 c.4070C>A p.P1357H Chr.11: 108511437-108511437 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755312889 Chr.11: 108511126-108511126 on Build GRCh38
rs376431745 Chr.11: 108511328-108511328 on Build GRCh38
rs913142127 Chr.11: 108511417-108511417 on Build GRCh38
rs752867229 Chr.11: 108511141-108511141 on Build GRCh38
rs768150921 Chr.11: 108511066-108511066 on Build GRCh38
rs200727347 Chr.11: 108511474-108511474 on Build GRCh38
rs1029977662 Chr.11: 108511196-108511196 on Build GRCh38
rs954372095 Chr.11: 108511202-108511202 on Build GRCh38
rs757762764 Chr.11: 108511317-108511317 on Build GRCh38
rs774464911 Chr.11: 108511397-108511397 on Build GRCh38
rs750351501 Chr.11: 108511104-108511104 on Build GRCh38
rs955792201 Chr.11: 108511398-108511398 on Build GRCh38
rs754611874 Chr.11: 108511179-108511181 on Build GRCh38
rs759792392 Chr.11: 108511080-108511080 on Build GRCh38
rs780464588 Chr.11: 108511478-108511478 on Build GRCh38
rs769343747 Chr.11: 108511349-108511349 on Build GRCh38
rs773903413 Chr.11: 108511242-108511242 on Build GRCh38
rs541249387 Chr.11: 108511432-108511432 on Build GRCh38
rs115484033 Chr.11: 108511485-108511485 on Build GRCh38
rs765692103 Chr.11: 108511470-108511470 on Build GRCh38
rs955760001 Chr.11: 108511050-108511050 on Build GRCh38
rs995104545 Chr.11: 108511499-108511499 on Build GRCh38
rs557351623 Chr.11: 108511122-108511122 on Build GRCh38
rs74725820 Chr.11: 108511435-108511435 on Build GRCh38
rs141172581 Chr.11: 108511171-108511171 on Build GRCh38
rs376266268 Chr.11: 108511229-108511229 on Build GRCh38
rs373291968 Chr.11: 108511325-108511325 on Build GRCh38
rs762868552 Chr.11: 108511236-108511236 on Build GRCh38
rs768345129 Chr.11: 108511221-108511221 on Build GRCh38
rs767159879 Chr.11: 108511293-108511293 on Build GRCh38
rs973974276 Chr.11: 108511253-108511253 on Build GRCh38
rs759352392 Chr.11: 108511429-108511429 on Build GRCh38
rs774340091 Chr.11: 108511274-108511274 on Build GRCh38
rs755955627 Chr.11: 108511500-108511500 on Build GRCh38
rs199636293 Chr.11: 108511292-108511292 on Build GRCh38
rs756175919 Chr.11: 108511143-108511143 on Build GRCh38
rs765181920 Chr.11: 108511093-108511093 on Build GRCh38
rs143775381 Chr.11: 108511494-108511494 on Build GRCh38
rs995940329 Chr.11: 108511180-108511180 on Build GRCh38
rs776174830 Chr.11: 108511073-108511073 on Build GRCh38
rs764220052 Chr.11: 108511449-108511449 on Build GRCh38
rs1052424792 Chr.11: 108511368-108511368 on Build GRCh38
rs969838832 Chr.11: 108511286-108511286 on Build GRCh38
rs989838172 Chr.11: 108511090-108511090 on Build GRCh38
rs1021492037 Chr.11: 108511145-108511145 on Build GRCh38
rs565043078 Chr.11: 108511373-108511373 on Build GRCh38
rs865894870 Chr.11: 108511285-108511285 on Build GRCh38
rs760324571 Chr.11: 108511056-108511056 on Build GRCh38
rs746651616 Chr.11: 108511205-108511205 on Build GRCh38
rs1002734861 Chr.11: 108511337-108511337 on Build GRCh38
rs141714483 Chr.11: 108511491-108511491 on Build GRCh38
rs963933501 Chr.11: 108511495-108511495 on Build GRCh38
rs866115440 Chr.11: 108511410-108511410 on Build GRCh38
rs775711259 Chr.11: 108511441-108511441 on Build GRCh38
rs34978242 Chr.11: 108511480-108511480 on Build GRCh38
rs779225336 Chr.11: 108511320-108511320 on Build GRCh38
rs765584375 Chr.11: 108511240-108511240 on Build GRCh38
rs542108290 Chr.11: 108511194-108511194 on Build GRCh38
rs895230628 Chr.11: 108511502-108511502 on Build GRCh38
rs776086890 Chr.11: 108511222-108511222 on Build GRCh38
rs186051270 Chr.11: 108511385-108511385 on Build GRCh38
rs369244769 Chr.11: 108511277-108511277 on Build GRCh38
rs771257746 Chr.11: 108511178-108511178 on Build GRCh38
rs749933244 Chr.11: 108511310-108511310 on Build GRCh38
rs190476119 Chr.11: 108511447-108511447 on Build GRCh38
rs771035473 Chr.11: 108511390-108511390 on Build GRCh38
rs750943563 Chr.11: 108511471-108511471 on Build GRCh38
rs912838927 Chr.11: 108511455-108511455 on Build GRCh38
rs375346460 Chr.11: 108511465-108511465 on Build GRCh38
rs900349346 Chr.11: 108511475-108511475 on Build GRCh38
rs753914140 Chr.11: 108511452-108511452 on Build GRCh38
rs200608516 Chr.11: 108511492-108511492 on Build GRCh38
rs369877753 Chr.11: 108511323-108511323 on Build GRCh38
rs766268403 Chr.11: 108511259-108511259 on Build GRCh38
rs778291825 Chr.11: 108511155-108511155 on Build GRCh38
rs745803304 Chr.11: 108511496-108511496 on Build GRCh38
rs747910610 Chr.11: 108511356-108511357 on Build GRCh38
rs762751070 Chr.11: 108511116-108511116 on Build GRCh38
rs781401387 Chr.11: 108511139-108511139 on Build GRCh38
rs868078653 Chr.11: 108511357-108511357 on Build GRCh38
rs758785641 Chr.11: 108511324-108511324 on Build GRCh38
rs376155497 Chr.11: 108511315-108511315 on Build GRCh38
rs747761481 Chr.11: 108511334-108511334 on Build GRCh38
rs773136704 Chr.11: 108511230-108511230 on Build GRCh38
rs146870938 Chr.11: 108511288-108511288 on Build GRCh38
rs201753150 Chr.11: 108511289-108511289 on Build GRCh38
rs780889832 Chr.11: 108511305-108511305 on Build GRCh38
rs753724545 Chr.11: 108511490-108511490 on Build GRCh38
rs755775962 Chr.11: 108511482-108511482 on Build GRCh38
rs779315623 Chr.11: 108511200-108511200 on Build GRCh38
rs74858839 Chr.11: 108511118-108511118 on Build GRCh38
rs775062364 Chr.11: 108511049-108511049 on Build GRCh38

View Full Product Details