Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110435751-110436253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDX1
Entrez Gene ID
2230
Gene Name
ferredoxin 1
Gene Aliases
ADX, FDX, LOH11CR1D
UniGene
Hs.744
Gene Chromosome Location
Chr.11: 110429937-110464884 on Build GRCh38
Transcripts
NM_004109.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5749493 c.280G>T p.V94F Chr.11: 110435928-110435928 on Build GRCh38
1206963 c.283G>T p.E95* Chr.11: 110435931-110435931 on Build GRCh38
7178550 c.188C>T p.S63L Chr.11: 110435836-110435836 on Build GRCh38
4988586 c.239C>A p.T80K Chr.11: 110435887-110435887 on Build GRCh38
3738857 c.287A>G p.N96S Chr.11: 110435935-110435935 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs964498024 Chr.11: 110436057-110436057 on Build GRCh38
rs1017386438 Chr.11: 110435816-110435817 on Build GRCh38
rs896152656 Chr.11: 110435952-110435952 on Build GRCh38
rs909203388 Chr.11: 110436124-110436124 on Build GRCh38
rs960062093 Chr.11: 110436063-110436063 on Build GRCh38
rs773338470 Chr.11: 110435829-110435829 on Build GRCh38
rs775772933 Chr.11: 110435998-110435998 on Build GRCh38
rs139446247 Chr.11: 110435902-110435902 on Build GRCh38
rs777832214 Chr.11: 110436225-110436225 on Build GRCh38
rs528785756 Chr.11: 110436076-110436076 on Build GRCh38
rs4754454 Chr.11: 110436213-110436213 on Build GRCh38
rs780150621 Chr.11: 110435794-110435794 on Build GRCh38
rs771019824 Chr.11: 110435965-110435965 on Build GRCh38
rs909264831 Chr.11: 110435858-110435858 on Build GRCh38
rs757253507 Chr.11: 110435820-110435821 on Build GRCh38
rs768532188 Chr.11: 110435847-110435847 on Build GRCh38
rs746484343 Chr.11: 110436024-110436025 on Build GRCh38
rs144228166 Chr.11: 110435951-110435951 on Build GRCh38
rs764822594 Chr.11: 110435872-110435872 on Build GRCh38
rs772711311 Chr.11: 110436058-110436058 on Build GRCh38
rs754755113 Chr.11: 110435817-110435817 on Build GRCh38
rs978706269 Chr.11: 110435810-110435810 on Build GRCh38
rs781457182 Chr.11: 110435819-110435819 on Build GRCh38
rs759894054 Chr.11: 110435855-110435855 on Build GRCh38
rs369911593 Chr.11: 110435986-110435986 on Build GRCh38
rs773023611 Chr.11: 110436009-110436009 on Build GRCh38
rs1001236559 Chr.11: 110436051-110436051 on Build GRCh38
rs568418824 Chr.11: 110436130-110436130 on Build GRCh38
rs769913106 Chr.11: 110435825-110435825 on Build GRCh38
rs765304776 Chr.11: 110435821-110435821 on Build GRCh38
rs191114093 Chr.11: 110436018-110436018 on Build GRCh38
rs927462796 Chr.11: 110436215-110436215 on Build GRCh38
rs35520653 Chr.11: 110436025-110436026 on Build GRCh38
rs142079713 Chr.11: 110436094-110436094 on Build GRCh38
rs375254527 Chr.11: 110435887-110435887 on Build GRCh38
rs761111999 Chr.11: 110436001-110436001 on Build GRCh38
rs771424853 Chr.11: 110436037-110436037 on Build GRCh38
rs746402720 Chr.11: 110435995-110435995 on Build GRCh38
rs185852628 Chr.11: 110435867-110435867 on Build GRCh38
rs953344244 Chr.11: 110436222-110436222 on Build GRCh38
rs774868154 Chr.11: 110435851-110435851 on Build GRCh38
rs756272504 Chr.11: 110435944-110435944 on Build GRCh38
rs375998812 Chr.11: 110435922-110435922 on Build GRCh38
rs529266216 Chr.11: 110436123-110436123 on Build GRCh38
rs1020552388 Chr.11: 110436139-110436139 on Build GRCh38
rs925913685 Chr.11: 110436221-110436221 on Build GRCh38
rs4754453 Chr.11: 110436003-110436003 on Build GRCh38
rs150960595 Chr.11: 110435891-110435891 on Build GRCh38
rs1014065343 Chr.11: 110436064-110436064 on Build GRCh38
rs748391161 Chr.11: 110436052-110436052 on Build GRCh38
rs950787181 Chr.11: 110436120-110436121 on Build GRCh38
rs749385109 Chr.11: 110435833-110435833 on Build GRCh38
rs547010438 Chr.11: 110436079-110436079 on Build GRCh38
rs780919632 Chr.11: 110435919-110435919 on Build GRCh38
rs967268012 Chr.11: 110436197-110436197 on Build GRCh38
rs772475820 Chr.11: 110435996-110435996 on Build GRCh38
rs925944834 Chr.11: 110435820-110435820 on Build GRCh38
rs58842739 Chr.11: 110435799-110435799 on Build GRCh38
rs773868601 Chr.11: 110435924-110435924 on Build GRCh38
rs199894632 Chr.11: 110435868-110435868 on Build GRCh38
rs1017418941 Chr.11: 110436056-110436056 on Build GRCh38
rs748363171 Chr.11: 110435823-110435823 on Build GRCh38
rs1030220913 Chr.11: 110436104-110436104 on Build GRCh38
rs879199313 Chr.11: 110436122-110436122 on Build GRCh38
rs963491093 Chr.11: 110436156-110436156 on Build GRCh38
rs761435469 Chr.11: 110435869-110435869 on Build GRCh38
rs1034066738 Chr.11: 110436059-110436059 on Build GRCh38
rs749892432 Chr.11: 110435879-110435879 on Build GRCh38

View Full Product Details