Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 110579550-110580056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP20
Entrez Gene ID
57569
Gene Name
Rho GTPase activating protein 20
Gene Aliases
RARHOGAP
UniGene
Hs.6136
Gene Chromosome Location
Chr.11: 110577035-110713189 on Build GRCh38
Transcripts
NM_001258415.1 NM_001258416.1 NM_001258417.1 NM_001258418.1 NM_020809.3 XM_005271628.3 XM_006718893.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1183374 c.3230C>T p.A1077V Chr.11: 110579716-110579716 on Build GRCh38
4865182 c.3119C>A p.S1040Y Chr.11: 110579749-110579749 on Build GRCh38
4184189 c.2951G>C p.C984S Chr.11: 110579917-110579917 on Build GRCh38
1127912 c.3288G>A p.R1096R Chr.11: 110579658-110579658 on Build GRCh38
6306667 c.3290G>T p.R1097I Chr.11: 110579587-110579587 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756855502 Chr.11: 110579732-110579732 on Build GRCh38
rs752960539 Chr.11: 110579883-110579883 on Build GRCh38
rs147768508 Chr.11: 110579931-110579931 on Build GRCh38
rs759947104 Chr.11: 110579798-110579798 on Build GRCh38
rs143937766 Chr.11: 110579733-110579733 on Build GRCh38
rs748232042 Chr.11: 110579709-110579709 on Build GRCh38
rs770257363 Chr.11: 110579591-110579592 on Build GRCh38
rs773730819 Chr.11: 110579719-110579719 on Build GRCh38
rs534496852 Chr.11: 110579597-110579597 on Build GRCh38
rs957740355 Chr.11: 110579684-110579684 on Build GRCh38
rs1001530796 Chr.11: 110579594-110579594 on Build GRCh38
rs752905344 Chr.11: 110579981-110579981 on Build GRCh38
rs759634070 Chr.11: 110579951-110579951 on Build GRCh38
rs779503759 Chr.11: 110579915-110579915 on Build GRCh38
rs149514977 Chr.11: 110579729-110579729 on Build GRCh38
rs769914124 Chr.11: 110579779-110579779 on Build GRCh38
rs777889763 Chr.11: 110579886-110579886 on Build GRCh38
rs748178488 Chr.11: 110579765-110579765 on Build GRCh38
rs376924148 Chr.11: 110579970-110579970 on Build GRCh38
rs142882906 Chr.11: 110579573-110579573 on Build GRCh38
rs779262652 Chr.11: 110580025-110580025 on Build GRCh38
rs774519462 Chr.11: 110579797-110579797 on Build GRCh38
rs763579927 Chr.11: 110579626-110579626 on Build GRCh38
rs144971465 Chr.11: 110579612-110579612 on Build GRCh38
rs750434395 Chr.11: 110579737-110579737 on Build GRCh38
rs181642846 Chr.11: 110579839-110579839 on Build GRCh38
rs759224482 Chr.11: 110579969-110579969 on Build GRCh38
rs758180280 Chr.11: 110579739-110579739 on Build GRCh38
rs753328755 Chr.11: 110579730-110579730 on Build GRCh38
rs765747510 Chr.11: 110579599-110579599 on Build GRCh38
rs758149438 Chr.11: 110579676-110579676 on Build GRCh38
rs985548058 Chr.11: 110580030-110580030 on Build GRCh38
rs768775438 Chr.11: 110579700-110579700 on Build GRCh38
rs199551649 Chr.11: 110579948-110579948 on Build GRCh38
rs773761383 Chr.11: 110579637-110579638 on Build GRCh38
rs1048379456 Chr.11: 110579805-110579805 on Build GRCh38
rs1036693363 Chr.11: 110580023-110580023 on Build GRCh38
rs139127000 Chr.11: 110579725-110579725 on Build GRCh38
rs898243809 Chr.11: 110579716-110579716 on Build GRCh38
rs772613968 Chr.11: 110579932-110579932 on Build GRCh38
rs773502786 Chr.11: 110579861-110579861 on Build GRCh38
rs375823554 Chr.11: 110579589-110579589 on Build GRCh38
rs753818198 Chr.11: 110580003-110580003 on Build GRCh38
rs771273556 Chr.11: 110579912-110579912 on Build GRCh38
rs760923229 Chr.11: 110579942-110579942 on Build GRCh38
rs377611284 Chr.11: 110579897-110579897 on Build GRCh38
rs749821280 Chr.11: 110579852-110579853 on Build GRCh38
rs377207066 Chr.11: 110579631-110579631 on Build GRCh38
rs755091346 Chr.11: 110579754-110579754 on Build GRCh38
rs1051994482 Chr.11: 110579656-110579656 on Build GRCh38
rs918391594 Chr.11: 110579772-110579772 on Build GRCh38
rs770247166 Chr.11: 110579715-110579715 on Build GRCh38
rs149310586 Chr.11: 110579872-110579872 on Build GRCh38
rs139449100 Chr.11: 110579949-110579949 on Build GRCh38
rs781424737 Chr.11: 110579755-110579755 on Build GRCh38
rs751274213 Chr.11: 110579871-110579871 on Build GRCh38
rs772783670 Chr.11: 110579947-110579947 on Build GRCh38
rs751511045 Chr.11: 110580002-110580002 on Build GRCh38
rs200535574 Chr.11: 110579858-110579858 on Build GRCh38
rs765414431 Chr.11: 110579993-110579993 on Build GRCh38
rs773681800 Chr.11: 110579786-110579786 on Build GRCh38
rs202001398 Chr.11: 110579939-110579939 on Build GRCh38
rs73553932 Chr.11: 110579639-110579639 on Build GRCh38
rs756186455 Chr.11: 110579884-110579884 on Build GRCh38
rs144340070 Chr.11: 110579790-110579790 on Build GRCh38
rs768150332 Chr.11: 110579809-110579809 on Build GRCh38
rs776441588 Chr.11: 110579819-110579819 on Build GRCh38
rs953612058 Chr.11: 110579995-110579995 on Build GRCh38
rs750379178 Chr.11: 110579835-110579835 on Build GRCh38
rs546583933 Chr.11: 110579815-110579815 on Build GRCh38
rs140374907 Chr.11: 110579753-110579753 on Build GRCh38
rs372534511 Chr.11: 110579604-110579604 on Build GRCh38
rs150854099 Chr.11: 110579620-110579620 on Build GRCh38
rs780300469 Chr.11: 110579680-110579680 on Build GRCh38
rs755491292 Chr.11: 110579823-110579823 on Build GRCh38
rs767153914 Chr.11: 110579614-110579614 on Build GRCh38
rs776392265 Chr.11: 110579936-110579936 on Build GRCh38
rs1041289013 Chr.11: 110579746-110579746 on Build GRCh38
rs532744277 Chr.11: 110579731-110579731 on Build GRCh38
rs1055783150 Chr.11: 110579927-110579927 on Build GRCh38
rs761384767 Chr.11: 110579820-110579820 on Build GRCh38
rs369256834 Chr.11: 110579593-110579593 on Build GRCh38
rs146241130 Chr.11: 110579940-110579940 on Build GRCh38
rs753810570 Chr.11: 110579630-110579630 on Build GRCh38
rs757376519 Chr.11: 110580022-110580022 on Build GRCh38
rs111989729 Chr.11: 110579622-110579622 on Build GRCh38
rs151328327 Chr.11: 110579957-110579957 on Build GRCh38
rs747234738 Chr.11: 110579690-110579690 on Build GRCh38
rs537163504 Chr.11: 110579875-110579875 on Build GRCh38
rs966704845 Chr.11: 110579840-110579840 on Build GRCh38
rs138835399 Chr.11: 110579617-110579617 on Build GRCh38
rs368567456 Chr.11: 110579910-110579910 on Build GRCh38
rs374179856 Chr.11: 110579722-110579722 on Build GRCh38
rs369088115 Chr.11: 110579703-110579703 on Build GRCh38

View Full Product Details