Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111512103-111512612 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
UniGene
Hs.128180
Gene Chromosome Location
Chr.11: 111383828-111512388 on Build GRCh38
Transcripts
NM_017589.3 XM_011542876.2 XM_011542878.2 XM_011542879.2 XM_011542880.2

SNP Detail

SNP ID Chromosome Location
rs113782040 Chr.11: 111512347-111512347 on Build GRCh38
rs1006196846 Chr.11: 111512203-111512203 on Build GRCh38
rs1028634449 Chr.11: 111512298-111512298 on Build GRCh38
rs560656636 Chr.11: 111512333-111512334 on Build GRCh38
rs530386716 Chr.11: 111512215-111512215 on Build GRCh38
rs759087710 Chr.11: 111512337-111512337 on Build GRCh38
rs1055042809 Chr.11: 111512506-111512506 on Build GRCh38
rs968884065 Chr.11: 111512469-111512469 on Build GRCh38
rs1033726908 Chr.11: 111512204-111512204 on Build GRCh38
rs373333956 Chr.11: 111512371-111512371 on Build GRCh38
rs902463397 Chr.11: 111512202-111512202 on Build GRCh38
rs1032178274 Chr.11: 111512233-111512233 on Build GRCh38
rs895820534 Chr.11: 111512128-111512128 on Build GRCh38
rs929459285 Chr.11: 111512427-111512427 on Build GRCh38
rs971650964 Chr.11: 111512574-111512574 on Build GRCh38
rs998676903 Chr.11: 111512584-111512584 on Build GRCh38
rs923419929 Chr.11: 111512414-111512414 on Build GRCh38
rs369158551 Chr.11: 111512275-111512275 on Build GRCh38
rs971364490 Chr.11: 111512246-111512246 on Build GRCh38
rs1048464470 Chr.11: 111512439-111512439 on Build GRCh38
rs935243113 Chr.11: 111512518-111512518 on Build GRCh38
rs957281199 Chr.11: 111512331-111512331 on Build GRCh38
rs753449225 Chr.11: 111512158-111512158 on Build GRCh38
rs954508065 Chr.11: 111512310-111512310 on Build GRCh38
rs1006641351 Chr.11: 111512479-111512479 on Build GRCh38
rs937416012 Chr.11: 111512420-111512420 on Build GRCh38
rs996047182 Chr.11: 111512572-111512572 on Build GRCh38
rs887995007 Chr.11: 111512474-111512474 on Build GRCh38
rs1046331026 Chr.11: 111512199-111512199 on Build GRCh38
rs1024092528 Chr.11: 111512234-111512234 on Build GRCh38
rs916075620 Chr.11: 111512408-111512408 on Build GRCh38
rs924523653 Chr.11: 111512322-111512322 on Build GRCh38
rs895275457 Chr.11: 111512507-111512507 on Build GRCh38
rs917367698 Chr.11: 111512435-111512435 on Build GRCh38
rs1053588171 Chr.11: 111512529-111512531 on Build GRCh38
rs959413844 Chr.11: 111512214-111512214 on Build GRCh38
rs990591742 Chr.11: 111512402-111512402 on Build GRCh38
rs1013607360 Chr.11: 111512513-111512513 on Build GRCh38
rs1024123783 Chr.11: 111512557-111512557 on Build GRCh38
rs1040587328 Chr.11: 111512399-111512399 on Build GRCh38
rs887785596 Chr.11: 111512178-111512178 on Build GRCh38
rs10789838 Chr.11: 111512390-111512390 on Build GRCh38
rs879513868 Chr.11: 111512544-111512544 on Build GRCh38
rs532648209 Chr.11: 111512368-111512368 on Build GRCh38
rs973505124 Chr.11: 111512391-111512391 on Build GRCh38
rs80073960 Chr.11: 111512223-111512223 on Build GRCh38

View Full Product Details