Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111874303-111874807 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXACB1
Entrez Gene ID
91893
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
hCG_2033039
UniGene
Hs.697132
Gene Chromosome Location
Chr.11: 111874056-111879457 on Build GRCh38
Transcripts
NM_138378.2 NR_038364.1

SNP Detail

SNP ID Chromosome Location
rs569683361 Chr.11: 111874758-111874758 on Build GRCh38
rs782573932 Chr.11: 111874477-111874477 on Build GRCh38
rs605655 Chr.11: 111874735-111874735 on Build GRCh38
rs993790118 Chr.11: 111874397-111874397 on Build GRCh38
rs945349270 Chr.11: 111874520-111874520 on Build GRCh38
rs966767602 Chr.11: 111874714-111874714 on Build GRCh38
rs984921813 Chr.11: 111874470-111874470 on Build GRCh38
rs78960577 Chr.11: 111874760-111874760 on Build GRCh38
rs952258502 Chr.11: 111874629-111874629 on Build GRCh38
rs574872281 Chr.11: 111874530-111874530 on Build GRCh38
rs535629792 Chr.11: 111874761-111874761 on Build GRCh38
rs1054258711 Chr.11: 111874545-111874545 on Build GRCh38
rs550191900 Chr.11: 111874566-111874566 on Build GRCh38
rs543286957 Chr.11: 111874369-111874369 on Build GRCh38
rs571837697 Chr.11: 111874567-111874567 on Build GRCh38
rs952248506 Chr.11: 111874466-111874466 on Build GRCh38
rs554503092 Chr.11: 111874334-111874334 on Build GRCh38
rs556948516 Chr.11: 111874478-111874478 on Build GRCh38
rs558735122 Chr.11: 111874374-111874374 on Build GRCh38
rs146307924 Chr.11: 111874398-111874398 on Build GRCh38
rs959465773 Chr.11: 111874674-111874674 on Build GRCh38
rs924513268 Chr.11: 111874533-111874533 on Build GRCh38
rs782776215 Chr.11: 111874421-111874421 on Build GRCh38
rs575937193 Chr.11: 111874352-111874352 on Build GRCh38
rs1033779539 Chr.11: 111874673-111874673 on Build GRCh38
rs926712520 Chr.11: 111874472-111874472 on Build GRCh38
rs76503924 Chr.11: 111874761-111874761 on Build GRCh38
rs565847254 Chr.11: 111874621-111874621 on Build GRCh38
rs541847817 Chr.11: 111874519-111874519 on Build GRCh38
rs538343566 Chr.11: 111874370-111874370 on Build GRCh38
rs535817559 Chr.11: 111874577-111874577 on Build GRCh38
rs781911406 Chr.11: 111874393-111874393 on Build GRCh38
rs917979681 Chr.11: 111874705-111874705 on Build GRCh38
rs10670929 Chr.11: 111874604-111874605 on Build GRCh38
rs901539727 Chr.11: 111874615-111874615 on Build GRCh38
rs181778357 Chr.11: 111874517-111874517 on Build GRCh38
rs536301739 Chr.11: 111874740-111874740 on Build GRCh38
rs541164416 Chr.11: 111874473-111874473 on Build GRCh38
rs536237399 Chr.11: 111874332-111874332 on Build GRCh38
rs186364603 Chr.11: 111874578-111874578 on Build GRCh38
rs11418999 Chr.11: 111874594-111874595 on Build GRCh38
rs762970523 Chr.11: 111874732-111874732 on Build GRCh38
rs372706355 Chr.11: 111874420-111874420 on Build GRCh38
rs782475028 Chr.11: 111874476-111874476 on Build GRCh38
rs1006353045 Chr.11: 111874646-111874646 on Build GRCh38
rs57964658 Chr.11: 111874605-111874606 on Build GRCh38
rs781827804 Chr.11: 111874440-111874440 on Build GRCh38
rs1040510413 Chr.11: 111874582-111874582 on Build GRCh38
rs191646487 Chr.11: 111874691-111874691 on Build GRCh38
rs532059042 Chr.11: 111874538-111874538 on Build GRCh38
rs547911036 Chr.11: 111874616-111874616 on Build GRCh38
rs978492881 Chr.11: 111874729-111874729 on Build GRCh38
rs554568105 Chr.11: 111874742-111874742 on Build GRCh38
rs992278506 Chr.11: 111874679-111874679 on Build GRCh38
rs376881205 Chr.11: 111874479-111874479 on Build GRCh38

View Full Product Details