Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111875586-111876093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXACB1
Entrez Gene ID
91893
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
hCG_2033039
UniGene
Hs.697132
Gene Chromosome Location
Chr.11: 111874056-111879457 on Build GRCh38
Transcripts
NM_138378.2 NR_038364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5371646 c.772C>T p.P258S Chr.11: 111876025-111876025 on Build GRCh38
127993 c.1077C>T p.I359I Chr.11: 111875720-111875720 on Build GRCh38
4620282 c.1124C>G p.P375R Chr.11: 111875673-111875673 on Build GRCh38
7100064 c.829C>T p.L277F Chr.11: 111875968-111875968 on Build GRCh38
4801828 c.807G>T p.L269L Chr.11: 111875990-111875990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782556237 Chr.11: 111875878-111875879 on Build GRCh38
rs782224550 Chr.11: 111875886-111875886 on Build GRCh38
rs782029467 Chr.11: 111875998-111875998 on Build GRCh38
rs781976406 Chr.11: 111875910-111875910 on Build GRCh38
rs782812093 Chr.11: 111875947-111875947 on Build GRCh38
rs901198217 Chr.11: 111875945-111875945 on Build GRCh38
rs370683740 Chr.11: 111875781-111875781 on Build GRCh38
rs781979749 Chr.11: 111875654-111875654 on Build GRCh38
rs782546963 Chr.11: 111875792-111875792 on Build GRCh38
rs782353556 Chr.11: 111875652-111875652 on Build GRCh38
rs373040520 Chr.11: 111875673-111875673 on Build GRCh38
rs782404074 Chr.11: 111875638-111875638 on Build GRCh38
rs267602691 Chr.11: 111876026-111876026 on Build GRCh38
rs782239950 Chr.11: 111875752-111875752 on Build GRCh38
rs782250299 Chr.11: 111875856-111875856 on Build GRCh38
rs781788299 Chr.11: 111875720-111875720 on Build GRCh38
rs782651405 Chr.11: 111875743-111875743 on Build GRCh38
rs546895043 Chr.11: 111875890-111875890 on Build GRCh38
rs372889858 Chr.11: 111875728-111875728 on Build GRCh38
rs367754447 Chr.11: 111875881-111875881 on Build GRCh38
rs782807549 Chr.11: 111875899-111875899 on Build GRCh38
rs782147764 Chr.11: 111875898-111875898 on Build GRCh38
rs782127058 Chr.11: 111875919-111875919 on Build GRCh38
rs781861315 Chr.11: 111875801-111875801 on Build GRCh38
rs782188535 Chr.11: 111875620-111875620 on Build GRCh38
rs267602690 Chr.11: 111875997-111875997 on Build GRCh38
rs371997183 Chr.11: 111875897-111875897 on Build GRCh38
rs924214638 Chr.11: 111875835-111875835 on Build GRCh38
rs782712776 Chr.11: 111875782-111875782 on Build GRCh38
rs781943119 Chr.11: 111875764-111875764 on Build GRCh38
rs755792614 Chr.11: 111875691-111875691 on Build GRCh38
rs963165619 Chr.11: 111876066-111876066 on Build GRCh38
rs781795968 Chr.11: 111875938-111875938 on Build GRCh38
rs782777581 Chr.11: 111875694-111875694 on Build GRCh38
rs782061518 Chr.11: 111875756-111875756 on Build GRCh38
rs782020485 Chr.11: 111876045-111876045 on Build GRCh38
rs782205189 Chr.11: 111875830-111875830 on Build GRCh38
rs374999467 Chr.11: 111875639-111875639 on Build GRCh38
rs376903298 Chr.11: 111875940-111875940 on Build GRCh38
rs782284495 Chr.11: 111875729-111875729 on Build GRCh38
rs557152920 Chr.11: 111875971-111875971 on Build GRCh38
rs370077571 Chr.11: 111876023-111876023 on Build GRCh38
rs782450166 Chr.11: 111875722-111875722 on Build GRCh38
rs368610130 Chr.11: 111875822-111875822 on Build GRCh38
rs563448840 Chr.11: 111875771-111875771 on Build GRCh38
rs534644379 Chr.11: 111875857-111875857 on Build GRCh38
rs61739323 Chr.11: 111875642-111875642 on Build GRCh38
rs781881926 Chr.11: 111875722-111875723 on Build GRCh38
rs552308305 Chr.11: 111875852-111875852 on Build GRCh38
rs782648947 Chr.11: 111876069-111876070 on Build GRCh38
rs782167123 Chr.11: 111876059-111876059 on Build GRCh38
rs374384559 Chr.11: 111875796-111875796 on Build GRCh38
rs368384184 Chr.11: 111876008-111876008 on Build GRCh38
rs782736410 Chr.11: 111876034-111876034 on Build GRCh38
rs782215561 Chr.11: 111875968-111875968 on Build GRCh38
rs376141657 Chr.11: 111875775-111875775 on Build GRCh38
rs782721202 Chr.11: 111875776-111875776 on Build GRCh38
rs782162578 Chr.11: 111875688-111875688 on Build GRCh38
rs781891264 Chr.11: 111875680-111875680 on Build GRCh38
rs781986780 Chr.11: 111875774-111875774 on Build GRCh38
rs370579560 Chr.11: 111875713-111875713 on Build GRCh38
rs189903941 Chr.11: 111876024-111876024 on Build GRCh38
rs782469814 Chr.11: 111875717-111875717 on Build GRCh38
rs781886688 Chr.11: 111875784-111875784 on Build GRCh38
rs782363909 Chr.11: 111875734-111875734 on Build GRCh38
rs782383946 Chr.11: 111876013-111876013 on Build GRCh38
rs782559863 Chr.11: 111875980-111875980 on Build GRCh38
rs375728601 Chr.11: 111876068-111876068 on Build GRCh38
rs374128358 Chr.11: 111875871-111875871 on Build GRCh38
rs782504862 Chr.11: 111875817-111875817 on Build GRCh38
rs782400704 Chr.11: 111875986-111875986 on Build GRCh38
rs983243600 Chr.11: 111875658-111875658 on Build GRCh38
rs782714957 Chr.11: 111875922-111875922 on Build GRCh38
rs372325121 Chr.11: 111875981-111875981 on Build GRCh38
rs781909860 Chr.11: 111875949-111875949 on Build GRCh38

View Full Product Details