Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111876401-111876911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FDXACB1
Entrez Gene ID
91893
Gene Name
ferredoxin-fold anticodon binding domain containing 1
Gene Aliases
hCG_2033039
UniGene
Hs.697132
Gene Chromosome Location
Chr.11: 111874056-111879457 on Build GRCh38
Transcripts
NM_138378.2 NR_038364.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3998228 c.600T>C p.F200F Chr.11: 111876573-111876573 on Build GRCh38
5961924 c.478G>A p.D160N Chr.11: 111876863-111876863 on Build GRCh38
7154114 c.643C>A p.Q215K Chr.11: 111876530-111876530 on Build GRCh38
6552663 c.494G>A p.S165N Chr.11: 111876847-111876847 on Build GRCh38
5522688 c.653C>T p.S218F Chr.11: 111876520-111876520 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145927035 Chr.11: 111876457-111876457 on Build GRCh38
rs149256640 Chr.11: 111876864-111876864 on Build GRCh38
rs34493659 Chr.11: 111876460-111876461 on Build GRCh38
rs782681033 Chr.11: 111876882-111876883 on Build GRCh38
rs782682396 Chr.11: 111876817-111876817 on Build GRCh38
rs782750045 Chr.11: 111876572-111876573 on Build GRCh38
rs989758095 Chr.11: 111876723-111876723 on Build GRCh38
rs922684237 Chr.11: 111876825-111876825 on Build GRCh38
rs781791480 Chr.11: 111876874-111876874 on Build GRCh38
rs201248796 Chr.11: 111876578-111876578 on Build GRCh38
rs782275453 Chr.11: 111876818-111876818 on Build GRCh38
rs782395814 Chr.11: 111876615-111876615 on Build GRCh38
rs782420239 Chr.11: 111876635-111876635 on Build GRCh38
rs1030642733 Chr.11: 111876700-111876700 on Build GRCh38
rs781990717 Chr.11: 111876506-111876506 on Build GRCh38
rs782121939 Chr.11: 111876642-111876642 on Build GRCh38
rs782535465 Chr.11: 111876808-111876808 on Build GRCh38
rs782709281 Chr.11: 111876769-111876769 on Build GRCh38
rs781866894 Chr.11: 111876577-111876577 on Build GRCh38
rs915488331 Chr.11: 111876754-111876754 on Build GRCh38
rs75258745 Chr.11: 111876863-111876863 on Build GRCh38
rs782385232 Chr.11: 111876819-111876819 on Build GRCh38
rs782689448 Chr.11: 111876599-111876599 on Build GRCh38
rs782300536 Chr.11: 111876849-111876849 on Build GRCh38
rs367619899 Chr.11: 111876624-111876624 on Build GRCh38
rs781790559 Chr.11: 111876431-111876431 on Build GRCh38
rs138528048 Chr.11: 111876669-111876669 on Build GRCh38
rs782482760 Chr.11: 111876783-111876783 on Build GRCh38
rs782740584 Chr.11: 111876644-111876644 on Build GRCh38
rs781825889 Chr.11: 111876762-111876762 on Build GRCh38
rs782148977 Chr.11: 111876634-111876634 on Build GRCh38
rs542018782 Chr.11: 111876514-111876514 on Build GRCh38
rs782353304 Chr.11: 111876492-111876492 on Build GRCh38
rs781784423 Chr.11: 111876875-111876876 on Build GRCh38
rs1004782244 Chr.11: 111876505-111876505 on Build GRCh38
rs199785091 Chr.11: 111876800-111876800 on Build GRCh38
rs782502313 Chr.11: 111876882-111876882 on Build GRCh38
rs367952793 Chr.11: 111876860-111876860 on Build GRCh38
rs781830391 Chr.11: 111876648-111876648 on Build GRCh38
rs371392509 Chr.11: 111876596-111876596 on Build GRCh38
rs782246443 Chr.11: 111876610-111876610 on Build GRCh38
rs781973551 Chr.11: 111876538-111876538 on Build GRCh38
rs782193078 Chr.11: 111876802-111876802 on Build GRCh38
rs782288335 Chr.11: 111876440-111876440 on Build GRCh38
rs782024980 Chr.11: 111876854-111876854 on Build GRCh38
rs376752367 Chr.11: 111876581-111876581 on Build GRCh38
rs782347826 Chr.11: 111876515-111876515 on Build GRCh38
rs372752086 Chr.11: 111876432-111876432 on Build GRCh38
rs886368667 Chr.11: 111876496-111876496 on Build GRCh38
rs201093596 Chr.11: 111876630-111876630 on Build GRCh38
rs782148713 Chr.11: 111876687-111876687 on Build GRCh38
rs781984213 Chr.11: 111876639-111876639 on Build GRCh38
rs782053847 Chr.11: 111876546-111876546 on Build GRCh38
rs200419859 Chr.11: 111876861-111876861 on Build GRCh38
rs374653051 Chr.11: 111876621-111876621 on Build GRCh38
rs782647610 Chr.11: 111876889-111876889 on Build GRCh38
rs782713079 Chr.11: 111876564-111876564 on Build GRCh38
rs781946532 Chr.11: 111876828-111876828 on Build GRCh38
rs782282119 Chr.11: 111876499-111876499 on Build GRCh38
rs634983 Chr.11: 111876703-111876703 on Build GRCh38
rs782420720 Chr.11: 111876502-111876502 on Build GRCh38
rs782647899 Chr.11: 111876758-111876758 on Build GRCh38
rs529154867 Chr.11: 111876776-111876776 on Build GRCh38
rs550817278 Chr.11: 111876792-111876792 on Build GRCh38
rs782052793 Chr.11: 111876450-111876450 on Build GRCh38
rs782767907 Chr.11: 111876858-111876858 on Build GRCh38
rs782173873 Chr.11: 111876856-111876856 on Build GRCh38
rs782227098 Chr.11: 111876623-111876623 on Build GRCh38
rs530922734 Chr.11: 111876589-111876589 on Build GRCh38
rs781881813 Chr.11: 111876571-111876571 on Build GRCh38
rs901253629 Chr.11: 111876435-111876435 on Build GRCh38
rs782046574 Chr.11: 111876664-111876664 on Build GRCh38
rs782596239 Chr.11: 111876617-111876617 on Build GRCh38
rs781913851 Chr.11: 111876685-111876685 on Build GRCh38
rs782671190 Chr.11: 111876439-111876439 on Build GRCh38
rs782507091 Chr.11: 111876456-111876456 on Build GRCh38
rs782748041 Chr.11: 111876576-111876576 on Build GRCh38
rs554893289 Chr.11: 111876878-111876878 on Build GRCh38
rs782508285 Chr.11: 111876686-111876686 on Build GRCh38
rs782592918 Chr.11: 111876494-111876494 on Build GRCh38
rs782206205 Chr.11: 111876488-111876488 on Build GRCh38
rs569000920 Chr.11: 111876829-111876829 on Build GRCh38
rs1047021765 Chr.11: 111876487-111876487 on Build GRCh38
rs7124696 Chr.11: 111876573-111876573 on Build GRCh38
rs568373524 Chr.11: 111876708-111876708 on Build GRCh38
rs782127283 Chr.11: 111876534-111876534 on Build GRCh38

View Full Product Details