Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112028659-112029188 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLAT
Entrez Gene ID
1737
Gene Name
dihydrolipoamide S-acetyltransferase
Gene Aliases
DLTA, PDC-E2, PDCE2
UniGene
Hs.335551
Gene Chromosome Location
Chr.11: 112024814-112064278 on Build GRCh38
Transcripts
NM_001931.4 XM_011542647.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5479486 c.507-9_507-6delCTTC p.? Chr.11: 112028783-112028786 on Build GRCh38
1351263 c.605C>T p.S202L Chr.11: 112028890-112028890 on Build GRCh38
5548986 c.572C>T p.A191V Chr.11: 112028857-112028857 on Build GRCh38
3716751 c.515A>C p.D172A Chr.11: 112028800-112028800 on Build GRCh38
2111892 c.606G>A p.S202S Chr.11: 112028891-112028891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141869115 Chr.11: 112028895-112028895 on Build GRCh38
rs782356632 Chr.11: 112028838-112028839 on Build GRCh38
rs782500644 Chr.11: 112028985-112028985 on Build GRCh38
rs201046000 Chr.11: 112028809-112028809 on Build GRCh38
rs148153443 Chr.11: 112028891-112028891 on Build GRCh38
rs896040101 Chr.11: 112029019-112029019 on Build GRCh38
rs782718789 Chr.11: 112028786-112028786 on Build GRCh38
rs782453317 Chr.11: 112028849-112028849 on Build GRCh38
rs367605727 Chr.11: 112028795-112028795 on Build GRCh38
rs79063223 Chr.11: 112029164-112029164 on Build GRCh38
rs140678772 Chr.11: 112028913-112028913 on Build GRCh38
rs140241473 Chr.11: 112028824-112028824 on Build GRCh38
rs143107853 Chr.11: 112028855-112028855 on Build GRCh38
rs781951438 Chr.11: 112028890-112028890 on Build GRCh38
rs782423670 Chr.11: 112028871-112028871 on Build GRCh38
rs144782209 Chr.11: 112028870-112028870 on Build GRCh38
rs1041993012 Chr.11: 112029149-112029149 on Build GRCh38
rs1032795731 Chr.11: 112028815-112028815 on Build GRCh38
rs371720128 Chr.11: 112028803-112028803 on Build GRCh38
rs782723590 Chr.11: 112028959-112028959 on Build GRCh38
rs369523624 Chr.11: 112028992-112028992 on Build GRCh38
rs782488131 Chr.11: 112028954-112028954 on Build GRCh38
rs375316997 Chr.11: 112028808-112028808 on Build GRCh38
rs35867231 Chr.11: 112028785-112028786 on Build GRCh38
rs781869552 Chr.11: 112028821-112028821 on Build GRCh38
rs145146632 Chr.11: 112028851-112028851 on Build GRCh38
rs782379548 Chr.11: 112028685-112028685 on Build GRCh38
rs374343565 Chr.11: 112028776-112028776 on Build GRCh38
rs1056433614 Chr.11: 112028711-112028711 on Build GRCh38
rs11553595 Chr.11: 112028911-112028911 on Build GRCh38
rs782036767 Chr.11: 112028908-112028908 on Build GRCh38
rs376677023 Chr.11: 112028996-112028996 on Build GRCh38
rs782225633 Chr.11: 112028853-112028853 on Build GRCh38
rs371678198 Chr.11: 112029147-112029147 on Build GRCh38
rs1055900020 Chr.11: 112028958-112028958 on Build GRCh38
rs896092514 Chr.11: 112028721-112028721 on Build GRCh38
rs782677556 Chr.11: 112028847-112028847 on Build GRCh38
rs200500508 Chr.11: 112028857-112028857 on Build GRCh38
rs34321117 Chr.11: 112028871-112028872 on Build GRCh38
rs782197801 Chr.11: 112029020-112029020 on Build GRCh38
rs782369386 Chr.11: 112028888-112028888 on Build GRCh38
rs781985127 Chr.11: 112029042-112029042 on Build GRCh38
rs149440666 Chr.11: 112028868-112028868 on Build GRCh38
rs782596110 Chr.11: 112028844-112028844 on Build GRCh38
rs781916095 Chr.11: 112028743-112028743 on Build GRCh38
rs35957705 Chr.11: 112028847-112028848 on Build GRCh38
rs782767621 Chr.11: 112028896-112028896 on Build GRCh38
rs200488420 Chr.11: 112028848-112028848 on Build GRCh38
rs781789669 Chr.11: 112028936-112028936 on Build GRCh38
rs763843823 Chr.11: 112028723-112028724 on Build GRCh38
rs781863843 Chr.11: 112028971-112028971 on Build GRCh38
rs367824868 Chr.11: 112028765-112028765 on Build GRCh38
rs782366427 Chr.11: 112028880-112028880 on Build GRCh38
rs57586330 Chr.11: 112029028-112029028 on Build GRCh38
rs888607150 Chr.11: 112028691-112028691 on Build GRCh38
rs781963289 Chr.11: 112029124-112029124 on Build GRCh38
rs1013844258 Chr.11: 112028734-112028734 on Build GRCh38
rs587598588 Chr.11: 112028704-112028704 on Build GRCh38

View Full Product Details