Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112073902-112074399 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf57
Entrez Gene ID
55216
Gene Name
chromosome 11 open reading frame 57
Gene Aliases
-
UniGene
Hs.195060
Gene Chromosome Location
Chr.11: 112074244-112085150 on Build GRCh38
Transcripts
NM_001082969.1 NM_001082970.1 NM_001301017.1 NM_001301019.1 NM_001301021.1 NM_018195.3 NR_103469.1 NM_001082619.1 NM_138789.3 XM_011542593.2 XM_017017201.1 XM_017017202.1 XM_017017203.1 XM_017017204.1 XM_017017205.1

Gene Symbol
PIH1D2
Entrez Gene ID
120379
Gene Name
PIH1 domain containing 2
Gene Aliases
-
UniGene
Hs.420662
Gene Chromosome Location
Chr.11: 112052161-112074288 on Build GRCh38
Transcripts
NM_001082969.1 NM_001082970.1 NM_001301017.1 NM_001301019.1 NM_001301021.1 NM_018195.3 NR_103469.1 NM_001082619.1 NM_138789.3 XM_011542593.2 XM_017017201.1 XM_017017202.1 XM_017017203.1 XM_017017204.1 XM_017017205.1

SNP Detail

SNP ID Chromosome Location
rs533168207 Chr.11: 112074158-112074158 on Build GRCh38
rs980743421 Chr.11: 112074243-112074243 on Build GRCh38
rs182483100 Chr.11: 112074326-112074326 on Build GRCh38
rs782642856 Chr.11: 112073948-112073948 on Build GRCh38
rs890779292 Chr.11: 112074372-112074372 on Build GRCh38
rs369525741 Chr.11: 112074333-112074333 on Build GRCh38
rs782555026 Chr.11: 112074082-112074082 on Build GRCh38
rs573820019 Chr.11: 112074335-112074335 on Build GRCh38
rs927998490 Chr.11: 112074245-112074245 on Build GRCh38
rs376223922 Chr.11: 112074181-112074181 on Build GRCh38
rs532049417 Chr.11: 112073975-112073975 on Build GRCh38
rs996431476 Chr.11: 112074127-112074127 on Build GRCh38
rs921494677 Chr.11: 112074336-112074336 on Build GRCh38
rs961017479 Chr.11: 112074315-112074315 on Build GRCh38
rs566578816 Chr.11: 112074085-112074085 on Build GRCh38
rs371448798 Chr.11: 112074249-112074249 on Build GRCh38
rs939468377 Chr.11: 112074024-112074024 on Build GRCh38
rs890841873 Chr.11: 112074150-112074150 on Build GRCh38
rs969368971 Chr.11: 112073966-112073966 on Build GRCh38
rs2303437 Chr.11: 112074177-112074177 on Build GRCh38
rs553802591 Chr.11: 112074334-112074334 on Build GRCh38
rs782220921 Chr.11: 112074241-112074241 on Build GRCh38
rs542780344 Chr.11: 112074344-112074344 on Build GRCh38
rs150668153 Chr.11: 112074362-112074362 on Build GRCh38
rs969965463 Chr.11: 112074239-112074239 on Build GRCh38
rs546665702 Chr.11: 112074007-112074007 on Build GRCh38
rs900103191 Chr.11: 112074112-112074112 on Build GRCh38
rs973998092 Chr.11: 112073954-112073954 on Build GRCh38
rs535596347 Chr.11: 112074097-112074097 on Build GRCh38
rs962571676 Chr.11: 112074193-112074193 on Build GRCh38
rs1043950496 Chr.11: 112074104-112074104 on Build GRCh38
rs549219540 Chr.11: 112074155-112074155 on Build GRCh38
rs532835830 Chr.11: 112073996-112073996 on Build GRCh38
rs1053001908 Chr.11: 112074064-112074064 on Build GRCh38
rs782332529 Chr.11: 112074279-112074279 on Build GRCh38
rs374660879 Chr.11: 112074299-112074299 on Build GRCh38
rs562814950 Chr.11: 112073956-112073956 on Build GRCh38
rs538124190 Chr.11: 112074232-112074232 on Build GRCh38
rs1050737414 Chr.11: 112074146-112074146 on Build GRCh38
rs782524649 Chr.11: 112073929-112073929 on Build GRCh38
rs782646109 Chr.11: 112074327-112074327 on Build GRCh38
rs79835487 Chr.11: 112073978-112073978 on Build GRCh38
rs78421692 Chr.11: 112074322-112074322 on Build GRCh38
rs995373818 Chr.11: 112074203-112074203 on Build GRCh38
rs932918844 Chr.11: 112074340-112074340 on Build GRCh38
rs142914267 Chr.11: 112073994-112073994 on Build GRCh38

View Full Product Details