Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112083405-112083914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf57
Entrez Gene ID
55216
Gene Name
chromosome 11 open reading frame 57
Gene Aliases
-
UniGene
Hs.195060
Gene Chromosome Location
Chr.11: 112074244-112085150 on Build GRCh38
Transcripts
NM_001082969.1 NM_001082970.1 NM_001301017.1 NM_001301019.1 NM_001301021.1 NM_018195.3 NR_103469.1

SNP Detail

SNP ID Chromosome Location
rs911279440 Chr.11: 112083645-112083645 on Build GRCh38
rs138507332 Chr.11: 112083584-112083584 on Build GRCh38
rs909836950 Chr.11: 112083866-112083866 on Build GRCh38
rs559247624 Chr.11: 112083622-112083622 on Build GRCh38
rs147281084 Chr.11: 112083737-112083737 on Build GRCh38
rs570373153 Chr.11: 112083715-112083715 on Build GRCh38
rs572797447 Chr.11: 112083636-112083636 on Build GRCh38
rs190574277 Chr.11: 112083482-112083482 on Build GRCh38
rs984275280 Chr.11: 112083617-112083626 on Build GRCh38
rs994427609 Chr.11: 112083442-112083442 on Build GRCh38
rs142285332 Chr.11: 112083839-112083839 on Build GRCh38
rs1001611977 Chr.11: 112083473-112083473 on Build GRCh38
rs929760565 Chr.11: 112083847-112083847 on Build GRCh38
rs75262267 Chr.11: 112083541-112083541 on Build GRCh38
rs542349863 Chr.11: 112083783-112083783 on Build GRCh38
rs905842970 Chr.11: 112083891-112083891 on Build GRCh38
rs766325560 Chr.11: 112083545-112083545 on Build GRCh38
rs918394022 Chr.11: 112083529-112083529 on Build GRCh38
rs959796040 Chr.11: 112083532-112083532 on Build GRCh38
rs944090397 Chr.11: 112083651-112083651 on Build GRCh38
rs888715066 Chr.11: 112083741-112083741 on Build GRCh38
rs968949517 Chr.11: 112083711-112083711 on Build GRCh38
rs184242653 Chr.11: 112083593-112083593 on Build GRCh38
rs1013955039 Chr.11: 112083864-112083864 on Build GRCh38
rs985881997 Chr.11: 112083637-112083637 on Build GRCh38
rs754994964 Chr.11: 112083712-112083712 on Build GRCh38
rs1035874820 Chr.11: 112083623-112083623 on Build GRCh38
rs761729307 Chr.11: 112083451-112083451 on Build GRCh38
rs767354901 Chr.11: 112083463-112083463 on Build GRCh38
rs140693312 Chr.11: 112083803-112083803 on Build GRCh38
rs117042730 Chr.11: 112083660-112083660 on Build GRCh38
rs1013991020 Chr.11: 112083535-112083535 on Build GRCh38
rs11546875 Chr.11: 112083523-112083523 on Build GRCh38
rs1022331694 Chr.11: 112083691-112083691 on Build GRCh38
rs911010845 Chr.11: 112083481-112083481 on Build GRCh38
rs369867535 Chr.11: 112083585-112083585 on Build GRCh38
rs1049156873 Chr.11: 112083445-112083445 on Build GRCh38
rs143907986 Chr.11: 112083728-112083728 on Build GRCh38
rs181924990 Chr.11: 112083492-112083492 on Build GRCh38
rs144786392 Chr.11: 112083542-112083542 on Build GRCh38
rs947954936 Chr.11: 112083868-112083870 on Build GRCh38
rs888086802 Chr.11: 112083468-112083468 on Build GRCh38
rs965144812 Chr.11: 112083793-112083793 on Build GRCh38
rs1044393648 Chr.11: 112083871-112083871 on Build GRCh38
rs1055378150 Chr.11: 112083826-112083826 on Build GRCh38
rs1010488661 Chr.11: 112083667-112083667 on Build GRCh38

View Full Product Details