Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 112084093-112084620 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf57
Entrez Gene ID
55216
Gene Name
chromosome 11 open reading frame 57
Gene Aliases
-
UniGene
Hs.195060
Gene Chromosome Location
Chr.11: 112074244-112085150 on Build GRCh38
Transcripts
NM_001082969.1 NM_001082970.1 NM_001301017.1 NM_001301019.1 NM_001301021.1 NM_018195.3 NR_103469.1

SNP Detail

SNP ID Chromosome Location
rs1057123419 Chr.11: 112084269-112084269 on Build GRCh38
rs1033245045 Chr.11: 112084221-112084221 on Build GRCh38
rs774123547 Chr.11: 112084400-112084400 on Build GRCh38
rs960641626 Chr.11: 112084559-112084559 on Build GRCh38
rs532724905 Chr.11: 112084317-112084317 on Build GRCh38
rs927299483 Chr.11: 112084245-112084245 on Build GRCh38
rs1005272147 Chr.11: 112084489-112084489 on Build GRCh38
rs1041377681 Chr.11: 112084441-112084441 on Build GRCh38
rs188971213 Chr.11: 112084304-112084304 on Build GRCh38
rs1025295337 Chr.11: 112084473-112084473 on Build GRCh38
rs993258764 Chr.11: 112084574-112084574 on Build GRCh38
rs555285281 Chr.11: 112084482-112084482 on Build GRCh38
rs775167892 Chr.11: 112084256-112084256 on Build GRCh38
rs7123075 Chr.11: 112084524-112084524 on Build GRCh38
rs917500600 Chr.11: 112084422-112084422 on Build GRCh38
rs909694736 Chr.11: 112084154-112084154 on Build GRCh38
rs1000003560 Chr.11: 112084206-112084206 on Build GRCh38
rs561120601 Chr.11: 112084205-112084205 on Build GRCh38
rs1007402439 Chr.11: 112084513-112084513 on Build GRCh38
rs935730801 Chr.11: 112084459-112084459 on Build GRCh38
rs946137138 Chr.11: 112084315-112084318 on Build GRCh38
rs1139693 Chr.11: 112084195-112084195 on Build GRCh38
rs895476464 Chr.11: 112084142-112084142 on Build GRCh38
rs750313050 Chr.11: 112084598-112084598 on Build GRCh38
rs965333990 Chr.11: 112084293-112084293 on Build GRCh38
rs1025706881 Chr.11: 112084324-112084324 on Build GRCh38
rs530146695 Chr.11: 112084250-112084250 on Build GRCh38
rs752086870 Chr.11: 112084131-112084131 on Build GRCh38
rs769377402 Chr.11: 112084145-112084145 on Build GRCh38
rs777378476 Chr.11: 112084183-112084183 on Build GRCh38
rs1056014261 Chr.11: 112084141-112084141 on Build GRCh38
rs559141148 Chr.11: 112084229-112084229 on Build GRCh38
rs576245355 Chr.11: 112084447-112084447 on Build GRCh38
rs765664632 Chr.11: 112084133-112084134 on Build GRCh38
rs949726761 Chr.11: 112084148-112084148 on Build GRCh38
rs552819545 Chr.11: 112084330-112084330 on Build GRCh38
rs901423317 Chr.11: 112084452-112084452 on Build GRCh38
rs1025855173 Chr.11: 112084117-112084117 on Build GRCh38
rs549964939 Chr.11: 112084255-112084255 on Build GRCh38
rs901775139 Chr.11: 112084530-112084530 on Build GRCh38
rs984246739 Chr.11: 112084371-112084371 on Build GRCh38
rs567111887 Chr.11: 112084518-112084518 on Build GRCh38
rs765094137 Chr.11: 112084169-112084169 on Build GRCh38
rs761451653 Chr.11: 112084502-112084502 on Build GRCh38
rs746388981 Chr.11: 112084121-112084121 on Build GRCh38
rs993444031 Chr.11: 112084582-112084582 on Build GRCh38
rs902948438 Chr.11: 112084196-112084196 on Build GRCh38
rs757980223 Chr.11: 112084133-112084133 on Build GRCh38
rs181665407 Chr.11: 112084340-112084340 on Build GRCh38
rs535331966 Chr.11: 112084388-112084388 on Build GRCh38
rs746717114 Chr.11: 112084271-112084271 on Build GRCh38
rs910040263 Chr.11: 112084395-112084395 on Build GRCh38
rs888912997 Chr.11: 112084485-112084485 on Build GRCh38
rs980621337 Chr.11: 112084222-112084222 on Build GRCh38
rs1139692 Chr.11: 112084240-112084240 on Build GRCh38
rs904665338 Chr.11: 112084328-112084328 on Build GRCh38
rs949696135 Chr.11: 112084424-112084424 on Build GRCh38
rs991665749 Chr.11: 112084401-112084401 on Build GRCh38
rs927917605 Chr.11: 112084546-112084546 on Build GRCh38

View Full Product Details