Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113399225-113399720 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKK1
Entrez Gene ID
255239
Gene Name
ankyrin repeat and kinase domain containing 1
Gene Aliases
PKK2
UniGene
Hs.448473
Gene Chromosome Location
Chr.11: 113386014-113400418 on Build GRCh38
Transcripts
NM_178510.1 XM_011542736.2 XM_011542737.2 XM_011542738.2 XM_017017475.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3443514 c.1722G>A p.L574L Chr.11: 113399691-113399691 on Build GRCh38
5360898 c.1350C>T p.H450H Chr.11: 113399319-113399319 on Build GRCh38
4145314 c.1683C>T p.Y561Y Chr.11: 113399652-113399652 on Build GRCh38
6537522 c.1290C>G p.A430A Chr.11: 113399259-113399259 on Build GRCh38
6053387 c.1583G>T p.R528I Chr.11: 113399552-113399552 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759973935 Chr.11: 113399663-113399663 on Build GRCh38
rs771657563 Chr.11: 113399649-113399650 on Build GRCh38
rs778583811 Chr.11: 113399359-113399359 on Build GRCh38
rs757199134 Chr.11: 113399472-113399472 on Build GRCh38
rs201891002 Chr.11: 113399303-113399303 on Build GRCh38
rs199986656 Chr.11: 113399292-113399292 on Build GRCh38
rs757194170 Chr.11: 113399353-113399353 on Build GRCh38
rs751344795 Chr.11: 113399621-113399621 on Build GRCh38
rs754746606 Chr.11: 113399692-113399692 on Build GRCh38
rs867811775 Chr.11: 113399464-113399464 on Build GRCh38
rs2734849 Chr.11: 113399438-113399438 on Build GRCh38
rs755664137 Chr.11: 113399345-113399345 on Build GRCh38
rs1002357219 Chr.11: 113399486-113399486 on Build GRCh38
rs766798510 Chr.11: 113399575-113399575 on Build GRCh38
rs750392041 Chr.11: 113399609-113399609 on Build GRCh38
rs754400717 Chr.11: 113399459-113399459 on Build GRCh38
rs34983219 Chr.11: 113399320-113399320 on Build GRCh38
rs770440138 Chr.11: 113399357-113399357 on Build GRCh38
rs772246720 Chr.11: 113399379-113399379 on Build GRCh38
rs765529414 Chr.11: 113399317-113399317 on Build GRCh38
rs200848862 Chr.11: 113399402-113399402 on Build GRCh38
rs530835710 Chr.11: 113399517-113399517 on Build GRCh38
rs760737435 Chr.11: 113399301-113399301 on Build GRCh38
rs780918392 Chr.11: 113399694-113399694 on Build GRCh38
rs769920822 Chr.11: 113399265-113399265 on Build GRCh38
rs1020995096 Chr.11: 113399541-113399541 on Build GRCh38
rs768460251 Chr.11: 113399665-113399665 on Build GRCh38
rs748655600 Chr.11: 113399326-113399326 on Build GRCh38
rs967985757 Chr.11: 113399577-113399577 on Build GRCh38
rs751085500 Chr.11: 113399681-113399681 on Build GRCh38
rs745688052 Chr.11: 113399482-113399482 on Build GRCh38
rs200848219 Chr.11: 113399457-113399457 on Build GRCh38
rs752191051 Chr.11: 113399344-113399344 on Build GRCh38
rs374849691 Chr.11: 113399300-113399300 on Build GRCh38
rs752421459 Chr.11: 113399446-113399446 on Build GRCh38
rs2587554 Chr.11: 113399503-113399503 on Build GRCh38
rs773796958 Chr.11: 113399386-113399387 on Build GRCh38
rs763823018 Chr.11: 113399582-113399582 on Build GRCh38
rs974220272 Chr.11: 113399425-113399425 on Build GRCh38
rs751399807 Chr.11: 113399688-113399688 on Build GRCh38
rs770107477 Chr.11: 113399277-113399277 on Build GRCh38
rs372461990 Chr.11: 113399677-113399677 on Build GRCh38
rs752326791 Chr.11: 113399571-113399571 on Build GRCh38
rs763885896 Chr.11: 113399578-113399578 on Build GRCh38
rs917886985 Chr.11: 113399460-113399460 on Build GRCh38
rs774912504 Chr.11: 113399659-113399659 on Build GRCh38
rs767125783 Chr.11: 113399443-113399443 on Build GRCh38
rs545925009 Chr.11: 113399417-113399417 on Build GRCh38
rs201207634 Chr.11: 113399456-113399456 on Build GRCh38
rs2734848 Chr.11: 113399652-113399652 on Build GRCh38
rs749145473 Chr.11: 113399280-113399280 on Build GRCh38
rs748225627 Chr.11: 113399542-113399542 on Build GRCh38
rs955623999 Chr.11: 113399369-113399369 on Build GRCh38
rs369229013 Chr.11: 113399473-113399473 on Build GRCh38
rs761318999 Chr.11: 113399675-113399675 on Build GRCh38
rs56005821 Chr.11: 113399319-113399319 on Build GRCh38
rs775748000 Chr.11: 113399401-113399401 on Build GRCh38
rs777675256 Chr.11: 113399648-113399648 on Build GRCh38
rs370088482 Chr.11: 113399356-113399356 on Build GRCh38
rs768742229 Chr.11: 113399410-113399410 on Build GRCh38
rs376661130 Chr.11: 113399453-113399453 on Build GRCh38
rs745661789 Chr.11: 113399362-113399362 on Build GRCh38
rs372776088 Chr.11: 113399514-113399514 on Build GRCh38
rs555490493 Chr.11: 113399680-113399680 on Build GRCh38
rs979276927 Chr.11: 113399376-113399376 on Build GRCh38
rs775487122 Chr.11: 113399294-113399294 on Build GRCh38
rs4938016 Chr.11: 113399293-113399293 on Build GRCh38
rs200507206 Chr.11: 113399456-113399456 on Build GRCh38
rs376080076 Chr.11: 113399682-113399682 on Build GRCh38
rs147474296 Chr.11: 113399647-113399647 on Build GRCh38
rs765134129 Chr.11: 113399599-113399599 on Build GRCh38
rs184645039 Chr.11: 113399342-113399342 on Build GRCh38
rs749805635 Chr.11: 113399649-113399649 on Build GRCh38
rs749849059 Chr.11: 113399679-113399679 on Build GRCh38
rs775129985 Chr.11: 113399652-113399653 on Build GRCh38
rs199896606 Chr.11: 113399442-113399442 on Build GRCh38
rs762324123 Chr.11: 113399428-113399428 on Build GRCh38
rs758090485 Chr.11: 113399579-113399579 on Build GRCh38
rs770245667 Chr.11: 113399545-113399545 on Build GRCh38
rs200099073 Chr.11: 113399622-113399622 on Build GRCh38
rs781363250 Chr.11: 113399645-113399645 on Build GRCh38
rs745509772 Chr.11: 113399643-113399644 on Build GRCh38
rs535913227 Chr.11: 113399669-113399669 on Build GRCh38
rs114700049 Chr.11: 113399646-113399646 on Build GRCh38
rs747682799 Chr.11: 113399253-113399253 on Build GRCh38
rs761024615 Chr.11: 113399339-113399339 on Build GRCh38
rs979853585 Chr.11: 113399592-113399592 on Build GRCh38
rs758224977 Chr.11: 113399615-113399615 on Build GRCh38
rs780584250 Chr.11: 113399250-113399250 on Build GRCh38
rs199570081 Chr.11: 113399538-113399538 on Build GRCh38
rs761353002 Chr.11: 113399597-113399597 on Build GRCh38
rs754594883 Chr.11: 113399248-113399248 on Build GRCh38
rs758186532 Chr.11: 113399485-113399485 on Build GRCh38
rs912302675 Chr.11: 113399437-113399437 on Build GRCh38
rs2587555 Chr.11: 113399311-113399311 on Build GRCh38
rs533347208 Chr.11: 113399416-113399416 on Build GRCh38
rs753463597 Chr.11: 113399587-113399587 on Build GRCh38
rs777823114 Chr.11: 113399273-113399273 on Build GRCh38
rs78229381 Chr.11: 113399302-113399302 on Build GRCh38
rs773748715 Chr.11: 113399547-113399547 on Build GRCh38
rs538752260 Chr.11: 113399352-113399352 on Build GRCh38

View Full Product Details