Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 113779706-113780203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN25
Entrez Gene ID
644672
Gene Name
claudin 25
Gene Aliases
-
UniGene
Hs.705360
Gene Chromosome Location
Chr.11: 113779796-113780485 on Build GRCh38
Transcripts
NM_001101389.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4672692 c.68G>A p.C23Y Chr.11: 113779863-113779863 on Build GRCh38
2103480 c.261C>A p.S87S Chr.11: 113780056-113780056 on Build GRCh38
1561086 c.55C>A p.L19I Chr.11: 113779850-113779850 on Build GRCh38
4610821 c.197delC p.F67fs*13 Chr.11: 113779992-113779992 on Build GRCh38
1284146 c.332G>T p.R111I Chr.11: 113780127-113780127 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773431568 Chr.11: 113779975-113779975 on Build GRCh38
rs182753809 Chr.11: 113780035-113780035 on Build GRCh38
rs749373712 Chr.11: 113779976-113779976 on Build GRCh38
rs779897572 Chr.11: 113779957-113779957 on Build GRCh38
rs374737042 Chr.11: 113779981-113779981 on Build GRCh38
rs747219472 Chr.11: 113780158-113780158 on Build GRCh38
rs201922794 Chr.11: 113780174-113780174 on Build GRCh38
rs17115958 Chr.11: 113779918-113779918 on Build GRCh38
rs751896747 Chr.11: 113779828-113779828 on Build GRCh38
rs1015858364 Chr.11: 113779887-113779887 on Build GRCh38
rs202065177 Chr.11: 113780145-113780145 on Build GRCh38
rs765466437 Chr.11: 113779776-113779776 on Build GRCh38
rs368739845 Chr.11: 113779832-113779832 on Build GRCh38
rs746964341 Chr.11: 113779958-113779958 on Build GRCh38
rs888715901 Chr.11: 113780047-113780047 on Build GRCh38
rs1016348772 Chr.11: 113780071-113780071 on Build GRCh38
rs377605266 Chr.11: 113779750-113779750 on Build GRCh38
rs774785286 Chr.11: 113779983-113779983 on Build GRCh38
rs556650979 Chr.11: 113779927-113779927 on Build GRCh38
rs375569415 Chr.11: 113779817-113779817 on Build GRCh38
rs918393061 Chr.11: 113779811-113779811 on Build GRCh38
rs764892613 Chr.11: 113779812-113779812 on Build GRCh38
rs779232461 Chr.11: 113780169-113780169 on Build GRCh38
rs907346246 Chr.11: 113779953-113779953 on Build GRCh38
rs779993191 Chr.11: 113779843-113779843 on Build GRCh38
rs538478199 Chr.11: 113779941-113779941 on Build GRCh38
rs765901652 Chr.11: 113780030-113780030 on Build GRCh38
rs201592139 Chr.11: 113780180-113780180 on Build GRCh38
rs772015749 Chr.11: 113779828-113779829 on Build GRCh38
rs757357516 Chr.11: 113780062-113780062 on Build GRCh38
rs368993117 Chr.11: 113780144-113780144 on Build GRCh38
rs766813269 Chr.11: 113779898-113779898 on Build GRCh38
rs769944171 Chr.11: 113779858-113779858 on Build GRCh38
rs776123054 Chr.11: 113780092-113780092 on Build GRCh38
rs778582555 Chr.11: 113779833-113779833 on Build GRCh38
rs188108588 Chr.11: 113779783-113779783 on Build GRCh38
rs748354515 Chr.11: 113779857-113779857 on Build GRCh38
rs971804554 Chr.11: 113779914-113779914 on Build GRCh38
rs768889315 Chr.11: 113779806-113779806 on Build GRCh38
rs769061794 Chr.11: 113780099-113780099 on Build GRCh38
rs779056527 Chr.11: 113780076-113780076 on Build GRCh38
rs373997049 Chr.11: 113779854-113779854 on Build GRCh38
rs370403067 Chr.11: 113779846-113779846 on Build GRCh38
rs749924583 Chr.11: 113779940-113779940 on Build GRCh38
rs372181019 Chr.11: 113780072-113780072 on Build GRCh38
rs375224880 Chr.11: 113779990-113779990 on Build GRCh38
rs761076385 Chr.11: 113780000-113780000 on Build GRCh38
rs190938380 Chr.11: 113779997-113779997 on Build GRCh38
rs375691427 Chr.11: 113779957-113779958 on Build GRCh38
rs751099592 Chr.11: 113780126-113780126 on Build GRCh38
rs955204579 Chr.11: 113780140-113780140 on Build GRCh38
rs757534339 Chr.11: 113780165-113780165 on Build GRCh38
rs954727747 Chr.11: 113780166-113780166 on Build GRCh38
rs368118465 Chr.11: 113780008-113780008 on Build GRCh38
rs765879630 Chr.11: 113780110-113780110 on Build GRCh38
rs1044652121 Chr.11: 113780042-113780042 on Build GRCh38
rs761162254 Chr.11: 113780098-113780098 on Build GRCh38
rs893220944 Chr.11: 113779901-113779901 on Build GRCh38
rs754824119 Chr.11: 113779964-113779964 on Build GRCh38
rs1023165732 Chr.11: 113779863-113779863 on Build GRCh38
rs199984262 Chr.11: 113780150-113780150 on Build GRCh38
rs969055014 Chr.11: 113779875-113779875 on Build GRCh38
rs374252300 Chr.11: 113779763-113779763 on Build GRCh38
rs770540964 Chr.11: 113779757-113779757 on Build GRCh38
rs753506514 Chr.11: 113779830-113779830 on Build GRCh38
rs911120544 Chr.11: 113780157-113780157 on Build GRCh38
rs371018780 Chr.11: 113779816-113779816 on Build GRCh38
rs200887183 Chr.11: 113780036-113780036 on Build GRCh38
rs777892536 Chr.11: 113780067-113780067 on Build GRCh38
rs772732542 Chr.11: 113780089-113780089 on Build GRCh38
rs773169116 Chr.11: 113779877-113779877 on Build GRCh38
rs866329317 Chr.11: 113779991-113779991 on Build GRCh38
rs549200914 Chr.11: 113779933-113779934 on Build GRCh38
rs765295086 Chr.11: 113779815-113779815 on Build GRCh38
rs367566026 Chr.11: 113779829-113779829 on Build GRCh38
rs758302636 Chr.11: 113779836-113779836 on Build GRCh38
rs760135741 Chr.11: 113779986-113779986 on Build GRCh38
rs370949818 Chr.11: 113779758-113779758 on Build GRCh38
rs776813904 Chr.11: 113779809-113779809 on Build GRCh38
rs140547989 Chr.11: 113780102-113780102 on Build GRCh38
rs367712273 Chr.11: 113779924-113779924 on Build GRCh38
rs772247740 Chr.11: 113780179-113780179 on Build GRCh38
rs866477480 Chr.11: 113779841-113779841 on Build GRCh38
rs371879188 Chr.11: 113779919-113779919 on Build GRCh38
rs770971339 Chr.11: 113780075-113780075 on Build GRCh38
rs972532381 Chr.11: 113779793-113779793 on Build GRCh38
rs747871346 Chr.11: 113779967-113779967 on Build GRCh38
rs757904523 Chr.11: 113779946-113779946 on Build GRCh38
rs764847351 Chr.11: 113780005-113780005 on Build GRCh38
rs147182065 Chr.11: 113780054-113780054 on Build GRCh38
rs745321540 Chr.11: 113779834-113779834 on Build GRCh38
rs750060970 Chr.11: 113780006-113780006 on Build GRCh38
rs377711789 Chr.11: 113779773-113779773 on Build GRCh38
rs756977984 Chr.11: 113779923-113779923 on Build GRCh38
rs767488166 Chr.11: 113780132-113780132 on Build GRCh38
rs768410561 Chr.11: 113779851-113779851 on Build GRCh38
rs758997931 Chr.11: 113780129-113780129 on Build GRCh38
rs371445026 Chr.11: 113779965-113779965 on Build GRCh38
rs541402454 Chr.11: 113779878-113779878 on Build GRCh38
rs962474584 Chr.11: 113779790-113779790 on Build GRCh38
rs1054481490 Chr.11: 113779896-113779896 on Build GRCh38
rs896006391 Chr.11: 113780019-113780019 on Build GRCh38
rs753511689 Chr.11: 113780163-113780163 on Build GRCh38
rs774895409 Chr.11: 113779908-113779908 on Build GRCh38
rs201126561 Chr.11: 113780084-113780084 on Build GRCh38
rs766841470 Chr.11: 113779819-113779819 on Build GRCh38
rs962361402 Chr.11: 113780023-113780023 on Build GRCh38
rs746830020 Chr.11: 113779845-113779845 on Build GRCh38
rs781067415 Chr.11: 113780037-113780037 on Build GRCh38
rs868828897 Chr.11: 113779928-113779928 on Build GRCh38

View Full Product Details