Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125445665-125446164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ1
Entrez Gene ID
9638
Gene Name
fasciculation and elongation protein zeta 1
Gene Aliases
UNC-76
UniGene
Hs.224008
Gene Chromosome Location
Chr.11: 125445745-125496310 on Build GRCh38
Transcripts
NM_005103.4 XM_005271734.2 XM_005271735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1975554 c.1163-1G>C p.? Chr.11: 125446112-125446112 on Build GRCh38
7227824 c.1163-7G>A p.? Chr.11: 125446118-125446118 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370358975 Chr.11: 125446139-125446139 on Build GRCh38
rs771409092 Chr.11: 125446044-125446044 on Build GRCh38
rs3763 Chr.11: 125445965-125445965 on Build GRCh38
rs958001323 Chr.11: 125446007-125446007 on Build GRCh38
rs966814980 Chr.11: 125445966-125445966 on Build GRCh38
rs991049284 Chr.11: 125446012-125446012 on Build GRCh38
rs140279964 Chr.11: 125445836-125445836 on Build GRCh38
rs1006821962 Chr.11: 125445916-125445916 on Build GRCh38
rs916759713 Chr.11: 125445857-125445857 on Build GRCh38
rs532641544 Chr.11: 125445964-125445964 on Build GRCh38
rs773837751 Chr.11: 125445954-125445954 on Build GRCh38
rs765287709 Chr.11: 125446089-125446089 on Build GRCh38
rs34007049 Chr.11: 125445943-125445944 on Build GRCh38
rs978107699 Chr.11: 125445831-125445831 on Build GRCh38
rs575176579 Chr.11: 125445844-125445844 on Build GRCh38
rs976588054 Chr.11: 125445793-125445793 on Build GRCh38
rs1030395941 Chr.11: 125445823-125445823 on Build GRCh38
rs542716708 Chr.11: 125445691-125445691 on Build GRCh38
rs547232851 Chr.11: 125445875-125445875 on Build GRCh38
rs756308363 Chr.11: 125446124-125446124 on Build GRCh38
rs775326083 Chr.11: 125446072-125446072 on Build GRCh38
rs771192076 Chr.11: 125446137-125446137 on Build GRCh38
rs759179605 Chr.11: 125446064-125446064 on Build GRCh38
rs187900168 Chr.11: 125445941-125445941 on Build GRCh38
rs150225868 Chr.11: 125446118-125446118 on Build GRCh38
rs113645449 Chr.11: 125445979-125445979 on Build GRCh38
rs896890155 Chr.11: 125445892-125445892 on Build GRCh38
rs750347817 Chr.11: 125446102-125446102 on Build GRCh38
rs959642016 Chr.11: 125445924-125445924 on Build GRCh38
rs1048089414 Chr.11: 125445909-125445909 on Build GRCh38
rs544212962 Chr.11: 125445884-125445884 on Build GRCh38
rs560916786 Chr.11: 125445886-125445886 on Build GRCh38
rs1018177734 Chr.11: 125445934-125445934 on Build GRCh38
rs1048141533 Chr.11: 125446098-125446098 on Build GRCh38
rs775094507 Chr.11: 125446140-125446140 on Build GRCh38
rs565082202 Chr.11: 125445842-125445842 on Build GRCh38
rs778010518 Chr.11: 125446131-125446131 on Build GRCh38
rs748399440 Chr.11: 125446119-125446119 on Build GRCh38
rs572912864 Chr.11: 125445718-125445718 on Build GRCh38
rs866081602 Chr.11: 125446040-125446040 on Build GRCh38
rs546737323 Chr.11: 125445956-125445956 on Build GRCh38
rs925574913 Chr.11: 125445834-125445834 on Build GRCh38
rs967140041 Chr.11: 125445827-125445827 on Build GRCh38
rs565623423 Chr.11: 125445953-125445953 on Build GRCh38
rs974580803 Chr.11: 125446002-125446002 on Build GRCh38
rs1030863402 Chr.11: 125445957-125445957 on Build GRCh38
rs555067790 Chr.11: 125445992-125445992 on Build GRCh38
rs753613206 Chr.11: 125446086-125446086 on Build GRCh38
rs773735644 Chr.11: 125446054-125446054 on Build GRCh38
rs763821201 Chr.11: 125446076-125446076 on Build GRCh38
rs374802495 Chr.11: 125446143-125446143 on Build GRCh38
rs750273632 Chr.11: 125445898-125445898 on Build GRCh38
rs757148188 Chr.11: 125446088-125446088 on Build GRCh38
rs767118680 Chr.11: 125446068-125446068 on Build GRCh38
rs982294629 Chr.11: 125446065-125446065 on Build GRCh38
rs921891645 Chr.11: 125445715-125445715 on Build GRCh38
rs770039112 Chr.11: 125445940-125445940 on Build GRCh38
rs888076121 Chr.11: 125445910-125445910 on Build GRCh38
rs200854660 Chr.11: 125446133-125446133 on Build GRCh38
rs1001074771 Chr.11: 125445904-125445904 on Build GRCh38
rs1032313112 Chr.11: 125445997-125445997 on Build GRCh38
rs771990093 Chr.11: 125446122-125446123 on Build GRCh38
rs921753847 Chr.11: 125446018-125446018 on Build GRCh38
rs560624868 Chr.11: 125445962-125445962 on Build GRCh38
rs755099205 Chr.11: 125446109-125446109 on Build GRCh38
rs115757040 Chr.11: 125445692-125445692 on Build GRCh38
rs760366665 Chr.11: 125446073-125446073 on Build GRCh38

View Full Product Details