Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125452254-125452757 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FEZ1
Entrez Gene ID
9638
Gene Name
fasciculation and elongation protein zeta 1
Gene Aliases
UNC-76
UniGene
Hs.224008
Gene Chromosome Location
Chr.11: 125445745-125496310 on Build GRCh38
Transcripts
NM_005103.4 XM_005271734.2 XM_005271735.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6863939 c.1096+5G>T p.? Chr.11: 125452329-125452329 on Build GRCh38
5734166 c.1058C>A p.S353Y Chr.11: 125452372-125452372 on Build GRCh38
4965577 c.1096+8C>A p.? Chr.11: 125452326-125452326 on Build GRCh38
6531343 c.1070T>G p.V357G Chr.11: 125452360-125452360 on Build GRCh38
6891817 c.1063C>T p.P355S Chr.11: 125452367-125452367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979542734 Chr.11: 125452641-125452641 on Build GRCh38
rs917863737 Chr.11: 125452275-125452275 on Build GRCh38
rs537095838 Chr.11: 125452421-125452421 on Build GRCh38
rs1006834397 Chr.11: 125452682-125452682 on Build GRCh38
rs762801339 Chr.11: 125452396-125452396 on Build GRCh38
rs896879547 Chr.11: 125452622-125452622 on Build GRCh38
rs192854061 Chr.11: 125452662-125452662 on Build GRCh38
rs772941155 Chr.11: 125452289-125452289 on Build GRCh38
rs769660697 Chr.11: 125452285-125452285 on Build GRCh38
rs762054096 Chr.11: 125452385-125452385 on Build GRCh38
rs750753028 Chr.11: 125452344-125452344 on Build GRCh38
rs750692283 Chr.11: 125452531-125452531 on Build GRCh38
rs368811806 Chr.11: 125452468-125452468 on Build GRCh38
rs765413775 Chr.11: 125452463-125452463 on Build GRCh38
rs372289859 Chr.11: 125452312-125452312 on Build GRCh38
rs751491750 Chr.11: 125452309-125452309 on Build GRCh38
rs754125308 Chr.11: 125452320-125452320 on Build GRCh38
rs187154563 Chr.11: 125452717-125452717 on Build GRCh38
rs762889660 Chr.11: 125452293-125452293 on Build GRCh38
rs185204523 Chr.11: 125452670-125452670 on Build GRCh38
rs768252510 Chr.11: 125452362-125452362 on Build GRCh38
rs1055045670 Chr.11: 125452537-125452537 on Build GRCh38
rs952362006 Chr.11: 125452317-125452317 on Build GRCh38
rs917726458 Chr.11: 125452724-125452724 on Build GRCh38
rs113374379 Chr.11: 125452393-125452393 on Build GRCh38
rs112732577 Chr.11: 125452358-125452358 on Build GRCh38
rs147677117 Chr.11: 125452542-125452542 on Build GRCh38
rs755451038 Chr.11: 125452604-125452604 on Build GRCh38
rs1046115442 Chr.11: 125452617-125452617 on Build GRCh38
rs992426613 Chr.11: 125452673-125452673 on Build GRCh38
rs757376190 Chr.11: 125452323-125452323 on Build GRCh38
rs747858697 Chr.11: 125452370-125452370 on Build GRCh38
rs923337704 Chr.11: 125452377-125452377 on Build GRCh38
rs758831644 Chr.11: 125452353-125452353 on Build GRCh38
rs779436234 Chr.11: 125452327-125452327 on Build GRCh38
rs1042172795 Chr.11: 125452491-125452491 on Build GRCh38
rs993925757 Chr.11: 125452637-125452637 on Build GRCh38
rs766509029 Chr.11: 125452541-125452541 on Build GRCh38
rs997456078 Chr.11: 125452483-125452483 on Build GRCh38
rs763004094 Chr.11: 125452538-125452538 on Build GRCh38
rs557202039 Chr.11: 125452438-125452438 on Build GRCh38
rs1029815607 Chr.11: 125452502-125452502 on Build GRCh38
rs1020980427 Chr.11: 125452628-125452628 on Build GRCh38
rs150797411 Chr.11: 125452365-125452365 on Build GRCh38
rs985062881 Chr.11: 125452337-125452337 on Build GRCh38
rs534353743 Chr.11: 125452311-125452311 on Build GRCh38
rs554438390 Chr.11: 125452700-125452700 on Build GRCh38
rs766234927 Chr.11: 125452306-125452306 on Build GRCh38
rs925123204 Chr.11: 125452522-125452522 on Build GRCh38
rs147040802 Chr.11: 125452505-125452505 on Build GRCh38
rs769377953 Chr.11: 125452378-125452378 on Build GRCh38
rs891405548 Chr.11: 125452524-125452524 on Build GRCh38
rs142332090 Chr.11: 125452675-125452675 on Build GRCh38

View Full Product Details