Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125957370-125957906 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDON
Entrez Gene ID
50937
Gene Name
cell adhesion associated, oncogene regulated
Gene Aliases
CDO, CDON1, HPE11, ORCAM
UniGene
Hs.38034
Gene Chromosome Location
Chr.11: 125956813-126063335 on Build GRCh38
Transcripts
NM_001243597.1 NM_016952.4 XM_011542862.2 XM_011542863.2 XM_011542864.2 XM_011542865.2 XM_011542866.2 XM_017017873.1

SNP Detail

SNP ID Chromosome Location
rs574688562 Chr.11: 125957487-125957487 on Build GRCh38
rs996560312 Chr.11: 125957461-125957461 on Build GRCh38
rs369076776 Chr.11: 125957712-125957712 on Build GRCh38
rs558279401 Chr.11: 125957402-125957402 on Build GRCh38
rs935640599 Chr.11: 125957635-125957635 on Build GRCh38
rs531690322 Chr.11: 125957648-125957648 on Build GRCh38
rs911727642 Chr.11: 125957500-125957500 on Build GRCh38
rs547803371 Chr.11: 125957772-125957772 on Build GRCh38
rs978360295 Chr.11: 125957819-125957819 on Build GRCh38
rs77815115 Chr.11: 125957428-125957428 on Build GRCh38
rs966553971 Chr.11: 125957802-125957802 on Build GRCh38
rs1024446294 Chr.11: 125957688-125957688 on Build GRCh38
rs191432564 Chr.11: 125957801-125957801 on Build GRCh38
rs886047946 Chr.11: 125957732-125957732 on Build GRCh38
rs1045261392 Chr.11: 125957882-125957882 on Build GRCh38
rs145839768 Chr.11: 125957400-125957400 on Build GRCh38
rs894728448 Chr.11: 125957673-125957673 on Build GRCh38
rs1004034010 Chr.11: 125957404-125957404 on Build GRCh38
rs576469776 Chr.11: 125957599-125957599 on Build GRCh38
rs182984159 Chr.11: 125957823-125957823 on Build GRCh38
rs924376955 Chr.11: 125957847-125957847 on Build GRCh38
rs952255945 Chr.11: 125957742-125957742 on Build GRCh38
rs548586562 Chr.11: 125957717-125957717 on Build GRCh38
rs190613771 Chr.11: 125957491-125957491 on Build GRCh38
rs187258536 Chr.11: 125957766-125957766 on Build GRCh38
rs539903009 Chr.11: 125957721-125957721 on Build GRCh38
rs1015400849 Chr.11: 125957696-125957696 on Build GRCh38
rs972182465 Chr.11: 125957401-125957401 on Build GRCh38
rs1013177083 Chr.11: 125957676-125957676 on Build GRCh38
rs147791703 Chr.11: 125957518-125957518 on Build GRCh38
rs560994615 Chr.11: 125957469-125957469 on Build GRCh38
rs767240187 Chr.11: 125957782-125957782 on Build GRCh38
rs141160538 Chr.11: 125957533-125957533 on Build GRCh38
rs1054757239 Chr.11: 125957862-125957862 on Build GRCh38
rs565040160 Chr.11: 125957800-125957800 on Build GRCh38
rs886047945 Chr.11: 125957461-125957461 on Build GRCh38
rs182961107 Chr.11: 125957596-125957596 on Build GRCh38
rs936338928 Chr.11: 125957590-125957590 on Build GRCh38
rs1003644000 Chr.11: 125957691-125957691 on Build GRCh38
rs11220285 Chr.11: 125957452-125957452 on Build GRCh38
rs1045245873 Chr.11: 125957667-125957667 on Build GRCh38
rs1012445475 Chr.11: 125957656-125957656 on Build GRCh38
rs907381947 Chr.11: 125957707-125957707 on Build GRCh38
rs993965808 Chr.11: 125957711-125957711 on Build GRCh38
rs984799857 Chr.11: 125957760-125957760 on Build GRCh38
rs886047947 Chr.11: 125957809-125957809 on Build GRCh38
rs985997406 Chr.11: 125957492-125957492 on Build GRCh38
rs773724685 Chr.11: 125957512-125957512 on Build GRCh38
rs986008732 Chr.11: 125957770-125957770 on Build GRCh38
rs140155591 Chr.11: 125957857-125957860 on Build GRCh38
rs948922090 Chr.11: 125957881-125957881 on Build GRCh38
rs1014934336 Chr.11: 125957407-125957407 on Build GRCh38
rs894089594 Chr.11: 125957651-125957651 on Build GRCh38
rs886047944 Chr.11: 125957419-125957420 on Build GRCh38
rs886047948 Chr.11: 125957838-125957838 on Build GRCh38
rs763387577 Chr.11: 125957514-125957514 on Build GRCh38
rs923108388 Chr.11: 125957516-125957516 on Build GRCh38
rs1026704190 Chr.11: 125957427-125957427 on Build GRCh38
rs936458935 Chr.11: 125957834-125957834 on Build GRCh38
rs1042159508 Chr.11: 125957531-125957531 on Build GRCh38
rs73019370 Chr.11: 125957866-125957866 on Build GRCh38

View Full Product Details