Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126269272-126269704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
UniGene
Hs.317190
Gene Chromosome Location
Chr.11: 126269040-126278132 on Build GRCh38
Transcripts
NM_017547.3 NR_037647.1 NR_037648.1 XM_006718881.3 XM_011542895.2 XM_011542896.2 XM_017018000.1 XM_017018001.1 XM_017018002.1 XM_017018003.1 XM_017018004.1 XM_017018006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4390690 c.37G>A p.E13K Chr.11: 126269409-126269409 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs750902202 Chr.11: 126269356-126269356 on Build GRCh38
rs997581952 Chr.11: 126269607-126269607 on Build GRCh38
rs868435847 Chr.11: 126269531-126269531 on Build GRCh38
rs938663711 Chr.11: 126269488-126269488 on Build GRCh38
rs540583652 Chr.11: 126269530-126269530 on Build GRCh38
rs536188622 Chr.11: 126269296-126269296 on Build GRCh38
rs755886789 Chr.11: 126269451-126269451 on Build GRCh38
rs745956160 Chr.11: 126269306-126269306 on Build GRCh38
rs546838613 Chr.11: 126269301-126269301 on Build GRCh38
rs763593398 Chr.11: 126269428-126269428 on Build GRCh38
rs776915158 Chr.11: 126269341-126269341 on Build GRCh38
rs765662904 Chr.11: 126269346-126269346 on Build GRCh38
rs560808228 Chr.11: 126269538-126269538 on Build GRCh38
rs575850448 Chr.11: 126269394-126269394 on Build GRCh38
rs759082553 Chr.11: 126269435-126269435 on Build GRCh38
rs1035141232 Chr.11: 126269555-126269555 on Build GRCh38
rs758003639 Chr.11: 126269358-126269358 on Build GRCh38
rs1041283480 Chr.11: 126269560-126269560 on Build GRCh38
rs978516305 Chr.11: 126269610-126269610 on Build GRCh38
rs796162188 Chr.11: 126269507-126269507 on Build GRCh38
rs985015233 Chr.11: 126269399-126269399 on Build GRCh38
rs888842678 Chr.11: 126269680-126269680 on Build GRCh38
rs1012151948 Chr.11: 126269417-126269417 on Build GRCh38
rs968385359 Chr.11: 126269609-126269609 on Build GRCh38
rs546336544 Chr.11: 126269621-126269621 on Build GRCh38
rs768823305 Chr.11: 126269415-126269415 on Build GRCh38
rs766973553 Chr.11: 126269430-126269430 on Build GRCh38
rs781284592 Chr.11: 126269570-126269572 on Build GRCh38
rs374280595 Chr.11: 126269678-126269678 on Build GRCh38
rs1023073542 Chr.11: 126269588-126269588 on Build GRCh38
rs191342721 Chr.11: 126269548-126269548 on Build GRCh38
rs747074565 Chr.11: 126269413-126269413 on Build GRCh38
rs773196403 Chr.11: 126269302-126269302 on Build GRCh38
rs377294474 Chr.11: 126269300-126269300 on Build GRCh38
rs186530916 Chr.11: 126269360-126269360 on Build GRCh38
rs953188366 Chr.11: 126269372-126269372 on Build GRCh38
rs997234988 Chr.11: 126269347-126269347 on Build GRCh38
rs773886379 Chr.11: 126269424-126269424 on Build GRCh38
rs369871514 Chr.11: 126269317-126269317 on Build GRCh38
rs1055833856 Chr.11: 126269494-126269494 on Build GRCh38
rs371930810 Chr.11: 126269339-126269339 on Build GRCh38
rs147836146 Chr.11: 126269648-126269648 on Build GRCh38
rs749178954 Chr.11: 126269408-126269408 on Build GRCh38
rs1012483738 Chr.11: 126269571-126269571 on Build GRCh38
rs924318812 Chr.11: 126269639-126269639 on Build GRCh38
rs1786703 Chr.11: 126269418-126269418 on Build GRCh38
rs762229626 Chr.11: 126269344-126269344 on Build GRCh38
rs555034172 Chr.11: 126269422-126269422 on Build GRCh38
rs943286342 Chr.11: 126269403-126269403 on Build GRCh38
rs755876630 Chr.11: 126269382-126269382 on Build GRCh38
rs771833596 Chr.11: 126269397-126269398 on Build GRCh38
rs767081061 Chr.11: 126269437-126269437 on Build GRCh38
rs965691143 Chr.11: 126269328-126269328 on Build GRCh38
rs760849767 Chr.11: 126269334-126269334 on Build GRCh38
rs751174601 Chr.11: 126269433-126269433 on Build GRCh38
rs780154875 Chr.11: 126269411-126269411 on Build GRCh38
rs747960550 Chr.11: 126269366-126269366 on Build GRCh38
rs759367781 Chr.11: 126269340-126269341 on Build GRCh38
rs574757590 Chr.11: 126269509-126269509 on Build GRCh38
rs928582995 Chr.11: 126269441-126269441 on Build GRCh38
rs566573657 Chr.11: 126269327-126269327 on Build GRCh38
rs1047768848 Chr.11: 126269449-126269449 on Build GRCh38
rs959922073 Chr.11: 126269568-126269568 on Build GRCh38
rs901472127 Chr.11: 126269580-126269580 on Build GRCh38
rs934879497 Chr.11: 126269551-126269551 on Build GRCh38
rs752340687 Chr.11: 126269440-126269440 on Build GRCh38
rs1030594324 Chr.11: 126269314-126269314 on Build GRCh38
rs911837626 Chr.11: 126269376-126269377 on Build GRCh38
rs779478966 Chr.11: 126269657-126269657 on Build GRCh38
rs776842603 Chr.11: 126269416-126269416 on Build GRCh38
rs947518599 Chr.11: 126269537-126269537 on Build GRCh38
rs754729021 Chr.11: 126269365-126269365 on Build GRCh38

View Full Product Details