Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126272890-126273385 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
UniGene
Hs.317190
Gene Chromosome Location
Chr.11: 126269040-126278132 on Build GRCh38
Transcripts
NM_017547.3 NR_037647.1 NR_037648.1 XM_006718879.3 XM_006718881.3 XM_011542895.2 XM_011542896.2 XM_017018000.1 XM_017018001.1 XM_017018002.1 XM_017018003.1 XM_017018004.1 XM_017018005.1 XM_017018006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5487648 c.388G>A p.V130I Chr.11: 126273348-126273348 on Build GRCh38
5578597 c.424C>T p.P142S Chr.11: 126273384-126273384 on Build GRCh38
6666581 c.406C>T p.R136W Chr.11: 126273068-126273068 on Build GRCh38
5487647 c.430G>A p.V144I Chr.11: 126273348-126273348 on Build GRCh38
5578596 c.466C>T p.P156S Chr.11: 126273384-126273384 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1004494741 Chr.11: 126273340-126273340 on Build GRCh38
rs541684918 Chr.11: 126272955-126272955 on Build GRCh38
rs759917961 Chr.11: 126273363-126273363 on Build GRCh38
rs951249107 Chr.11: 126272994-126272994 on Build GRCh38
rs766610401 Chr.11: 126273293-126273293 on Build GRCh38
rs753941444 Chr.11: 126272937-126272937 on Build GRCh38
rs2298453 Chr.11: 126273202-126273202 on Build GRCh38
rs748813627 Chr.11: 126273324-126273324 on Build GRCh38
rs774342140 Chr.11: 126273121-126273121 on Build GRCh38
rs112855709 Chr.11: 126273289-126273289 on Build GRCh38
rs746956683 Chr.11: 126273347-126273347 on Build GRCh38
rs751899225 Chr.11: 126273078-126273078 on Build GRCh38
rs180800246 Chr.11: 126273089-126273089 on Build GRCh38
rs762004291 Chr.11: 126273277-126273277 on Build GRCh38
rs577380096 Chr.11: 126273100-126273100 on Build GRCh38
rs755681660 Chr.11: 126273320-126273320 on Build GRCh38
rs756285292 Chr.11: 126272933-126272933 on Build GRCh38
rs995269350 Chr.11: 126273230-126273230 on Build GRCh38
rs1039805478 Chr.11: 126273207-126273207 on Build GRCh38
rs34542988 Chr.11: 126273351-126273351 on Build GRCh38
rs185293323 Chr.11: 126272989-126272989 on Build GRCh38
rs886944589 Chr.11: 126273333-126273333 on Build GRCh38
rs927432478 Chr.11: 126273182-126273182 on Build GRCh38
rs879515535 Chr.11: 126273195-126273196 on Build GRCh38
rs747471338 Chr.11: 126272949-126272949 on Build GRCh38
rs754006850 Chr.11: 126273337-126273337 on Build GRCh38
rs777189187 Chr.11: 126273321-126273321 on Build GRCh38
rs563391934 Chr.11: 126273195-126273195 on Build GRCh38
rs147235743 Chr.11: 126273353-126273353 on Build GRCh38
rs1052024776 Chr.11: 126273150-126273150 on Build GRCh38
rs770589780 Chr.11: 126273108-126273108 on Build GRCh38
rs138188150 Chr.11: 126273074-126273074 on Build GRCh38
rs937590657 Chr.11: 126273194-126273194 on Build GRCh38
rs754032276 Chr.11: 126273049-126273049 on Build GRCh38
rs759591132 Chr.11: 126273022-126273022 on Build GRCh38
rs757604987 Chr.11: 126272939-126272939 on Build GRCh38
rs535066892 Chr.11: 126273029-126273029 on Build GRCh38
rs995326316 Chr.11: 126272946-126272946 on Build GRCh38
rs763082363 Chr.11: 126273354-126273354 on Build GRCh38
rs950167546 Chr.11: 126273136-126273136 on Build GRCh38
rs113976905 Chr.11: 126273095-126273095 on Build GRCh38
rs748853628 Chr.11: 126273098-126273098 on Build GRCh38
rs765187255 Chr.11: 126273287-126273287 on Build GRCh38
rs1045925344 Chr.11: 126273156-126273156 on Build GRCh38
rs2298454 Chr.11: 126273236-126273236 on Build GRCh38
rs1004141278 Chr.11: 126273011-126273011 on Build GRCh38
rs769120291 Chr.11: 126272950-126272950 on Build GRCh38
rs367645630 Chr.11: 126272935-126272935 on Build GRCh38
rs765506915 Chr.11: 126273067-126273067 on Build GRCh38
rs755535853 Chr.11: 126273084-126273084 on Build GRCh38
rs751985297 Chr.11: 126273295-126273295 on Build GRCh38
rs756921599 Chr.11: 126273329-126273329 on Build GRCh38
rs375413966 Chr.11: 126273309-126273309 on Build GRCh38
rs774500202 Chr.11: 126273359-126273359 on Build GRCh38
rs543529584 Chr.11: 126273184-126273184 on Build GRCh38
rs771941278 Chr.11: 126273335-126273335 on Build GRCh38
rs945595564 Chr.11: 126273148-126273148 on Build GRCh38
rs777504068 Chr.11: 126273097-126273097 on Build GRCh38
rs764093077 Chr.11: 126272922-126272923 on Build GRCh38
rs775507828 Chr.11: 126273276-126273276 on Build GRCh38
rs761834891 Chr.11: 126272942-126272942 on Build GRCh38
rs762822720 Chr.11: 126272996-126272996 on Build GRCh38
rs943708416 Chr.11: 126273206-126273206 on Build GRCh38
rs75183025 Chr.11: 126273057-126273057 on Build GRCh38
rs778707231 Chr.11: 126273330-126273330 on Build GRCh38
rs878971256 Chr.11: 126273308-126273308 on Build GRCh38
rs531412126 Chr.11: 126273366-126273366 on Build GRCh38
rs780345055 Chr.11: 126273331-126273331 on Build GRCh38
rs181493362 Chr.11: 126272941-126272941 on Build GRCh38
rs375883538 Chr.11: 126272931-126272931 on Build GRCh38
rs547461665 Chr.11: 126273209-126273210 on Build GRCh38
rs775595762 Chr.11: 126273341-126273341 on Build GRCh38
rs755266959 Chr.11: 126273306-126273306 on Build GRCh38
rs750482528 Chr.11: 126273288-126273288 on Build GRCh38
rs769554446 Chr.11: 126272976-126272976 on Build GRCh38
rs556261091 Chr.11: 126272998-126272998 on Build GRCh38
rs201873350 Chr.11: 126273355-126273355 on Build GRCh38
rs953976894 Chr.11: 126273266-126273266 on Build GRCh38
rs1029510972 Chr.11: 126273260-126273260 on Build GRCh38
rs863224018 Chr.11: 126273352-126273352 on Build GRCh38
rs758629736 Chr.11: 126272944-126272944 on Build GRCh38
rs899560769 Chr.11: 126273229-126273229 on Build GRCh38
rs1016844031 Chr.11: 126273296-126273296 on Build GRCh38
rs753156614 Chr.11: 126273313-126273313 on Build GRCh38
rs747641223 Chr.11: 126272960-126272960 on Build GRCh38
rs376227631 Chr.11: 126273365-126273365 on Build GRCh38
rs189739398 Chr.11: 126273015-126273015 on Build GRCh38
rs373075574 Chr.11: 126273068-126273068 on Build GRCh38
rs780629612 Chr.11: 126272948-126272948 on Build GRCh38
rs758819066 Chr.11: 126273069-126273069 on Build GRCh38
rs375703314 Chr.11: 126273273-126273273 on Build GRCh38

View Full Product Details