Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 126275923-126276452 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FOXRED1
Entrez Gene ID
55572
Gene Name
FAD dependent oxidoreductase domain containing 1
Gene Aliases
FP634, H17
UniGene
Hs.317190
Gene Chromosome Location
Chr.11: 126269040-126278132 on Build GRCh38
Transcripts
NM_017547.3 NR_037647.1 NR_037648.1 XM_006718879.3 XM_006718881.3 XM_011542895.2 XM_011542896.2 XM_017018000.1 XM_017018001.1 XM_017018002.1 XM_017018003.1 XM_017018004.1 XM_017018005.1 XM_017018006.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5620842 c.794A>G p.Y265C Chr.11: 126276084-126276084 on Build GRCh38
5361157 c.866A>G p.N289S Chr.11: 126276114-126276114 on Build GRCh38
1352425 c.962C>T p.P321L Chr.11: 126276210-126276210 on Build GRCh38
5633448 c.831C>T p.A277A Chr.11: 126276121-126276121 on Build GRCh38
6505215 c.859G>A p.V287M Chr.11: 126276107-126276107 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376595518 Chr.11: 126276315-126276316 on Build GRCh38
rs777315728 Chr.11: 126276169-126276169 on Build GRCh38
rs761993742 Chr.11: 126276084-126276084 on Build GRCh38
rs200551914 Chr.11: 126276115-126276115 on Build GRCh38
rs56321210 Chr.11: 126276349-126276349 on Build GRCh38
rs748195712 Chr.11: 126276200-126276200 on Build GRCh38
rs780616719 Chr.11: 126276116-126276116 on Build GRCh38
rs776400237 Chr.11: 126276369-126276396 on Build GRCh38
rs369592628 Chr.11: 126276320-126276320 on Build GRCh38
rs111388246 Chr.11: 126276313-126276313 on Build GRCh38
rs765356156 Chr.11: 126276382-126276383 on Build GRCh38
rs373422248 Chr.11: 126276321-126276321 on Build GRCh38
rs149010068 Chr.11: 126276428-126276428 on Build GRCh38
rs867067790 Chr.11: 126276321-126276321 on Build GRCh38
rs749110767 Chr.11: 126276122-126276122 on Build GRCh38
rs750702096 Chr.11: 126276388-126276388 on Build GRCh38
rs763336673 Chr.11: 126276144-126276144 on Build GRCh38
rs780468984 Chr.11: 126276027-126276027 on Build GRCh38
rs111357379 Chr.11: 126276317-126276317 on Build GRCh38
rs781102596 Chr.11: 126276391-126276391 on Build GRCh38
rs778903746 Chr.11: 126276189-126276189 on Build GRCh38
rs779145530 Chr.11: 126276397-126276397 on Build GRCh38
rs775933882 Chr.11: 126276402-126276402 on Build GRCh38
rs142440426 Chr.11: 126276315-126276317 on Build GRCh38
rs760282774 Chr.11: 126276325-126276325 on Build GRCh38
rs766220242 Chr.11: 126276386-126276386 on Build GRCh38
rs888954197 Chr.11: 126276132-126276132 on Build GRCh38
rs752020360 Chr.11: 126276109-126276109 on Build GRCh38
rs754947608 Chr.11: 126276389-126276389 on Build GRCh38
rs770317136 Chr.11: 126276247-126276248 on Build GRCh38
rs781541770 Chr.11: 126276227-126276227 on Build GRCh38
rs368695946 Chr.11: 126276356-126276356 on Build GRCh38
rs868038947 Chr.11: 126276316-126276316 on Build GRCh38
rs772443706 Chr.11: 126276363-126276363 on Build GRCh38
rs1008727781 Chr.11: 126276060-126276060 on Build GRCh38
rs758160028 Chr.11: 126276263-126276263 on Build GRCh38
rs11220434 Chr.11: 126276235-126276235 on Build GRCh38
rs753379714 Chr.11: 126276233-126276233 on Build GRCh38
rs559064999 Chr.11: 126276121-126276121 on Build GRCh38
rs755536248 Chr.11: 126276155-126276155 on Build GRCh38
rs974737910 Chr.11: 126276120-126276120 on Build GRCh38
rs112008079 Chr.11: 126276286-126276286 on Build GRCh38
rs145888229 Chr.11: 126276168-126276168 on Build GRCh38
rs1043289472 Chr.11: 126276020-126276020 on Build GRCh38
rs760372928 Chr.11: 126276363-126276371 on Build GRCh38
rs760863038 Chr.11: 126276072-126276072 on Build GRCh38
rs755790428 Chr.11: 126276118-126276118 on Build GRCh38
rs369938396 Chr.11: 126276300-126276300 on Build GRCh38
rs779189406 Chr.11: 126276023-126276023 on Build GRCh38
rs780823573 Chr.11: 126276319-126276320 on Build GRCh38
rs771128237 Chr.11: 126276210-126276210 on Build GRCh38
rs775689348 Chr.11: 126276367-126276367 on Build GRCh38
rs761399539 Chr.11: 126276374-126276374 on Build GRCh38
rs772134174 Chr.11: 126276398-126276398 on Build GRCh38
rs35097682 Chr.11: 126276421-126276421 on Build GRCh38
rs997056117 Chr.11: 126276320-126276320 on Build GRCh38
rs1056638868 Chr.11: 126276228-126276228 on Build GRCh38
rs777259232 Chr.11: 126276415-126276415 on Build GRCh38
rs776744235 Chr.11: 126276073-126276073 on Build GRCh38
rs769876098 Chr.11: 126276203-126276203 on Build GRCh38
rs564591840 Chr.11: 126276410-126276410 on Build GRCh38
rs774764033 Chr.11: 126276211-126276211 on Build GRCh38
rs759879360 Chr.11: 126276223-126276223 on Build GRCh38
rs999958415 Chr.11: 126276351-126276351 on Build GRCh38
rs761263442 Chr.11: 126276370-126276370 on Build GRCh38
rs12804446 Chr.11: 126276323-126276323 on Build GRCh38
rs766865608 Chr.11: 126276394-126276395 on Build GRCh38
rs753201206 Chr.11: 126276227-126276227 on Build GRCh38
rs567511779 Chr.11: 126276032-126276032 on Build GRCh38
rs766891622 Chr.11: 126276107-126276107 on Build GRCh38
rs775259906 Chr.11: 126276308-126276308 on Build GRCh38
rs377120102 Chr.11: 126276328-126276328 on Build GRCh38
rs768705873 Chr.11: 126276366-126276366 on Build GRCh38
rs771886524 Chr.11: 126276348-126276354 on Build GRCh38
rs773431898 Chr.11: 126276143-126276143 on Build GRCh38
rs201751002 Chr.11: 126276324-126276324 on Build GRCh38
rs771903600 Chr.11: 126276192-126276192 on Build GRCh38
rs585209 Chr.11: 126276108-126276108 on Build GRCh38
rs149851763 Chr.11: 126276147-126276147 on Build GRCh38
rs763203813 Chr.11: 126276390-126276396 on Build GRCh38
rs750252540 Chr.11: 126276249-126276249 on Build GRCh38
rs1018849379 Chr.11: 126276114-126276114 on Build GRCh38
rs148955548 Chr.11: 126276105-126276105 on Build GRCh38
rs765488299 Chr.11: 126276089-126276089 on Build GRCh38
rs1000331906 Chr.11: 126275983-126275983 on Build GRCh38
rs375044137 Chr.11: 126276392-126276393 on Build GRCh38
rs374816209 Chr.11: 126275960-126275960 on Build GRCh38
rs770998661 Chr.11: 126276057-126276057 on Build GRCh38
rs758967393 Chr.11: 126276024-126276024 on Build GRCh38
rs374968241 Chr.11: 126276074-126276074 on Build GRCh38
rs2276025 Chr.11: 126276036-126276036 on Build GRCh38
rs374211633 Chr.11: 126276232-126276232 on Build GRCh38
rs886047982 Chr.11: 126276135-126276135 on Build GRCh38
rs757950808 Chr.11: 126276384-126276384 on Build GRCh38
rs770802832 Chr.11: 126276125-126276125 on Build GRCh38
rs775205671 Chr.11: 126276348-126276357 on Build GRCh38
rs764634373 Chr.11: 126276371-126276371 on Build GRCh38
rs904297729 Chr.11: 126275957-126275957 on Build GRCh38
rs753504174 Chr.11: 126276172-126276172 on Build GRCh38
rs991538302 Chr.11: 126276309-126276309 on Build GRCh38
rs541052622 Chr.11: 126276229-126276229 on Build GRCh38
rs367938377 Chr.11: 126276142-126276142 on Build GRCh38
rs754433442 Chr.11: 126276029-126276029 on Build GRCh38
rs779980682 Chr.11: 126276193-126276193 on Build GRCh38
rs370131645 Chr.11: 126276053-126276053 on Build GRCh38
rs758701557 Chr.11: 126276103-126276103 on Build GRCh38
rs78535068 Chr.11: 126276331-126276331 on Build GRCh38
rs753398194 Chr.11: 126276372-126276372 on Build GRCh38
rs796575723 Chr.11: 126276346-126276349 on Build GRCh38
rs752023793 Chr.11: 126276396-126276397 on Build GRCh38
rs886047981 Chr.11: 126276081-126276081 on Build GRCh38
rs78997120 Chr.11: 126276362-126276362 on Build GRCh38
rs745844423 Chr.11: 126276131-126276131 on Build GRCh38
rs775406644 Chr.11: 126276171-126276171 on Build GRCh38
rs761882501 Chr.11: 126276139-126276139 on Build GRCh38
rs369670355 Chr.11: 126276377-126276377 on Build GRCh38
rs772938167 Chr.11: 126276043-126276043 on Build GRCh38
rs751255962 Chr.11: 126276388-126276388 on Build GRCh38
rs904159611 Chr.11: 126276296-126276296 on Build GRCh38
rs779885096 Chr.11: 126276348-126276348 on Build GRCh38
rs772028465 Chr.11: 126276136-126276136 on Build GRCh38
rs111338847 Chr.11: 126276191-126276191 on Build GRCh38
rs550604507 Chr.11: 126275954-126275954 on Build GRCh38
rs755297740 Chr.11: 126276415-126276415 on Build GRCh38
rs544726923 Chr.11: 126276396-126276396 on Build GRCh38
rs151046752 Chr.11: 126276246-126276246 on Build GRCh38
rs756941477 Chr.11: 126276181-126276181 on Build GRCh38
rs771890299 Chr.11: 126276306-126276306 on Build GRCh38
rs745475967 Chr.11: 126276345-126276346 on Build GRCh38
rs774375087 Chr.11: 126276071-126276071 on Build GRCh38
rs761997363 Chr.11: 126276375-126276381 on Build GRCh38
rs193267014 Chr.11: 126276287-126276287 on Build GRCh38
rs747681462 Chr.11: 126276117-126276117 on Build GRCh38
rs546801411 Chr.11: 126276100-126276100 on Build GRCh38
rs371937924 Chr.11: 126276226-126276226 on Build GRCh38
rs748899557 Chr.11: 126276246-126276246 on Build GRCh38
rs768389177 Chr.11: 126276254-126276257 on Build GRCh38
rs373491328 Chr.11: 126276378-126276378 on Build GRCh38
rs774500144 Chr.11: 126276361-126276361 on Build GRCh38
rs556202062 Chr.11: 126276292-126276293 on Build GRCh38
rs554655060 Chr.11: 126276209-126276209 on Build GRCh38
rs768064354 Chr.11: 126276157-126276157 on Build GRCh38
rs761452775 Chr.11: 126276420-126276420 on Build GRCh38
rs201062084 Chr.11: 126276395-126276395 on Build GRCh38
rs757675516 Chr.11: 126276019-126276019 on Build GRCh38
rs372184824 Chr.11: 126276314-126276314 on Build GRCh38
rs769464542 Chr.11: 126276037-126276037 on Build GRCh38
rs771146646 Chr.11: 126276357-126276357 on Build GRCh38

View Full Product Details