Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128693795-128694288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLI1
Entrez Gene ID
2313
Gene Name
Fli-1 proto-oncogene, ETS transcription factor
Gene Aliases
EWSR2, SIC-1
UniGene
Hs.504281
Gene Chromosome Location
Chr.11: 128685263-128813267 on Build GRCh38
Transcripts
NM_001167681.2 NM_001271010.1 NM_001271012.1 NM_002017.4 XM_011542701.2 XM_011542702.1 XM_017017405.1 XM_017017406.1 NR_038908.1

Gene Symbol
SENCR
Entrez Gene ID
100507392
Gene Name
smooth muscle and endothelial cell enriched migration/differentiation-associated long non-coding RNA
Gene Aliases
FLI1-AS1, lncRNA9
UniGene
Hs.657715
Gene Chromosome Location
Chr.11: 128691672-128696023 on Build GRCh38
Transcripts
NM_001167681.2 NM_001271010.1 NM_001271012.1 NM_002017.4 XM_011542701.2 XM_011542702.1 XM_017017405.1 XM_017017406.1 NR_038908.1

SNP Detail

SNP ID Chromosome Location
rs140864091 Chr.11: 128693831-128693831 on Build GRCh38
rs773038460 Chr.11: 128693942-128693945 on Build GRCh38
rs886585548 Chr.11: 128693993-128693993 on Build GRCh38
rs942335121 Chr.11: 128693941-128693941 on Build GRCh38
rs762436916 Chr.11: 128693944-128693961 on Build GRCh38
rs1010936809 Chr.11: 128693901-128693912 on Build GRCh38
rs867247972 Chr.11: 128693883-128693883 on Build GRCh38
rs145654087 Chr.11: 128694173-128694173 on Build GRCh38
rs766178380 Chr.11: 128694245-128694245 on Build GRCh38
rs746564927 Chr.11: 128694231-128694232 on Build GRCh38
rs756942288 Chr.11: 128693956-128693961 on Build GRCh38
rs543152818 Chr.11: 128693923-128693923 on Build GRCh38
rs987628233 Chr.11: 128693932-128693932 on Build GRCh38
rs535649413 Chr.11: 128693872-128693872 on Build GRCh38
rs770865917 Chr.11: 128693892-128693892 on Build GRCh38
rs1001533802 Chr.11: 128694043-128694043 on Build GRCh38
rs112860445 Chr.11: 128694231-128694231 on Build GRCh38
rs970333531 Chr.11: 128694034-128694034 on Build GRCh38
rs60096045 Chr.11: 128693861-128693862 on Build GRCh38
rs752471336 Chr.11: 128694266-128694266 on Build GRCh38
rs936109912 Chr.11: 128693845-128693845 on Build GRCh38
rs913334472 Chr.11: 128694117-128694117 on Build GRCh38
rs556987904 Chr.11: 128693944-128693944 on Build GRCh38
rs955665819 Chr.11: 128693926-128693928 on Build GRCh38
rs970425743 Chr.11: 128693836-128693836 on Build GRCh38
rs1047214343 Chr.11: 128694236-128694236 on Build GRCh38
rs372817629 Chr.11: 128694209-128694209 on Build GRCh38
rs767250889 Chr.11: 128694265-128694265 on Build GRCh38
rs142892110 Chr.11: 128693882-128693882 on Build GRCh38
rs779080271 Chr.11: 128694159-128694159 on Build GRCh38
rs1024580864 Chr.11: 128693832-128693832 on Build GRCh38
rs68049599 Chr.11: 128693992-128693993 on Build GRCh38
rs921542376 Chr.11: 128693996-128693996 on Build GRCh38
rs759952158 Chr.11: 128693942-128693961 on Build GRCh38
rs968621853 Chr.11: 128693856-128693856 on Build GRCh38
rs986657774 Chr.11: 128694166-128694166 on Build GRCh38
rs1057464718 Chr.11: 128694133-128694133 on Build GRCh38
rs1014428809 Chr.11: 128694014-128694014 on Build GRCh38
rs1046826186 Chr.11: 128693992-128693992 on Build GRCh38
rs573766725 Chr.11: 128694036-128694036 on Build GRCh38
rs889711931 Chr.11: 128693937-128693937 on Build GRCh38
rs957428954 Chr.11: 128694105-128694105 on Build GRCh38
rs1031551701 Chr.11: 128694242-128694242 on Build GRCh38
rs577175570 Chr.11: 128693902-128693902 on Build GRCh38
rs1037979628 Chr.11: 128694210-128694210 on Build GRCh38
rs774127474 Chr.11: 128694247-128694247 on Build GRCh38
rs942657889 Chr.11: 128694202-128694202 on Build GRCh38
rs1012718724 Chr.11: 128693833-128693833 on Build GRCh38
rs1038032017 Chr.11: 128693970-128693970 on Build GRCh38
rs762545912 Chr.11: 128694235-128694235 on Build GRCh38
rs748604139 Chr.11: 128694227-128694227 on Build GRCh38
rs1024944760 Chr.11: 128694028-128694028 on Build GRCh38
rs759182658 Chr.11: 128694255-128694255 on Build GRCh38
rs1051428867 Chr.11: 128693910-128693910 on Build GRCh38
rs536013878 Chr.11: 128693835-128693835 on Build GRCh38
rs887128453 Chr.11: 128694056-128694056 on Build GRCh38
rs955904174 Chr.11: 128694267-128694267 on Build GRCh38
rs988624647 Chr.11: 128693868-128693868 on Build GRCh38
rs919567291 Chr.11: 128694211-128694211 on Build GRCh38
rs778508207 Chr.11: 128694087-128694089 on Build GRCh38
rs879162296 Chr.11: 128693987-128693987 on Build GRCh38
rs898245794 Chr.11: 128693976-128693976 on Build GRCh38
rs57930585 Chr.11: 128693968-128693993 on Build GRCh38
rs376557878 Chr.11: 128694230-128694230 on Build GRCh38
rs989273541 Chr.11: 128694107-128694107 on Build GRCh38
rs544347560 Chr.11: 128693942-128693959 on Build GRCh38
rs149751111 Chr.11: 128693840-128693840 on Build GRCh38
rs1033034681 Chr.11: 128694087-128694087 on Build GRCh38
rs980107695 Chr.11: 128693837-128693837 on Build GRCh38
rs975604157 Chr.11: 128693989-128693989 on Build GRCh38
rs55635129 Chr.11: 128693882-128693882 on Build GRCh38
rs542842246 Chr.11: 128694128-128694128 on Build GRCh38
rs398018021 Chr.11: 128693889-128693889 on Build GRCh38
rs566061928 Chr.11: 128693852-128693852 on Build GRCh38
rs772912959 Chr.11: 128694233-128694233 on Build GRCh38

View Full Product Details