Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 128900636-128901146 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf45
Entrez Gene ID
219833
Gene Name
chromosome 11 open reading frame 45
Gene Aliases
-
UniGene
Hs.689860
Gene Chromosome Location
Chr.11: 128899565-128909520 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

Gene Symbol
KCNJ5
Entrez Gene ID
3762
Gene Name
potassium voltage-gated channel subfamily J member 5
Gene Aliases
CIR, GIRK4, KATP1, KIR3.4, LQT13
UniGene
Hs.444595
Gene Chromosome Location
Chr.11: 128891418-128921163 on Build GRCh38
Transcripts
NM_001256088.1 NM_145013.2 NR_045767.1 XM_011542654.2 XM_011542655.2 NM_000890.3

SNP Detail

SNP ID Chromosome Location
rs940016357 Chr.11: 128900731-128900731 on Build GRCh38
rs184454632 Chr.11: 128900680-128900680 on Build GRCh38
rs1052813665 Chr.11: 128900860-128900860 on Build GRCh38
rs917263693 Chr.11: 128900977-128900977 on Build GRCh38
rs995309879 Chr.11: 128900691-128900691 on Build GRCh38
rs908521169 Chr.11: 128900703-128900703 on Build GRCh38
rs763992892 Chr.11: 128900994-128900994 on Build GRCh38
rs948572793 Chr.11: 128900780-128900780 on Build GRCh38
rs945538585 Chr.11: 128900929-128900929 on Build GRCh38
rs78289503 Chr.11: 128900926-128900926 on Build GRCh38
rs969921728 Chr.11: 128900794-128900794 on Build GRCh38
rs1008540056 Chr.11: 128900873-128900873 on Build GRCh38
rs1014052176 Chr.11: 128900767-128900767 on Build GRCh38
rs760670581 Chr.11: 128900942-128900942 on Build GRCh38
rs970338969 Chr.11: 128901001-128901001 on Build GRCh38
rs1048281517 Chr.11: 128901064-128901064 on Build GRCh38
rs891446459 Chr.11: 128900872-128900872 on Build GRCh38
rs935863880 Chr.11: 128901010-128901010 on Build GRCh38
rs555342947 Chr.11: 128900878-128900878 on Build GRCh38
rs957182839 Chr.11: 128900876-128900876 on Build GRCh38
rs904358358 Chr.11: 128900802-128900802 on Build GRCh38
rs912160553 Chr.11: 128900968-128900968 on Build GRCh38
rs868389814 Chr.11: 128901110-128901110 on Build GRCh38
rs542142692 Chr.11: 128900949-128900949 on Build GRCh38
rs546951768 Chr.11: 128900885-128900885 on Build GRCh38
rs544819855 Chr.11: 128900875-128900875 on Build GRCh38
rs1005610862 Chr.11: 128900939-128900939 on Build GRCh38
rs1026183682 Chr.11: 128900722-128900722 on Build GRCh38
rs952929639 Chr.11: 128900935-128900935 on Build GRCh38
rs1032622168 Chr.11: 128900863-128900863 on Build GRCh38
rs552949720 Chr.11: 128900681-128900681 on Build GRCh38
rs201548686 Chr.11: 128900835-128900835 on Build GRCh38
rs899159947 Chr.11: 128901045-128901045 on Build GRCh38
rs943586535 Chr.11: 128901002-128901002 on Build GRCh38
rs147761572 Chr.11: 128900739-128900739 on Build GRCh38
rs980061409 Chr.11: 128901005-128901005 on Build GRCh38
rs992732216 Chr.11: 128900760-128900760 on Build GRCh38
rs76762583 Chr.11: 128900689-128900689 on Build GRCh38
rs988959907 Chr.11: 128900925-128900925 on Build GRCh38
rs925748514 Chr.11: 128901006-128901006 on Build GRCh38
rs1044587719 Chr.11: 128900798-128900798 on Build GRCh38
rs901458447 Chr.11: 128900924-128900924 on Build GRCh38
rs1015715621 Chr.11: 128900940-128900940 on Build GRCh38
rs575825025 Chr.11: 128900810-128900810 on Build GRCh38
rs930776539 Chr.11: 128901060-128901060 on Build GRCh38
rs560108128 Chr.11: 128901097-128901097 on Build GRCh38
rs1019820214 Chr.11: 128900910-128900910 on Build GRCh38
rs1024195953 Chr.11: 128900774-128900774 on Build GRCh38
rs913020995 Chr.11: 128901083-128901083 on Build GRCh38
rs1039076674 Chr.11: 128901026-128901026 on Build GRCh38
rs189775506 Chr.11: 128900755-128900755 on Build GRCh38

View Full Product Details