Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 974729-975237 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PERM1
Entrez Gene ID
84808
Gene Name
PPARGC1 and ESRR induced regulator, muscle 1
Gene Aliases
C1orf170
UniGene
Hs.271462
Gene Chromosome Location
Chr.1: 975198-982117 on Build GRCh38
Transcripts
NM_001291366.1 NM_001291367.1 XM_017002583.1 XM_017002584.1 XM_017002585.1 NM_001160184.1 NM_032129.2

Gene Symbol
PLEKHN1
Entrez Gene ID
84069
Gene Name
pleckstrin homology domain containing N1
Gene Aliases
-
UniGene
Hs.528691
Gene Chromosome Location
Chr.1: 965820-975108 on Build GRCh38
Transcripts
NM_001291366.1 NM_001291367.1 XM_017002583.1 XM_017002584.1 XM_017002585.1 NM_001160184.1 NM_032129.2

SNP Detail

SNP ID Chromosome Location
rs200964719 Chr.1: 975108-975108 on Build GRCh38
rs184104311 Chr.1: 975053-975053 on Build GRCh38
rs538116413 Chr.1: 974799-974799 on Build GRCh38
rs999350481 Chr.1: 975138-975149 on Build GRCh38
rs964798643 Chr.1: 974947-974947 on Build GRCh38
rs1011936257 Chr.1: 974756-974756 on Build GRCh38
rs556301852 Chr.1: 974929-974929 on Build GRCh38
rs6685581 Chr.1: 975058-975058 on Build GRCh38
rs954635450 Chr.1: 974981-974981 on Build GRCh38
rs1053507856 Chr.1: 974905-974905 on Build GRCh38
rs893905076 Chr.1: 975141-975147 on Build GRCh38
rs188543497 Chr.1: 975054-975054 on Build GRCh38
rs774489919 Chr.1: 975114-975115 on Build GRCh38
rs902415245 Chr.1: 975194-975194 on Build GRCh38
rs975155781 Chr.1: 974822-974822 on Build GRCh38
rs927738868 Chr.1: 975165-975165 on Build GRCh38
rs140154241 Chr.1: 975065-975065 on Build GRCh38
rs1016426612 Chr.1: 975074-975074 on Build GRCh38
rs921639155 Chr.1: 975179-975179 on Build GRCh38
rs374400565 Chr.1: 974901-974901 on Build GRCh38
rs978383087 Chr.1: 974983-974983 on Build GRCh38
rs982395062 Chr.1: 975159-975161 on Build GRCh38
rs957982260 Chr.1: 975147-975147 on Build GRCh38
rs765381441 Chr.1: 974843-974843 on Build GRCh38
rs564430369 Chr.1: 975085-975085 on Build GRCh38
rs997284120 Chr.1: 974844-974844 on Build GRCh38
rs1053655358 Chr.1: 975023-975023 on Build GRCh38
rs991556864 Chr.1: 975125-975141 on Build GRCh38
rs796451195 Chr.1: 975056-975058 on Build GRCh38
rs553895641 Chr.1: 974906-974906 on Build GRCh38
rs577628191 Chr.1: 975103-975104 on Build GRCh38
rs28477686 Chr.1: 975014-975014 on Build GRCh38
rs555848153 Chr.1: 974926-974927 on Build GRCh38
rs886689066 Chr.1: 975198-975198 on Build GRCh38
rs386627422 Chr.1: 975058-975093 on Build GRCh38
rs923289231 Chr.1: 974965-974965 on Build GRCh38
rs1046614617 Chr.1: 974926-974929 on Build GRCh38
rs558937500 Chr.1: 974966-974966 on Build GRCh38
rs1032870474 Chr.1: 975140-975146 on Build GRCh38
rs984928311 Chr.1: 974923-974923 on Build GRCh38
rs916332639 Chr.1: 975025-975025 on Build GRCh38
rs201771422 Chr.1: 975168-975169 on Build GRCh38
rs954847497 Chr.1: 975109-975109 on Build GRCh38
rs760587803 Chr.1: 974964-974964 on Build GRCh38
rs111470767 Chr.1: 974811-974811 on Build GRCh38
rs539677168 Chr.1: 974919-974919 on Build GRCh38
rs1051656998 Chr.1: 975192-975195 on Build GRCh38
rs377110644 Chr.1: 975057-975057 on Build GRCh38
rs950863341 Chr.1: 974922-974922 on Build GRCh38
rs1006557805 Chr.1: 974831-974831 on Build GRCh38
rs372814562 Chr.1: 975160-975164 on Build GRCh38
rs28561399 Chr.1: 975093-975093 on Build GRCh38
rs533931038 Chr.1: 974892-974892 on Build GRCh38
rs989297103 Chr.1: 975151-975151 on Build GRCh38
rs974767883 Chr.1: 974953-974953 on Build GRCh38
rs993218061 Chr.1: 974804-974804 on Build GRCh38
rs947822167 Chr.1: 975036-975036 on Build GRCh38
rs1030126326 Chr.1: 975039-975039 on Build GRCh38
rs941806216 Chr.1: 975168-975171 on Build GRCh38
rs935217399 Chr.1: 975204-975204 on Build GRCh38
rs375581213 Chr.1: 974943-974943 on Build GRCh38
rs567744734 Chr.1: 974910-974910 on Build GRCh38
rs1006451336 Chr.1: 975211-975211 on Build GRCh38
rs993867323 Chr.1: 975055-975055 on Build GRCh38
rs542458859 Chr.1: 975092-975092 on Build GRCh38
rs111830434 Chr.1: 974794-974794 on Build GRCh38
rs34754240 Chr.1: 974824-974825 on Build GRCh38
rs555778249 Chr.1: 975018-975018 on Build GRCh38
rs755534937 Chr.1: 975181-975181 on Build GRCh38
rs200122813 Chr.1: 975146-975149 on Build GRCh38
rs910010186 Chr.1: 975167-975167 on Build GRCh38
rs933249006 Chr.1: 975124-975125 on Build GRCh38
rs74424855 Chr.1: 975132-975132 on Build GRCh38
rs943333222 Chr.1: 974815-974815 on Build GRCh38
rs762227316 Chr.1: 974802-974802 on Build GRCh38
rs1006336757 Chr.1: 975059-975059 on Build GRCh38
rs370019776 Chr.1: 974797-974797 on Build GRCh38
rs548503468 Chr.1: 975130-975130 on Build GRCh38
rs982321753 Chr.1: 975095-975095 on Build GRCh38
rs772397939 Chr.1: 974762-974762 on Build GRCh38
rs775478962 Chr.1: 975115-975116 on Build GRCh38
rs548294591 Chr.1: 974860-974860 on Build GRCh38
rs900152061 Chr.1: 975118-975118 on Build GRCh38
rs759522236 Chr.1: 974908-974908 on Build GRCh38
rs571990964 Chr.1: 975165-975168 on Build GRCh38
rs916448450 Chr.1: 975159-975159 on Build GRCh38
rs1042202561 Chr.1: 974861-974861 on Build GRCh38
rs114241396 Chr.1: 974884-974884 on Build GRCh38
rs546044157 Chr.1: 975073-975073 on Build GRCh38
rs57882238 Chr.1: 975177-975177 on Build GRCh38
rs970425108 Chr.1: 974883-974883 on Build GRCh38
rs533480638 Chr.1: 975091-975091 on Build GRCh38
rs1017512949 Chr.1: 974841-974841 on Build GRCh38
rs930151436 Chr.1: 975106-975106 on Build GRCh38
rs983644360 Chr.1: 975108-975109 on Build GRCh38
rs918735837 Chr.1: 975009-975009 on Build GRCh38
rs531516036 Chr.1: 975116-975116 on Build GRCh38
rs936923445 Chr.1: 974777-974777 on Build GRCh38
rs981633463 Chr.1: 974789-974789 on Build GRCh38
rs141996759 Chr.1: 975148-975152 on Build GRCh38
rs759419736 Chr.1: 975056-975056 on Build GRCh38
rs1047869040 Chr.1: 974814-974814 on Build GRCh38
rs941718984 Chr.1: 975108-975116 on Build GRCh38
rs61358210 Chr.1: 975161-975161 on Build GRCh38
rs964746948 Chr.1: 975080-975080 on Build GRCh38
rs764611059 Chr.1: 974909-974909 on Build GRCh38
rs1026580936 Chr.1: 974920-974920 on Build GRCh38
rs1046548462 Chr.1: 975164-975164 on Build GRCh38
rs576901314 Chr.1: 974918-974918 on Build GRCh38
rs545009867 Chr.1: 975108-975111 on Build GRCh38
rs951652819 Chr.1: 975163-975165 on Build GRCh38
rs1023643771 Chr.1: 975150-975150 on Build GRCh38
rs972092826 Chr.1: 974786-974786 on Build GRCh38
rs28536514 Chr.1: 975029-975029 on Build GRCh38

View Full Product Details