Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1234227-1234764 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALT6
Entrez Gene ID
126792
Gene Name
beta-1,3-galactosyltransferase 6
Gene Aliases
EDSP2, SEMDJL1, beta3GalT6
UniGene
Hs.284284
Gene Chromosome Location
Chr.1: 1232249-1235041 on Build GRCh38
Transcripts
NM_080605.3

SNP Detail

SNP ID Chromosome Location
rs578059239 Chr.1: 1234536-1234536 on Build GRCh38
rs79425787 Chr.1: 1234257-1234257 on Build GRCh38
rs1027133948 Chr.1: 1234655-1234655 on Build GRCh38
rs1000550182 Chr.1: 1234432-1234432 on Build GRCh38
rs972842296 Chr.1: 1234533-1234533 on Build GRCh38
rs56852924 Chr.1: 1234593-1234593 on Build GRCh38
rs1049197885 Chr.1: 1234415-1234415 on Build GRCh38
rs910663784 Chr.1: 1234592-1234592 on Build GRCh38
rs563983349 Chr.1: 1234582-1234582 on Build GRCh38
rs972406433 Chr.1: 1234690-1234690 on Build GRCh38
rs375291472 Chr.1: 1234527-1234527 on Build GRCh38
rs1018488069 Chr.1: 1234680-1234680 on Build GRCh38
rs1000366062 Chr.1: 1234480-1234480 on Build GRCh38
rs889297589 Chr.1: 1234436-1234436 on Build GRCh38
rs79443011 Chr.1: 1234498-1234498 on Build GRCh38
rs748196490 Chr.1: 1234738-1234738 on Build GRCh38
rs1040631849 Chr.1: 1234454-1234454 on Build GRCh38
rs1040599313 Chr.1: 1234596-1234596 on Build GRCh38
rs147584042 Chr.1: 1234728-1234728 on Build GRCh38
rs114208940 Chr.1: 1234600-1234600 on Build GRCh38
rs1033785386 Chr.1: 1234445-1234445 on Build GRCh38
rs538800121 Chr.1: 1234375-1234375 on Build GRCh38
rs958510921 Chr.1: 1234524-1234524 on Build GRCh38
rs902865246 Chr.1: 1234619-1234619 on Build GRCh38
rs952124074 Chr.1: 1234734-1234734 on Build GRCh38
rs528705193 Chr.1: 1234632-1234632 on Build GRCh38
rs866119278 Chr.1: 1234546-1234546 on Build GRCh38
rs985711506 Chr.1: 1234563-1234563 on Build GRCh38
rs376392731 Chr.1: 1234360-1234360 on Build GRCh38
rs539084399 Chr.1: 1234303-1234303 on Build GRCh38
rs986155851 Chr.1: 1234678-1234678 on Build GRCh38
rs778066485 Chr.1: 1234382-1234382 on Build GRCh38
rs925785977 Chr.1: 1234281-1234281 on Build GRCh38
rs991309368 Chr.1: 1234528-1234528 on Build GRCh38
rs45591840 Chr.1: 1234348-1234348 on Build GRCh38
rs552005567 Chr.1: 1234692-1234692 on Build GRCh38
rs745879700 Chr.1: 1234503-1234503 on Build GRCh38
rs952942300 Chr.1: 1234541-1234543 on Build GRCh38
rs961528914 Chr.1: 1234505-1234505 on Build GRCh38
rs1055045877 Chr.1: 1234624-1234624 on Build GRCh38
rs930717774 Chr.1: 1234555-1234555 on Build GRCh38
rs557659114 Chr.1: 1234650-1234650 on Build GRCh38
rs145265329 Chr.1: 1234637-1234637 on Build GRCh38
rs991381411 Chr.1: 1234339-1234339 on Build GRCh38
rs576285504 Chr.1: 1234359-1234359 on Build GRCh38
rs997848645 Chr.1: 1234473-1234473 on Build GRCh38
rs749535599 Chr.1: 1234673-1234673 on Build GRCh38
rs564610226 Chr.1: 1234534-1234534 on Build GRCh38
rs919261526 Chr.1: 1234552-1234552 on Build GRCh38
rs570244002 Chr.1: 1234717-1234717 on Build GRCh38
rs116449540 Chr.1: 1234625-1234625 on Build GRCh38
rs1013214869 Chr.1: 1234629-1234629 on Build GRCh38
rs1020372408 Chr.1: 1234477-1234477 on Build GRCh38
rs909837843 Chr.1: 1234264-1234275 on Build GRCh38
rs958586164 Chr.1: 1234321-1234321 on Build GRCh38
rs572457553 Chr.1: 1234459-1234459 on Build GRCh38
rs896882896 Chr.1: 1234649-1234649 on Build GRCh38
rs1009829243 Chr.1: 1234601-1234601 on Build GRCh38
rs1033159909 Chr.1: 1234626-1234626 on Build GRCh38
rs903740928 Chr.1: 1234408-1234408 on Build GRCh38
rs971256147 Chr.1: 1234689-1234689 on Build GRCh38
rs757712673 Chr.1: 1234441-1234441 on Build GRCh38
rs971328161 Chr.1: 1234537-1234537 on Build GRCh38
rs911247460 Chr.1: 1234568-1234568 on Build GRCh38
rs747528218 Chr.1: 1234405-1234405 on Build GRCh38
rs1041353872 Chr.1: 1234610-1234610 on Build GRCh38
rs112067980 Chr.1: 1234478-1234478 on Build GRCh38
rs944050450 Chr.1: 1234575-1234576 on Build GRCh38
rs543867686 Chr.1: 1234567-1234567 on Build GRCh38
rs952868804 Chr.1: 1234656-1234656 on Build GRCh38

View Full Product Details