Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1312945-1313446 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5424020 c.1110C>A p.L370L Chr.1: 1313056-1313056 on Build GRCh38
6197350 c.1141C>T p.R381W Chr.1: 1313043-1313043 on Build GRCh38
3984146 c.1049T>C p.M350T Chr.1: 1313117-1313117 on Build GRCh38
5424021 c.1128C>A p.L376L Chr.1: 1313056-1313056 on Build GRCh38
6634873 c.1142G>A p.R381Q Chr.1: 1313042-1313042 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550340209 Chr.1: 1313270-1313270 on Build GRCh38
rs957465811 Chr.1: 1313328-1313328 on Build GRCh38
rs541205411 Chr.1: 1313241-1313241 on Build GRCh38
rs778939217 Chr.1: 1313131-1313131 on Build GRCh38
rs571545534 Chr.1: 1313345-1313345 on Build GRCh38
rs774338835 Chr.1: 1313005-1313005 on Build GRCh38
rs369623636 Chr.1: 1313326-1313326 on Build GRCh38
rs373477372 Chr.1: 1313376-1313376 on Build GRCh38
rs1041632800 Chr.1: 1313315-1313315 on Build GRCh38
rs74045438 Chr.1: 1313206-1313206 on Build GRCh38
rs879715121 Chr.1: 1313339-1313339 on Build GRCh38
rs780580202 Chr.1: 1313397-1313397 on Build GRCh38
rs997283135 Chr.1: 1313199-1313199 on Build GRCh38
rs752173663 Chr.1: 1313236-1313239 on Build GRCh38
rs746175790 Chr.1: 1312985-1312985 on Build GRCh38
rs527463409 Chr.1: 1313240-1313240 on Build GRCh38
rs570026046 Chr.1: 1313298-1313298 on Build GRCh38
rs992433298 Chr.1: 1313260-1313260 on Build GRCh38
rs768759266 Chr.1: 1313004-1313004 on Build GRCh38
rs941099747 Chr.1: 1313413-1313413 on Build GRCh38
rs751228573 Chr.1: 1313092-1313092 on Build GRCh38
rs373945578 Chr.1: 1313020-1313020 on Build GRCh38
rs775418738 Chr.1: 1313153-1313153 on Build GRCh38
rs899767309 Chr.1: 1313294-1313294 on Build GRCh38
rs140151309 Chr.1: 1313104-1313104 on Build GRCh38
rs758953398 Chr.1: 1313245-1313245 on Build GRCh38
rs563141128 Chr.1: 1313089-1313089 on Build GRCh38
rs768561022 Chr.1: 1313158-1313158 on Build GRCh38
rs533462638 Chr.1: 1313265-1313265 on Build GRCh38
rs771049158 Chr.1: 1313091-1313091 on Build GRCh38
rs780616170 Chr.1: 1313056-1313056 on Build GRCh38
rs1050321286 Chr.1: 1313197-1313197 on Build GRCh38
rs998906460 Chr.1: 1313207-1313207 on Build GRCh38
rs747119335 Chr.1: 1313149-1313149 on Build GRCh38
rs113577308 Chr.1: 1313327-1313327 on Build GRCh38
rs776566550 Chr.1: 1313019-1313019 on Build GRCh38
rs762829023 Chr.1: 1313090-1313090 on Build GRCh38
rs985559573 Chr.1: 1313419-1313419 on Build GRCh38
rs535033990 Chr.1: 1313336-1313336 on Build GRCh38
rs780741157 Chr.1: 1313116-1313116 on Build GRCh38
rs149153144 Chr.1: 1313354-1313354 on Build GRCh38
rs562299323 Chr.1: 1312964-1312964 on Build GRCh38
rs996747526 Chr.1: 1313302-1313302 on Build GRCh38
rs142980334 Chr.1: 1313087-1313087 on Build GRCh38
rs1008324045 Chr.1: 1313025-1313025 on Build GRCh38
rs1024908737 Chr.1: 1313378-1313378 on Build GRCh38
rs762585928 Chr.1: 1313155-1313155 on Build GRCh38
rs192157989 Chr.1: 1313341-1313341 on Build GRCh38
rs756534506 Chr.1: 1313050-1313050 on Build GRCh38
rs1031845888 Chr.1: 1313217-1313217 on Build GRCh38
rs762958897 Chr.1: 1313242-1313242 on Build GRCh38
rs898785221 Chr.1: 1312991-1312991 on Build GRCh38
rs749580829 Chr.1: 1313000-1313000 on Build GRCh38
rs961082890 Chr.1: 1313383-1313383 on Build GRCh38
rs752320898 Chr.1: 1313021-1313021 on Build GRCh38
rs369337063 Chr.1: 1312968-1312968 on Build GRCh38
rs774658242 Chr.1: 1313065-1313065 on Build GRCh38
rs944207618 Chr.1: 1313313-1313313 on Build GRCh38
rs545539612 Chr.1: 1313038-1313038 on Build GRCh38
rs1010341131 Chr.1: 1313330-1313330 on Build GRCh38
rs369448288 Chr.1: 1313012-1313012 on Build GRCh38
rs78535135 Chr.1: 1312973-1312973 on Build GRCh38
rs750944542 Chr.1: 1313113-1313113 on Build GRCh38
rs750038739 Chr.1: 1313398-1313398 on Build GRCh38
rs376193505 Chr.1: 1313134-1313134 on Build GRCh38
rs543500205 Chr.1: 1312967-1312967 on Build GRCh38
rs768724959 Chr.1: 1313063-1313063 on Build GRCh38
rs898732953 Chr.1: 1313188-1313188 on Build GRCh38
rs750983730 Chr.1: 1313029-1313029 on Build GRCh38
rs373229259 Chr.1: 1313253-1313253 on Build GRCh38
rs1029646516 Chr.1: 1313203-1313203 on Build GRCh38
rs376879612 Chr.1: 1313026-1313026 on Build GRCh38
rs769617133 Chr.1: 1313332-1313332 on Build GRCh38
rs758022638 Chr.1: 1313023-1313023 on Build GRCh38
rs376604506 Chr.1: 1312986-1312986 on Build GRCh38
rs890069378 Chr.1: 1313303-1313303 on Build GRCh38
rs749570256 Chr.1: 1313127-1313127 on Build GRCh38
rs550834556 Chr.1: 1313339-1313340 on Build GRCh38
rs765080194 Chr.1: 1313187-1313187 on Build GRCh38
rs773862725 Chr.1: 1313169-1313169 on Build GRCh38
rs756803839 Chr.1: 1313036-1313036 on Build GRCh38
rs941225091 Chr.1: 1313289-1313289 on Build GRCh38
rs529291826 Chr.1: 1313319-1313319 on Build GRCh38
rs764982365 Chr.1: 1313088-1313088 on Build GRCh38
rs772115268 Chr.1: 1313135-1313135 on Build GRCh38
rs566167631 Chr.1: 1313296-1313300 on Build GRCh38
rs776859024 Chr.1: 1313146-1313146 on Build GRCh38
rs761987136 Chr.1: 1313009-1313009 on Build GRCh38
rs529799687 Chr.1: 1313181-1313181 on Build GRCh38
rs755386598 Chr.1: 1313128-1313128 on Build GRCh38
rs756848979 Chr.1: 1313369-1313369 on Build GRCh38
rs769669296 Chr.1: 1313150-1313150 on Build GRCh38
rs9725147 Chr.1: 1313219-1313219 on Build GRCh38
rs772257638 Chr.1: 1313077-1313077 on Build GRCh38
rs764131825 Chr.1: 1313275-1313275 on Build GRCh38
rs1051104964 Chr.1: 1313314-1313314 on Build GRCh38
rs990328110 Chr.1: 1313234-1313234 on Build GRCh38
rs939797606 Chr.1: 1313287-1313287 on Build GRCh38
rs917696621 Chr.1: 1313132-1313132 on Build GRCh38
rs774434651 Chr.1: 1313016-1313022 on Build GRCh38
rs1038191812 Chr.1: 1313290-1313290 on Build GRCh38
rs746446373 Chr.1: 1313043-1313043 on Build GRCh38
rs776534078 Chr.1: 1313085-1313085 on Build GRCh38
rs766181999 Chr.1: 1313013-1313013 on Build GRCh38
rs756719002 Chr.1: 1313103-1313103 on Build GRCh38
rs1025247113 Chr.1: 1313381-1313382 on Build GRCh38
rs548248819 Chr.1: 1313198-1313198 on Build GRCh38
rs1012183598 Chr.1: 1313331-1313331 on Build GRCh38
rs549376222 Chr.1: 1313321-1313321 on Build GRCh38
rs144721249 Chr.1: 1313414-1313414 on Build GRCh38
rs748121981 Chr.1: 1313071-1313071 on Build GRCh38
rs899740247 Chr.1: 1313193-1313193 on Build GRCh38
rs761576095 Chr.1: 1313171-1313171 on Build GRCh38

View Full Product Details