Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1313420-1313914 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5434795 c.994C>T p.Q332* Chr.1: 1313556-1313556 on Build GRCh38
4021412 c.1012C>T p.R338W Chr.1: 1313538-1313538 on Build GRCh38
4021411 c.1030G>A p.E344K Chr.1: 1313520-1313520 on Build GRCh38
4021414 c.778A>G p.N260D Chr.1: 1313911-1313911 on Build GRCh38
6634878 c.1021C>T p.Q341* Chr.1: 1313547-1313547 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758575124 Chr.1: 1313503-1313503 on Build GRCh38
rs757512247 Chr.1: 1313708-1313708 on Build GRCh38
rs551665100 Chr.1: 1313872-1313872 on Build GRCh38
rs771001457 Chr.1: 1313515-1313515 on Build GRCh38
rs753669890 Chr.1: 1313693-1313693 on Build GRCh38
rs756218307 Chr.1: 1313756-1313756 on Build GRCh38
rs905990187 Chr.1: 1313588-1313588 on Build GRCh38
rs556125116 Chr.1: 1313752-1313753 on Build GRCh38
rs148768825 Chr.1: 1313533-1313533 on Build GRCh38
rs1003024903 Chr.1: 1313822-1313822 on Build GRCh38
rs962786059 Chr.1: 1313688-1313688 on Build GRCh38
rs773162180 Chr.1: 1313625-1313626 on Build GRCh38
rs950954196 Chr.1: 1313666-1313666 on Build GRCh38
rs747488263 Chr.1: 1313616-1313616 on Build GRCh38
rs747897328 Chr.1: 1313625-1313625 on Build GRCh38
rs749306909 Chr.1: 1313538-1313538 on Build GRCh38
rs559533918 Chr.1: 1313801-1313801 on Build GRCh38
rs774853966 Chr.1: 1313811-1313811 on Build GRCh38
rs779391005 Chr.1: 1313873-1313873 on Build GRCh38
rs373365375 Chr.1: 1313466-1313466 on Build GRCh38
rs763499397 Chr.1: 1313734-1313734 on Build GRCh38
rs778404375 Chr.1: 1313605-1313605 on Build GRCh38
rs577921220 Chr.1: 1313689-1313689 on Build GRCh38
rs12142199 Chr.1: 1313807-1313807 on Build GRCh38
rs1051047630 Chr.1: 1313464-1313464 on Build GRCh38
rs192196814 Chr.1: 1313690-1313690 on Build GRCh38
rs760557440 Chr.1: 1313637-1313637 on Build GRCh38
rs780898796 Chr.1: 1313842-1313842 on Build GRCh38
rs374585056 Chr.1: 1313882-1313882 on Build GRCh38
rs775962016 Chr.1: 1313733-1313733 on Build GRCh38
rs146520212 Chr.1: 1313735-1313735 on Build GRCh38
rs199574867 Chr.1: 1313714-1313714 on Build GRCh38
rs527406831 Chr.1: 1313777-1313777 on Build GRCh38
rs373444911 Chr.1: 1313883-1313883 on Build GRCh38
rs757313978 Chr.1: 1313607-1313607 on Build GRCh38
rs758367428 Chr.1: 1313768-1313768 on Build GRCh38
rs774657890 Chr.1: 1313741-1313741 on Build GRCh38
rs199575761 Chr.1: 1313765-1313765 on Build GRCh38
rs570365291 Chr.1: 1313711-1313711 on Build GRCh38
rs758257266 Chr.1: 1313719-1313719 on Build GRCh38
rs201455031 Chr.1: 1313723-1313723 on Build GRCh38
rs766735536 Chr.1: 1313827-1313827 on Build GRCh38
rs752230074 Chr.1: 1313813-1313814 on Build GRCh38
rs779703791 Chr.1: 1313853-1313853 on Build GRCh38
rs142134905 Chr.1: 1313578-1313578 on Build GRCh38
rs780891309 Chr.1: 1313782-1313782 on Build GRCh38
rs764053756 Chr.1: 1313836-1313836 on Build GRCh38
rs764499012 Chr.1: 1313703-1313703 on Build GRCh38
rs751844160 Chr.1: 1313603-1313603 on Build GRCh38
rs762013216 Chr.1: 1313492-1313502 on Build GRCh38
rs753916493 Chr.1: 1313829-1313829 on Build GRCh38
rs766281620 Chr.1: 1313687-1313688 on Build GRCh38
rs766431443 Chr.1: 1313761-1313761 on Build GRCh38
rs187619625 Chr.1: 1313675-1313675 on Build GRCh38
rs1023561012 Chr.1: 1313581-1313581 on Build GRCh38
rs367770919 Chr.1: 1313718-1313718 on Build GRCh38
rs199692417 Chr.1: 1313521-1313521 on Build GRCh38
rs965415964 Chr.1: 1313892-1313892 on Build GRCh38
rs201564312 Chr.1: 1313858-1313858 on Build GRCh38
rs952468027 Chr.1: 1313866-1313866 on Build GRCh38
rs371231719 Chr.1: 1313721-1313721 on Build GRCh38
rs897108688 Chr.1: 1313818-1313818 on Build GRCh38
rs891979169 Chr.1: 1313468-1313468 on Build GRCh38
rs770187372 Chr.1: 1313749-1313749 on Build GRCh38
rs745565232 Chr.1: 1313786-1313786 on Build GRCh38
rs962447761 Chr.1: 1313869-1313869 on Build GRCh38
rs769254461 Chr.1: 1313726-1313726 on Build GRCh38
rs763298002 Chr.1: 1313697-1313697 on Build GRCh38
rs768315997 Chr.1: 1313545-1313545 on Build GRCh38
rs555104938 Chr.1: 1313589-1313589 on Build GRCh38
rs369330200 Chr.1: 1313750-1313750 on Build GRCh38
rs764995765 Chr.1: 1313753-1313753 on Build GRCh38
rs772997141 Chr.1: 1313633-1313633 on Build GRCh38
rs776162938 Chr.1: 1313643-1313643 on Build GRCh38
rs200557566 Chr.1: 1313597-1313597 on Build GRCh38
rs150166328 Chr.1: 1313769-1313769 on Build GRCh38
rs773017208 Chr.1: 1313562-1313562 on Build GRCh38
rs755281612 Chr.1: 1313492-1313492 on Build GRCh38
rs572285730 Chr.1: 1313602-1313602 on Build GRCh38
rs910894352 Chr.1: 1313656-1313656 on Build GRCh38
rs747033468 Chr.1: 1313508-1313508 on Build GRCh38
rs1015679150 Chr.1: 1313868-1313868 on Build GRCh38
rs769104260 Chr.1: 1313808-1313808 on Build GRCh38
rs757059784 Chr.1: 1313839-1313839 on Build GRCh38
rs751760165 Chr.1: 1313707-1313707 on Build GRCh38
rs375687548 Chr.1: 1313843-1313843 on Build GRCh38
rs759699613 Chr.1: 1313832-1313832 on Build GRCh38
rs778847837 Chr.1: 1313716-1313716 on Build GRCh38
rs762174238 Chr.1: 1313814-1313814 on Build GRCh38
rs777992120 Chr.1: 1313504-1313504 on Build GRCh38
rs771977193 Chr.1: 1313616-1313616 on Build GRCh38
rs761120074 Chr.1: 1313889-1313889 on Build GRCh38
rs371596695 Chr.1: 1313781-1313781 on Build GRCh38
rs769325143 Chr.1: 1313798-1313798 on Build GRCh38
rs777747786 Chr.1: 1313720-1313720 on Build GRCh38
rs750468389 Chr.1: 1313712-1313712 on Build GRCh38
rs200727675 Chr.1: 1313753-1313753 on Build GRCh38
rs367695084 Chr.1: 1313490-1313490 on Build GRCh38
rs575917311 Chr.1: 1313527-1313527 on Build GRCh38
rs377592423 Chr.1: 1313563-1313563 on Build GRCh38
rs886708953 Chr.1: 1313828-1313828 on Build GRCh38
rs748893742 Chr.1: 1313630-1313630 on Build GRCh38
rs373852621 Chr.1: 1313482-1313482 on Build GRCh38
rs775865212 Chr.1: 1313693-1313693 on Build GRCh38
rs184751855 Chr.1: 1313640-1313640 on Build GRCh38
rs372312823 Chr.1: 1313692-1313692 on Build GRCh38
rs774243500 Chr.1: 1313547-1313547 on Build GRCh38
rs535451124 Chr.1: 1313551-1313551 on Build GRCh38
rs371636552 Chr.1: 1313501-1313501 on Build GRCh38
rs760858687 Chr.1: 1313819-1313819 on Build GRCh38
rs371108423 Chr.1: 1313463-1313463 on Build GRCh38
rs904640871 Chr.1: 1313500-1313500 on Build GRCh38
rs763923444 Chr.1: 1313884-1313884 on Build GRCh38
rs762108167 Chr.1: 1313745-1313745 on Build GRCh38
rs778741231 Chr.1: 1313621-1313621 on Build GRCh38
rs140248853 Chr.1: 1313579-1313579 on Build GRCh38
rs746704150 Chr.1: 1313637-1313637 on Build GRCh38
rs755682414 Chr.1: 1313848-1313848 on Build GRCh38
rs267597965 Chr.1: 1313594-1313594 on Build GRCh38
rs370598517 Chr.1: 1313572-1313572 on Build GRCh38
rs375843422 Chr.1: 1313627-1313627 on Build GRCh38
rs961196984 Chr.1: 1313542-1313542 on Build GRCh38
rs533508458 Chr.1: 1313646-1313646 on Build GRCh38
rs982435289 Chr.1: 1313673-1313673 on Build GRCh38
rs771806036 Chr.1: 1313554-1313554 on Build GRCh38
rs772387554 Chr.1: 1313816-1313816 on Build GRCh38
rs755785536 Chr.1: 1313467-1313467 on Build GRCh38

View Full Product Details