Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1314725-1315219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3L
Entrez Gene ID
54973
Gene Name
cleavage and polyadenylation specific factor 3-like
Gene Aliases
CPSF73L, INT11, INTS11, RC-68, RC68
UniGene
Hs.6449
Gene Chromosome Location
Chr.1: 1311585-1324687 on Build GRCh38
Transcripts
NM_001256456.1 NM_001256460.1 NM_001256462.1 NM_001256463.1 NM_017871.5 XM_011541647.1 XM_011541648.1 XM_011541650.1 XM_017001557.1 XM_017001558.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6042146 c.582-8T>C p.? Chr.1: 1314970-1314970 on Build GRCh38
1333357 c.671A>G p.K224R Chr.1: 1314855-1314855 on Build GRCh38
6042144 c.702C>T p.T234T Chr.1: 1314842-1314842 on Build GRCh38
6057753 c.687G>C p.K229N Chr.1: 1314857-1314857 on Build GRCh38
5550995 c.603C>T p.C201C Chr.1: 1314941-1314941 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028180429 Chr.1: 1314761-1314761 on Build GRCh38
rs545146495 Chr.1: 1314821-1314821 on Build GRCh38
rs181287914 Chr.1: 1315170-1315170 on Build GRCh38
rs368691656 Chr.1: 1314972-1314972 on Build GRCh38
rs755190827 Chr.1: 1314870-1314870 on Build GRCh38
rs780135771 Chr.1: 1314779-1314779 on Build GRCh38
rs776550612 Chr.1: 1314801-1314801 on Build GRCh38
rs897753289 Chr.1: 1315161-1315161 on Build GRCh38
rs575206993 Chr.1: 1315132-1315132 on Build GRCh38
rs774151732 Chr.1: 1314903-1314903 on Build GRCh38
rs189734561 Chr.1: 1315084-1315084 on Build GRCh38
rs113449676 Chr.1: 1315173-1315173 on Build GRCh38
rs781780682 Chr.1: 1315126-1315126 on Build GRCh38
rs575554390 Chr.1: 1314834-1314834 on Build GRCh38
rs546388361 Chr.1: 1315019-1315019 on Build GRCh38
rs376583652 Chr.1: 1314929-1314929 on Build GRCh38
rs988314529 Chr.1: 1315051-1315051 on Build GRCh38
rs2296473 Chr.1: 1314848-1314848 on Build GRCh38
rs949912946 Chr.1: 1315115-1315115 on Build GRCh38
rs774981700 Chr.1: 1314811-1314811 on Build GRCh38
rs779198517 Chr.1: 1314842-1314842 on Build GRCh38
rs1019069593 Chr.1: 1315178-1315178 on Build GRCh38
rs752971027 Chr.1: 1314932-1314932 on Build GRCh38
rs953467870 Chr.1: 1314762-1314762 on Build GRCh38
rs914151094 Chr.1: 1314819-1314819 on Build GRCh38
rs560713233 Chr.1: 1314902-1314902 on Build GRCh38
rs925712683 Chr.1: 1314852-1314852 on Build GRCh38
rs779132215 Chr.1: 1314872-1314872 on Build GRCh38
rs142031612 Chr.1: 1314829-1314829 on Build GRCh38
rs777068339 Chr.1: 1315000-1315000 on Build GRCh38
rs377386006 Chr.1: 1314790-1314790 on Build GRCh38
rs200181216 Chr.1: 1314971-1314971 on Build GRCh38
rs767194750 Chr.1: 1314904-1314904 on Build GRCh38
rs550814894 Chr.1: 1315008-1315008 on Build GRCh38
rs768810768 Chr.1: 1315192-1315192 on Build GRCh38
rs745928286 Chr.1: 1315150-1315150 on Build GRCh38
rs150689780 Chr.1: 1314843-1314843 on Build GRCh38
rs766829641 Chr.1: 1314911-1314911 on Build GRCh38
rs75629572 Chr.1: 1315027-1315027 on Build GRCh38
rs758447433 Chr.1: 1314933-1314933 on Build GRCh38
rs925609817 Chr.1: 1315065-1315065 on Build GRCh38
rs116331779 Chr.1: 1315050-1315050 on Build GRCh38
rs553165409 Chr.1: 1314786-1314786 on Build GRCh38
rs558521500 Chr.1: 1314845-1314845 on Build GRCh38
rs202164977 Chr.1: 1314983-1314983 on Build GRCh38
rs777868865 Chr.1: 1314936-1314936 on Build GRCh38
rs780285310 Chr.1: 1315169-1315169 on Build GRCh38
rs754622200 Chr.1: 1314874-1314874 on Build GRCh38
rs978849231 Chr.1: 1315064-1315064 on Build GRCh38
rs759319030 Chr.1: 1314802-1314802 on Build GRCh38
rs754186313 Chr.1: 1314914-1314914 on Build GRCh38
rs748275476 Chr.1: 1314920-1314921 on Build GRCh38
rs370089867 Chr.1: 1314877-1314877 on Build GRCh38
rs907443419 Chr.1: 1314862-1314862 on Build GRCh38
rs139153962 Chr.1: 1314939-1314939 on Build GRCh38
rs948496002 Chr.1: 1315005-1315005 on Build GRCh38
rs779868844 Chr.1: 1314940-1314940 on Build GRCh38
rs900560273 Chr.1: 1315120-1315120 on Build GRCh38
rs757749226 Chr.1: 1315093-1315093 on Build GRCh38
rs537869274 Chr.1: 1315162-1315162 on Build GRCh38
rs536071578 Chr.1: 1314776-1314776 on Build GRCh38
rs199777303 Chr.1: 1315195-1315204 on Build GRCh38
rs772203175 Chr.1: 1314876-1314876 on Build GRCh38
rs775314558 Chr.1: 1314888-1314888 on Build GRCh38
rs772812197 Chr.1: 1314993-1314993 on Build GRCh38
rs768598338 Chr.1: 1314788-1314788 on Build GRCh38
rs756203350 Chr.1: 1315163-1315163 on Build GRCh38
rs1019125301 Chr.1: 1315047-1315047 on Build GRCh38
rs748154379 Chr.1: 1314873-1314873 on Build GRCh38
rs768639694 Chr.1: 1314965-1314965 on Build GRCh38
rs999561953 Chr.1: 1315172-1315172 on Build GRCh38
rs201072145 Chr.1: 1314808-1314808 on Build GRCh38
rs761287688 Chr.1: 1314908-1314908 on Build GRCh38
rs544844033 Chr.1: 1314855-1314855 on Build GRCh38
rs200149331 Chr.1: 1314883-1314883 on Build GRCh38
rs560061998 Chr.1: 1315009-1315009 on Build GRCh38
rs34313161 Chr.1: 1314890-1314890 on Build GRCh38
rs988351527 Chr.1: 1314806-1314806 on Build GRCh38
rs753979888 Chr.1: 1314856-1314856 on Build GRCh38
rs914164756 Chr.1: 1315057-1315057 on Build GRCh38
rs762705843 Chr.1: 1314816-1314816 on Build GRCh38
rs929519295 Chr.1: 1315049-1315049 on Build GRCh38
rs749044981 Chr.1: 1314956-1314956 on Build GRCh38
rs765890808 Chr.1: 1314774-1314774 on Build GRCh38
rs372914859 Chr.1: 1314995-1314995 on Build GRCh38
rs916986813 Chr.1: 1315105-1315105 on Build GRCh38
rs751599310 Chr.1: 1315010-1315010 on Build GRCh38
rs764467184 Chr.1: 1315036-1315036 on Build GRCh38
rs763449895 Chr.1: 1314831-1314837 on Build GRCh38
rs759928482 Chr.1: 1315003-1315003 on Build GRCh38
rs370809438 Chr.1: 1314800-1314800 on Build GRCh38
rs997507615 Chr.1: 1314912-1314912 on Build GRCh38
rs146701215 Chr.1: 1314875-1314875 on Build GRCh38
rs374467956 Chr.1: 1314922-1314922 on Build GRCh38
rs12095333 Chr.1: 1314938-1314938 on Build GRCh38
rs145964770 Chr.1: 1314905-1314905 on Build GRCh38
rs756506402 Chr.1: 1314777-1314777 on Build GRCh38
rs536573666 Chr.1: 1315017-1315017 on Build GRCh38
rs745864321 Chr.1: 1314881-1314881 on Build GRCh38
rs141502981 Chr.1: 1315058-1315058 on Build GRCh38
rs368395926 Chr.1: 1314842-1314842 on Build GRCh38
rs771679573 Chr.1: 1314986-1314986 on Build GRCh38
rs1039634189 Chr.1: 1315087-1315087 on Build GRCh38
rs772214806 Chr.1: 1314972-1314972 on Build GRCh38
rs901910717 Chr.1: 1315189-1315189 on Build GRCh38

View Full Product Details