Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1392200-1392696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCNL2
Entrez Gene ID
81669
Gene Name
cyclin L2
Gene Aliases
ANIA-6B, CCNM, CCNS, HCLA-ISO, HLA-ISO, PCEE, SB138
UniGene
Hs.515704
Gene Chromosome Location
Chr.1: 1385711-1399338 on Build GRCh38
Transcripts
NM_001039577.3 NM_001320153.1 NM_001320155.1 NM_030937.4 NR_135154.1 XM_011542216.2 XM_011542219.2 XM_011542221.2 XM_017002420.1 XM_017002421.1 XM_017002422.1 XM_017002423.1

SNP Detail

SNP ID Chromosome Location
rs998471902 Chr.1: 1392438-1392438 on Build GRCh38
rs191939238 Chr.1: 1392247-1392247 on Build GRCh38
rs941498751 Chr.1: 1392676-1392676 on Build GRCh38
rs989328270 Chr.1: 1392589-1392589 on Build GRCh38
rs187897476 Chr.1: 1392362-1392362 on Build GRCh38
rs915015079 Chr.1: 1392421-1392421 on Build GRCh38
rs754137598 Chr.1: 1392675-1392675 on Build GRCh38
rs971711175 Chr.1: 1392514-1392514 on Build GRCh38
rs137906875 Chr.1: 1392374-1392374 on Build GRCh38
rs1025849746 Chr.1: 1392231-1392231 on Build GRCh38
rs74766556 Chr.1: 1392477-1392477 on Build GRCh38
rs963990104 Chr.1: 1392313-1392313 on Build GRCh38
rs184967896 Chr.1: 1392357-1392357 on Build GRCh38
rs549203292 Chr.1: 1392419-1392419 on Build GRCh38
rs918246648 Chr.1: 1392349-1392349 on Build GRCh38
rs923753209 Chr.1: 1392557-1392557 on Build GRCh38
rs563960289 Chr.1: 1392343-1392343 on Build GRCh38
rs983728871 Chr.1: 1392567-1392567 on Build GRCh38
rs939579103 Chr.1: 1392656-1392656 on Build GRCh38
rs929777963 Chr.1: 1392358-1392358 on Build GRCh38
rs537476101 Chr.1: 1392610-1392610 on Build GRCh38
rs950312373 Chr.1: 1392319-1392319 on Build GRCh38
rs964103625 Chr.1: 1392489-1392490 on Build GRCh38
rs914953314 Chr.1: 1392590-1392590 on Build GRCh38
rs775685810 Chr.1: 1392451-1392451 on Build GRCh38
rs1029702646 Chr.1: 1392511-1392511 on Build GRCh38
rs1024438335 Chr.1: 1392508-1392508 on Build GRCh38
rs947822827 Chr.1: 1392618-1392620 on Build GRCh38
rs757966015 Chr.1: 1392364-1392364 on Build GRCh38
rs954653076 Chr.1: 1392335-1392335 on Build GRCh38
rs566175240 Chr.1: 1392440-1392440 on Build GRCh38
rs369862332 Chr.1: 1392641-1392641 on Build GRCh38
rs534848929 Chr.1: 1392443-1392443 on Build GRCh38
rs1004078265 Chr.1: 1392230-1392230 on Build GRCh38
rs527438759 Chr.1: 1392277-1392277 on Build GRCh38
rs927679802 Chr.1: 1392639-1392639 on Build GRCh38
rs958747526 Chr.1: 1392469-1392469 on Build GRCh38
rs951103306 Chr.1: 1392564-1392564 on Build GRCh38
rs921811072 Chr.1: 1392326-1392326 on Build GRCh38
rs1045116117 Chr.1: 1392631-1392631 on Build GRCh38
rs1044736887 Chr.1: 1392439-1392439 on Build GRCh38
rs917515946 Chr.1: 1392316-1392316 on Build GRCh38
rs558118999 Chr.1: 1392449-1392449 on Build GRCh38
rs377404751 Chr.1: 1392237-1392237 on Build GRCh38
rs1016870707 Chr.1: 1392263-1392263 on Build GRCh38
rs541011184 Chr.1: 1392322-1392322 on Build GRCh38
rs1047286302 Chr.1: 1392659-1392659 on Build GRCh38
rs374736824 Chr.1: 1392378-1392378 on Build GRCh38
rs1000308546 Chr.1: 1392445-1392445 on Build GRCh38
rs918290063 Chr.1: 1392548-1392548 on Build GRCh38
rs972718812 Chr.1: 1392522-1392522 on Build GRCh38
rs751284253 Chr.1: 1392429-1392429 on Build GRCh38
rs1053558452 Chr.1: 1392415-1392415 on Build GRCh38
rs548354234 Chr.1: 1392530-1392530 on Build GRCh38
rs991629725 Chr.1: 1392309-1392309 on Build GRCh38
rs942556346 Chr.1: 1392412-1392412 on Build GRCh38
rs752163415 Chr.1: 1392325-1392325 on Build GRCh38
rs192786723 Chr.1: 1392490-1392490 on Build GRCh38

View Full Product Details