Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1495555-1496059 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD3B
Entrez Gene ID
83858
Gene Name
ATPase family, AAA domain containing 3B
Gene Aliases
AAA-TOB3, TOB3
UniGene
Hs.729021
Gene Chromosome Location
Chr.1: 1471732-1509466 on Build GRCh38
Transcripts
NM_001317238.1 NM_031921.5 XM_005244806.3 XM_011542243.2 XM_011542244.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5843390 c.1750C>T p.P584S Chr.1: 1495620-1495620 on Build GRCh38
1196902 c.1871G>T p.C624F Chr.1: 1495741-1495741 on Build GRCh38
4516399 c.1887_1888CC>TT p.P630S Chr.1: 1495757-1495758 on Build GRCh38
3747531 c.1735C>T p.R579C Chr.1: 1495605-1495605 on Build GRCh38
895042 c.1701G>T p.Q567H Chr.1: 1495571-1495571 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966453170 Chr.1: 1495814-1495814 on Build GRCh38
rs146723974 Chr.1: 1495796-1495796 on Build GRCh38
rs745957756 Chr.1: 1495722-1495722 on Build GRCh38
rs764412648 Chr.1: 1495610-1495610 on Build GRCh38
rs778410099 Chr.1: 1495752-1495752 on Build GRCh38
rs531492333 Chr.1: 1495620-1495620 on Build GRCh38
rs765968938 Chr.1: 1495576-1495576 on Build GRCh38
rs146505120 Chr.1: 1495793-1495793 on Build GRCh38
rs755022080 Chr.1: 1495684-1495684 on Build GRCh38
rs139902189 Chr.1: 1495605-1495605 on Build GRCh38
rs148302536 Chr.1: 1495798-1495798 on Build GRCh38
rs901593144 Chr.1: 1495602-1495602 on Build GRCh38
rs770756718 Chr.1: 1495648-1495648 on Build GRCh38
rs930537575 Chr.1: 1495580-1495580 on Build GRCh38
rs745740576 Chr.1: 1495734-1495734 on Build GRCh38
rs779885885 Chr.1: 1495739-1495739 on Build GRCh38
rs138274487 Chr.1: 1495670-1495670 on Build GRCh38
rs901207010 Chr.1: 1496029-1496029 on Build GRCh38
rs910447634 Chr.1: 1495666-1495666 on Build GRCh38
rs753648141 Chr.1: 1495827-1495827 on Build GRCh38
rs768740654 Chr.1: 1495640-1495640 on Build GRCh38
rs200599738 Chr.1: 1495694-1495694 on Build GRCh38
rs9792997 Chr.1: 1495680-1495680 on Build GRCh38
rs541984530 Chr.1: 1495819-1495819 on Build GRCh38
rs975898540 Chr.1: 1495803-1495803 on Build GRCh38
rs577199359 Chr.1: 1495607-1495607 on Build GRCh38
rs888549583 Chr.1: 1495992-1495992 on Build GRCh38
rs775785353 Chr.1: 1495861-1495861 on Build GRCh38
rs200835805 Chr.1: 1495807-1495807 on Build GRCh38
rs189192055 Chr.1: 1495588-1495588 on Build GRCh38
rs763836096 Chr.1: 1495778-1495779 on Build GRCh38
rs768121440 Chr.1: 1495706-1495706 on Build GRCh38
rs144198445 Chr.1: 1495729-1495729 on Build GRCh38
rs757570983 Chr.1: 1495628-1495628 on Build GRCh38
rs772182612 Chr.1: 1495608-1495608 on Build GRCh38
rs140821046 Chr.1: 1495989-1495989 on Build GRCh38
rs769624585 Chr.1: 1495742-1495742 on Build GRCh38
rs541175302 Chr.1: 1495846-1495846 on Build GRCh38
rs990507746 Chr.1: 1495924-1495924 on Build GRCh38
rs149788676 Chr.1: 1495627-1495627 on Build GRCh38
rs775129691 Chr.1: 1495696-1495696 on Build GRCh38
rs1018659709 Chr.1: 1495897-1495897 on Build GRCh38
rs746020409 Chr.1: 1495602-1495602 on Build GRCh38
rs373279247 Chr.1: 1495756-1495756 on Build GRCh38
rs771165746 Chr.1: 1495678-1495678 on Build GRCh38
rs145585259 Chr.1: 1495919-1495919 on Build GRCh38
rs754236762 Chr.1: 1495724-1495724 on Build GRCh38
rs904588 Chr.1: 1495971-1495971 on Build GRCh38
rs761123135 Chr.1: 1495717-1495717 on Build GRCh38
rs752241293 Chr.1: 1495622-1495622 on Build GRCh38
rs771614139 Chr.1: 1495801-1495801 on Build GRCh38
rs777093904 Chr.1: 1495778-1495778 on Build GRCh38
rs570553525 Chr.1: 1495673-1495673 on Build GRCh38
rs764296610 Chr.1: 1495818-1495818 on Build GRCh38
rs79205801 Chr.1: 1495695-1495695 on Build GRCh38
rs776553293 Chr.1: 1495655-1495655 on Build GRCh38
rs562772300 Chr.1: 1495619-1495619 on Build GRCh38
rs566186657 Chr.1: 1496005-1496005 on Build GRCh38
rs781141101 Chr.1: 1495731-1495731 on Build GRCh38
rs781725723 Chr.1: 1495855-1495855 on Build GRCh38
rs775076916 Chr.1: 1495667-1495667 on Build GRCh38
rs201844668 Chr.1: 1495779-1495779 on Build GRCh38
rs756785910 Chr.1: 1495849-1495849 on Build GRCh38
rs779373455 Chr.1: 1495751-1495751 on Build GRCh38
rs199599542 Chr.1: 1495725-1495725 on Build GRCh38
rs528543988 Chr.1: 1495972-1495974 on Build GRCh38
rs767182641 Chr.1: 1495840-1495841 on Build GRCh38
rs778175812 Chr.1: 1495624-1495624 on Build GRCh38
rs753489353 Chr.1: 1495577-1495577 on Build GRCh38
rs770321855 Chr.1: 1495857-1495857 on Build GRCh38
rs768185157 Chr.1: 1495697-1495697 on Build GRCh38
rs562182330 Chr.1: 1495646-1495646 on Build GRCh38
rs758359599 Chr.1: 1495730-1495730 on Build GRCh38
rs751625522 Chr.1: 1495786-1495786 on Build GRCh38
rs762430432 Chr.1: 1495625-1495625 on Build GRCh38
rs761861825 Chr.1: 1495823-1495823 on Build GRCh38
rs780393780 Chr.1: 1495635-1495635 on Build GRCh38
rs9792879 Chr.1: 1495785-1495785 on Build GRCh38
rs372995030 Chr.1: 1495669-1495669 on Build GRCh38
rs140300218 Chr.1: 1495604-1495604 on Build GRCh38
rs966543762 Chr.1: 1495917-1495917 on Build GRCh38
rs752528694 Chr.1: 1495833-1495833 on Build GRCh38
rs767907235 Chr.1: 1495671-1495671 on Build GRCh38
rs373709131 Chr.1: 1495614-1495614 on Build GRCh38
rs147286181 Chr.1: 1495591-1495591 on Build GRCh38
rs570265195 Chr.1: 1495698-1495698 on Build GRCh38
rs536016095 Chr.1: 1495713-1495713 on Build GRCh38
rs756775187 Chr.1: 1495592-1495592 on Build GRCh38
rs3842878 Chr.1: 1495905-1495905 on Build GRCh38
rs777575157 Chr.1: 1495641-1495641 on Build GRCh38
rs994324988 Chr.1: 1495876-1495876 on Build GRCh38
rs759975415 Chr.1: 1495806-1495806 on Build GRCh38
rs533689685 Chr.1: 1495748-1495748 on Build GRCh38
rs756473127 Chr.1: 1495632-1495632 on Build GRCh38
rs957987998 Chr.1: 1495922-1495922 on Build GRCh38
rs376741902 Chr.1: 1495828-1495828 on Build GRCh38
rs184136085 Chr.1: 1496034-1496034 on Build GRCh38
rs773865999 Chr.1: 1495771-1495771 on Build GRCh38
rs373416279 Chr.1: 1496003-1496003 on Build GRCh38
rs146476845 Chr.1: 1495931-1495931 on Build GRCh38
rs141015036 Chr.1: 1495794-1495794 on Build GRCh38
rs888527035 Chr.1: 1495893-1495893 on Build GRCh38
rs748171138 Chr.1: 1495692-1495692 on Build GRCh38
rs564244148 Chr.1: 1495883-1495883 on Build GRCh38
rs553799027 Chr.1: 1495777-1495777 on Build GRCh38
rs757866656 Chr.1: 1495856-1495856 on Build GRCh38
rs749626156 Chr.1: 1495869-1495869 on Build GRCh38
rs755916904 Chr.1: 1495736-1495736 on Build GRCh38
rs766788738 Chr.1: 1495722-1495722 on Build GRCh38
rs771638674 Chr.1: 1495763-1495763 on Build GRCh38
rs1030237574 Chr.1: 1496035-1496035 on Build GRCh38
rs556462938 Chr.1: 1495797-1495797 on Build GRCh38
rs978398900 Chr.1: 1495994-1495994 on Build GRCh38
rs535231437 Chr.1: 1496006-1496006 on Build GRCh38
rs200509735 Chr.1: 1495744-1495744 on Build GRCh38
rs918835746 Chr.1: 1495974-1495974 on Build GRCh38
rs775795375 Chr.1: 1495809-1495809 on Build GRCh38
rs781304793 Chr.1: 1495597-1495597 on Build GRCh38
rs140441570 Chr.1: 1495783-1495783 on Build GRCh38
rs535677144 Chr.1: 1495578-1495578 on Build GRCh38
rs533119376 Chr.1: 1495884-1495884 on Build GRCh38
rs766328822 Chr.1: 1495826-1495826 on Build GRCh38
rs984749409 Chr.1: 1495658-1495658 on Build GRCh38
rs761419080 Chr.1: 1495809-1495810 on Build GRCh38
rs1040128103 Chr.1: 1495595-1495595 on Build GRCh38
rs763220315 Chr.1: 1495813-1495813 on Build GRCh38
rs745487182 Chr.1: 1495661-1495661 on Build GRCh38
rs1049001294 Chr.1: 1495991-1495991 on Build GRCh38
rs756495698 Chr.1: 1495674-1495674 on Build GRCh38
rs749965388 Chr.1: 1495863-1495864 on Build GRCh38
rs367589270 Chr.1: 1495629-1495629 on Build GRCh38
rs776634721 Chr.1: 1495701-1495701 on Build GRCh38
rs978319492 Chr.1: 1495918-1495918 on Build GRCh38
rs745628384 Chr.1: 1495660-1495660 on Build GRCh38
rs936315018 Chr.1: 1495733-1495733 on Build GRCh38
rs1030185265 Chr.1: 1495915-1495915 on Build GRCh38
rs749399131 Chr.1: 1495637-1495637 on Build GRCh38
rs766735771 Chr.1: 1495676-1495676 on Build GRCh38
rs146492872 Chr.1: 1495613-1495613 on Build GRCh38
rs747278192 Chr.1: 1495673-1495673 on Build GRCh38
rs527777613 Chr.1: 1495830-1495830 on Build GRCh38
rs151150667 Chr.1: 1495787-1495787 on Build GRCh38
rs150884710 Chr.1: 1495579-1495579 on Build GRCh38
rs150201453 Chr.1: 1495686-1495686 on Build GRCh38
rs777545112 Chr.1: 1495843-1495843 on Build GRCh38
rs376596147 Chr.1: 1496035-1496036 on Build GRCh38
rs952068091 Chr.1: 1495634-1495635 on Build GRCh38
rs762758664 Chr.1: 1495771-1495773 on Build GRCh38
rs547026516 Chr.1: 1496028-1496029 on Build GRCh38
rs746906771 Chr.1: 1495647-1495647 on Build GRCh38
rs949633379 Chr.1: 1495966-1495966 on Build GRCh38
rs570901647 Chr.1: 1495895-1495896 on Build GRCh38
rs577360633 Chr.1: 1495792-1495793 on Build GRCh38
rs758967507 Chr.1: 1495586-1495586 on Build GRCh38
rs776157357 Chr.1: 1495618-1495618 on Build GRCh38
rs752824611 Chr.1: 1495782-1495782 on Build GRCh38
rs773754933 Chr.1: 1496009-1496009 on Build GRCh38
rs770691962 Chr.1: 1495617-1495617 on Build GRCh38
rs369662039 Chr.1: 1495790-1495790 on Build GRCh38
rs916219918 Chr.1: 1495935-1495935 on Build GRCh38
rs775348992 Chr.1: 1495750-1495751 on Build GRCh38
rs749035796 Chr.1: 1495745-1495745 on Build GRCh38
rs201429000 Chr.1: 1495668-1495668 on Build GRCh38
rs758164967 Chr.1: 1495840-1495840 on Build GRCh38

View Full Product Details