Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 1719050-1719565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK11A
Entrez Gene ID
728642
Gene Name
cyclin dependent kinase 11A
Gene Aliases
CDC2L2, CDC2L3, CDK11-p110, CDK11-p46, CDK11-p58, PITSLRE, p58GTA
UniGene
Hs.651228
Gene Chromosome Location
Chr.1: 1702383-1724565 on Build GRCh38
Transcripts
NM_001313896.1 NM_001313982.1 NM_024011.3 NM_033529.3 NR_132739.1 NR_132740.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
898877 c.245C>A p.S82Y Chr.1: 1719438-1719438 on Build GRCh38
4142970 c.277C>T p.R93W Chr.1: 1719406-1719406 on Build GRCh38
5010459 c.325+1G>A p.? Chr.1: 1719357-1719357 on Build GRCh38
385797 c.249G>A p.L83L Chr.1: 1719434-1719434 on Build GRCh38
2088794 c.319G>T p.E107* Chr.1: 1719364-1719364 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs78469202 Chr.1: 1719136-1719136 on Build GRCh38
rs74871563 Chr.1: 1719201-1719201 on Build GRCh38
rs768600257 Chr.1: 1719285-1719285 on Build GRCh38
rs765049205 Chr.1: 1719293-1719293 on Build GRCh38
rs547004452 Chr.1: 1719495-1719495 on Build GRCh38
rs878994186 Chr.1: 1719303-1719303 on Build GRCh38
rs771494265 Chr.1: 1719460-1719460 on Build GRCh38
rs775252031 Chr.1: 1719296-1719296 on Build GRCh38
rs749536579 Chr.1: 1719319-1719319 on Build GRCh38
rs1036669544 Chr.1: 1719334-1719334 on Build GRCh38
rs750911014 Chr.1: 1719353-1719353 on Build GRCh38
rs1534952 Chr.1: 1719189-1719189 on Build GRCh38
rs776580785 Chr.1: 1719290-1719290 on Build GRCh38
rs756337940 Chr.1: 1719278-1719278 on Build GRCh38
rs537114073 Chr.1: 1719262-1719262 on Build GRCh38
rs759940985 Chr.1: 1719419-1719419 on Build GRCh38
rs758644941 Chr.1: 1719426-1719426 on Build GRCh38
rs74763557 Chr.1: 1719180-1719180 on Build GRCh38
rs781271182 Chr.1: 1719329-1719329 on Build GRCh38
rs879147214 Chr.1: 1719115-1719115 on Build GRCh38
rs375417759 Chr.1: 1719108-1719109 on Build GRCh38
rs1137003 Chr.1: 1719348-1719348 on Build GRCh38
rs1137005 Chr.1: 1719368-1719368 on Build GRCh38
rs754215680 Chr.1: 1719413-1719413 on Build GRCh38
rs113532432 Chr.1: 1719502-1719502 on Build GRCh38
rs747883755 Chr.1: 1719494-1719494 on Build GRCh38
rs768006806 Chr.1: 1719314-1719314 on Build GRCh38
rs772149207 Chr.1: 1719357-1719363 on Build GRCh38
rs367689964 Chr.1: 1719121-1719121 on Build GRCh38
rs114056323 Chr.1: 1719530-1719530 on Build GRCh38
rs751745268 Chr.1: 1719484-1719484 on Build GRCh38
rs566998280 Chr.1: 1719173-1719173 on Build GRCh38
rs1015677392 Chr.1: 1719080-1719080 on Build GRCh38
rs878952355 Chr.1: 1719150-1719150 on Build GRCh38
rs567135494 Chr.1: 1719510-1719510 on Build GRCh38
rs752941599 Chr.1: 1719424-1719424 on Build GRCh38
rs898187479 Chr.1: 1719339-1719339 on Build GRCh38
rs113378618 Chr.1: 1719163-1719164 on Build GRCh38
rs754768405 Chr.1: 1719449-1719449 on Build GRCh38
rs758105482 Chr.1: 1719498-1719498 on Build GRCh38
rs79148452 Chr.1: 1719160-1719160 on Build GRCh38
rs765419172 Chr.1: 1719422-1719422 on Build GRCh38
rs746510763 Chr.1: 1719458-1719458 on Build GRCh38
rs369877223 Chr.1: 1719079-1719079 on Build GRCh38
rs78493985 Chr.1: 1719090-1719090 on Build GRCh38
rs36191908 Chr.1: 1719481-1719481 on Build GRCh38
rs200755605 Chr.1: 1719405-1719405 on Build GRCh38
rs749413674 Chr.1: 1719279-1719279 on Build GRCh38
rs751620983 Chr.1: 1719437-1719437 on Build GRCh38
rs75384210 Chr.1: 1719528-1719528 on Build GRCh38
rs78451819 Chr.1: 1719147-1719147 on Build GRCh38
rs79670681 Chr.1: 1719202-1719202 on Build GRCh38
rs879081683 Chr.1: 1719152-1719152 on Build GRCh38
rs913919605 Chr.1: 1719236-1719236 on Build GRCh38
rs111676866 Chr.1: 1719122-1719122 on Build GRCh38
rs201741924 Chr.1: 1719500-1719500 on Build GRCh38
rs769812604 Chr.1: 1719381-1719381 on Build GRCh38
rs187770842 Chr.1: 1719485-1719485 on Build GRCh38
rs747870830 Chr.1: 1719453-1719453 on Build GRCh38
rs200770770 Chr.1: 1719317-1719317 on Build GRCh38
rs368486018 Chr.1: 1719297-1719297 on Build GRCh38
rs779996662 Chr.1: 1719383-1719383 on Build GRCh38
rs780307440 Chr.1: 1719318-1719318 on Build GRCh38
rs557920252 Chr.1: 1719251-1719251 on Build GRCh38
rs946787849 Chr.1: 1719252-1719252 on Build GRCh38
rs72909030 Chr.1: 1719393-1719393 on Build GRCh38
rs748197686 Chr.1: 1719326-1719326 on Build GRCh38
rs759269705 Chr.1: 1719461-1719462 on Build GRCh38
rs770852204 Chr.1: 1719444-1719446 on Build GRCh38
rs772725526 Chr.1: 1719455-1719455 on Build GRCh38
rs547260232 Chr.1: 1719093-1719093 on Build GRCh38
rs72909028 Chr.1: 1719218-1719218 on Build GRCh38
rs748202839 Chr.1: 1719327-1719327 on Build GRCh38
rs771649177 Chr.1: 1719454-1719454 on Build GRCh38
rs148518955 Chr.1: 1719478-1719478 on Build GRCh38
rs754835097 Chr.1: 1719492-1719492 on Build GRCh38
rs78989514 Chr.1: 1719203-1719203 on Build GRCh38
rs762484242 Chr.1: 1719503-1719504 on Build GRCh38
rs112792283 Chr.1: 1719108-1719108 on Build GRCh38
rs751036878 Chr.1: 1719301-1719301 on Build GRCh38
rs759393005 Chr.1: 1719292-1719292 on Build GRCh38
rs768246706 Chr.1: 1719388-1719388 on Build GRCh38
rs75864145 Chr.1: 1719103-1719103 on Build GRCh38
rs776415978 Chr.1: 1719451-1719453 on Build GRCh38
rs779182274 Chr.1: 1719364-1719364 on Build GRCh38
rs746952748 Chr.1: 1719288-1719288 on Build GRCh38
rs768284935 Chr.1: 1719332-1719332 on Build GRCh38
rs1057041762 Chr.1: 1719308-1719308 on Build GRCh38
rs200418535 Chr.1: 1719480-1719480 on Build GRCh38
rs542603674 Chr.1: 1719385-1719385 on Build GRCh38
rs1059830 Chr.1: 1719358-1719358 on Build GRCh38
rs745697129 Chr.1: 1719330-1719330 on Build GRCh38
rs371830741 Chr.1: 1719302-1719302 on Build GRCh38
rs376809595 Chr.1: 1719452-1719452 on Build GRCh38
rs752787794 Chr.1: 1719365-1719365 on Build GRCh38
rs771596383 Chr.1: 1719401-1719401 on Build GRCh38
rs770837020 Chr.1: 1719289-1719289 on Build GRCh38
rs940267853 Chr.1: 1719540-1719540 on Build GRCh38
rs763533645 Chr.1: 1719299-1719299 on Build GRCh38
rs962430541 Chr.1: 1719084-1719084 on Build GRCh38
rs879157520 Chr.1: 1719113-1719113 on Build GRCh38
rs987115752 Chr.1: 1719096-1719096 on Build GRCh38
rs1028111893 Chr.1: 1719373-1719373 on Build GRCh38
rs777165590 Chr.1: 1719477-1719477 on Build GRCh38
rs745820114 Chr.1: 1719372-1719372 on Build GRCh38
rs766815750 Chr.1: 1719411-1719411 on Build GRCh38
rs879026274 Chr.1: 1719128-1719128 on Build GRCh38
rs1137004 Chr.1: 1719362-1719362 on Build GRCh38
rs918092453 Chr.1: 1719535-1719535 on Build GRCh38
rs386627544 Chr.1: 1719500-1719503 on Build GRCh38
rs756131673 Chr.1: 1719445-1719445 on Build GRCh38
rs577827165 Chr.1: 1719261-1719261 on Build GRCh38
rs201091435 Chr.1: 1719446-1719446 on Build GRCh38
rs774444010 Chr.1: 1719286-1719286 on Build GRCh38
rs1059831 Chr.1: 1719406-1719406 on Build GRCh38
rs770832144 Chr.1: 1719328-1719328 on Build GRCh38
rs374031384 Chr.1: 1719094-1719094 on Build GRCh38
rs768075476 Chr.1: 1719349-1719349 on Build GRCh38
rs112502411 Chr.1: 1719129-1719129 on Build GRCh38
rs75972011 Chr.1: 1719503-1719503 on Build GRCh38
rs773744208 Chr.1: 1719345-1719345 on Build GRCh38
rs755190607 Chr.1: 1719320-1719320 on Build GRCh38
rs778865504 Chr.1: 1719326-1719326 on Build GRCh38
rs749100082 Chr.1: 1719448-1719448 on Build GRCh38
rs72634834 Chr.1: 1719521-1719521 on Build GRCh38
rs747027760 Chr.1: 1719437-1719438 on Build GRCh38
rs777681332 Chr.1: 1719387-1719389 on Build GRCh38
rs775171326 Chr.1: 1719500-1719502 on Build GRCh38
rs201838992 Chr.1: 1719397-1719397 on Build GRCh38
rs112957564 Chr.1: 1719120-1719120 on Build GRCh38
rs766762922 Chr.1: 1719357-1719357 on Build GRCh38
rs201674700 Chr.1: 1719430-1719430 on Build GRCh38
rs988138001 Chr.1: 1719206-1719206 on Build GRCh38
rs779218469 Chr.1: 1719304-1719305 on Build GRCh38
rs769463634 Chr.1: 1719500-1719501 on Build GRCh38
rs112012562 Chr.1: 1719188-1719188 on Build GRCh38
rs74345479 Chr.1: 1719501-1719501 on Build GRCh38
rs775374870 Chr.1: 1719331-1719331 on Build GRCh38
rs376109033 Chr.1: 1719106-1719106 on Build GRCh38
rs201592416 Chr.1: 1719361-1719361 on Build GRCh38

View Full Product Details